Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45730915-45731405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
UniGene
Hs.101307
Gene Chromosome Location
Chr.18: 45724123-45752520 on Build GRCh38
Transcripts
NM_001128588.3 NM_001146036.2 NM_001146037.1 NM_001308278.1 NM_001308279.1 NM_015865.6 XM_005258329.1 XM_005258333.1 XM_006722526.3 XM_011526142.1 XM_011526144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397040 c.161T>C p.V54A Chr.18: 45731024-45731024 on Build GRCh38
6395235 c.185T>C p.I62T Chr.18: 45731048-45731048 on Build GRCh38
4965574 c.221A>G p.N74S Chr.18: 45731084-45731084 on Build GRCh38
1388750 c.421C>G p.L141V Chr.18: 45731116-45731116 on Build GRCh38
336087 c.291G>T p.W97C Chr.18: 45731154-45731154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928072537 Chr.18: 45731358-45731358 on Build GRCh38
rs559781749 Chr.18: 45731331-45731331 on Build GRCh38
rs745580527 Chr.18: 45731076-45731076 on Build GRCh38
rs562986926 Chr.18: 45731132-45731132 on Build GRCh38
rs113029149 Chr.18: 45731089-45731089 on Build GRCh38
rs763262711 Chr.18: 45731139-45731139 on Build GRCh38
rs777399384 Chr.18: 45730985-45730985 on Build GRCh38
rs752058156 Chr.18: 45731157-45731157 on Build GRCh38
rs754106291 Chr.18: 45731031-45731031 on Build GRCh38
rs962632371 Chr.18: 45731281-45731281 on Build GRCh38
rs987640812 Chr.18: 45731338-45731338 on Build GRCh38
rs758962746 Chr.18: 45731238-45731238 on Build GRCh38
rs749061328 Chr.18: 45731216-45731216 on Build GRCh38
rs189771012 Chr.18: 45731289-45731289 on Build GRCh38
rs751044851 Chr.18: 45731063-45731063 on Build GRCh38
rs781027243 Chr.18: 45730999-45730999 on Build GRCh38
rs368871785 Chr.18: 45731055-45731055 on Build GRCh38
rs28994287 Chr.18: 45731166-45731166 on Build GRCh38
rs973464654 Chr.18: 45730971-45730971 on Build GRCh38
rs543668941 Chr.18: 45731211-45731211 on Build GRCh38
rs1037701991 Chr.18: 45731062-45731062 on Build GRCh38
rs541182989 Chr.18: 45731310-45731310 on Build GRCh38
rs142529927 Chr.18: 45731065-45731065 on Build GRCh38
rs749964265 Chr.18: 45731196-45731196 on Build GRCh38
rs184913653 Chr.18: 45731071-45731071 on Build GRCh38
rs373450411 Chr.18: 45730955-45730962 on Build GRCh38
rs945178802 Chr.18: 45731020-45731020 on Build GRCh38
rs771038208 Chr.18: 45731127-45731127 on Build GRCh38
rs768487613 Chr.18: 45731021-45731021 on Build GRCh38
rs147790844 Chr.18: 45731131-45731131 on Build GRCh38
rs750167058 Chr.18: 45731020-45731029 on Build GRCh38
rs373010473 Chr.18: 45730996-45730996 on Build GRCh38
rs377124382 Chr.18: 45731023-45731023 on Build GRCh38
rs778298286 Chr.18: 45731376-45731376 on Build GRCh38
rs779059222 Chr.18: 45730951-45730951 on Build GRCh38
rs1033567236 Chr.18: 45730991-45730991 on Build GRCh38
rs745611570 Chr.18: 45731007-45731007 on Build GRCh38
rs139053682 Chr.18: 45731056-45731056 on Build GRCh38
rs924804539 Chr.18: 45731038-45731038 on Build GRCh38
rs749037771 Chr.18: 45731085-45731085 on Build GRCh38
rs140257106 Chr.18: 45731088-45731088 on Build GRCh38
rs534964434 Chr.18: 45731276-45731276 on Build GRCh38
rs748149855 Chr.18: 45731232-45731232 on Build GRCh38
rs977885321 Chr.18: 45731266-45731266 on Build GRCh38
rs200895613 Chr.18: 45731011-45731011 on Build GRCh38
rs764618306 Chr.18: 45731147-45731147 on Build GRCh38
rs775561079 Chr.18: 45731013-45731013 on Build GRCh38
rs959261830 Chr.18: 45730992-45730992 on Build GRCh38
rs767190652 Chr.18: 45731182-45731182 on Build GRCh38
rs770021358 Chr.18: 45731253-45731253 on Build GRCh38
rs753289147 Chr.18: 45730967-45730967 on Build GRCh38
rs146420195 Chr.18: 45731044-45731044 on Build GRCh38
rs770645099 Chr.18: 45730995-45730995 on Build GRCh38
rs910713900 Chr.18: 45731369-45731369 on Build GRCh38
rs959314162 Chr.18: 45731176-45731176 on Build GRCh38
rs955135936 Chr.18: 45731334-45731334 on Build GRCh38
rs776636321 Chr.18: 45731246-45731246 on Build GRCh38
rs766335775 Chr.18: 45731130-45731130 on Build GRCh38
rs552716005 Chr.18: 45731130-45731130 on Build GRCh38
rs202168702 Chr.18: 45731022-45731022 on Build GRCh38
rs141409831 Chr.18: 45731252-45731252 on Build GRCh38
rs373247991 Chr.18: 45731014-45731014 on Build GRCh38
rs989167938 Chr.18: 45730994-45730994 on Build GRCh38
rs755756472 Chr.18: 45731209-45731209 on Build GRCh38
rs778150490 Chr.18: 45731057-45731057 on Build GRCh38
rs760401643 Chr.18: 45731125-45731126 on Build GRCh38
rs552191196 Chr.18: 45731053-45731053 on Build GRCh38
rs753795913 Chr.18: 45731205-45731206 on Build GRCh38
rs754777890 Chr.18: 45731217-45731217 on Build GRCh38
rs374417360 Chr.18: 45731224-45731224 on Build GRCh38
rs770987026 Chr.18: 45731244-45731244 on Build GRCh38
rs920624993 Chr.18: 45731333-45731333 on Build GRCh38
rs775858393 Chr.18: 45731135-45731135 on Build GRCh38
rs757844601 Chr.18: 45730977-45730977 on Build GRCh38
rs750993870 Chr.18: 45731213-45731213 on Build GRCh38
rs757040554 Chr.18: 45731143-45731143 on Build GRCh38
rs779835840 Chr.18: 45731082-45731082 on Build GRCh38
rs766539715 Chr.18: 45731028-45731028 on Build GRCh38
rs114362217 Chr.18: 45731054-45731054 on Build GRCh38
rs746515374 Chr.18: 45730989-45730989 on Build GRCh38
rs980888242 Chr.18: 45731357-45731357 on Build GRCh38
rs147563846 Chr.18: 45731077-45731077 on Build GRCh38
rs201716471 Chr.18: 45731050-45731050 on Build GRCh38
rs760784196 Chr.18: 45731101-45731101 on Build GRCh38
rs988336714 Chr.18: 45731378-45731378 on Build GRCh38
rs966737411 Chr.18: 45731250-45731250 on Build GRCh38

View Full Product Details