Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46084368-46084877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5A1
Entrez Gene ID
498
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
Gene Aliases
ATP5A, ATP5AL2, ATPM, COXPD22, HEL-S-123m, MC5DN4, MOM2, OMR, ORM, hATP1
UniGene
Hs.298280
Gene Chromosome Location
Chr.18: 46084144-46104233 on Build GRCh38
Transcripts
NM_001001935.2 NM_001001937.1 NM_001257334.1 NM_001257335.1 NM_004046.5 XM_017025789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709483 c.1556A>T p.H519L Chr.18: 46084528-46084528 on Build GRCh38
7082990 c.1546G>C p.V516L Chr.18: 46084538-46084538 on Build GRCh38
4627595 c.1520T>C p.F507S Chr.18: 46084564-46084564 on Build GRCh38
4072305 c.1483T>C p.Y495H Chr.18: 46084601-46084601 on Build GRCh38
6444948 c.1452A>G p.Q484Q Chr.18: 46084632-46084632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73953979 Chr.18: 46084481-46084481 on Build GRCh38
rs898825940 Chr.18: 46084586-46084586 on Build GRCh38
rs542107636 Chr.18: 46084497-46084497 on Build GRCh38
rs370149807 Chr.18: 46084826-46084827 on Build GRCh38
rs754895860 Chr.18: 46084573-46084573 on Build GRCh38
rs563441821 Chr.18: 46084518-46084518 on Build GRCh38
rs746180623 Chr.18: 46084511-46084511 on Build GRCh38
rs772672687 Chr.18: 46084706-46084706 on Build GRCh38
rs368878416 Chr.18: 46084396-46084396 on Build GRCh38
rs557384276 Chr.18: 46084681-46084682 on Build GRCh38
rs139399471 Chr.18: 46084650-46084650 on Build GRCh38
rs147667560 Chr.18: 46084836-46084837 on Build GRCh38
rs757503918 Chr.18: 46084612-46084612 on Build GRCh38
rs200501560 Chr.18: 46084754-46084754 on Build GRCh38
rs1042724443 Chr.18: 46084823-46084823 on Build GRCh38
rs17856350 Chr.18: 46084555-46084555 on Build GRCh38
rs749872023 Chr.18: 46084664-46084664 on Build GRCh38
rs760326198 Chr.18: 46084538-46084538 on Build GRCh38
rs756369878 Chr.18: 46084620-46084620 on Build GRCh38
rs370098146 Chr.18: 46084492-46084492 on Build GRCh38
rs771766792 Chr.18: 46084682-46084682 on Build GRCh38
rs781224378 Chr.18: 46084614-46084614 on Build GRCh38
rs753837503 Chr.18: 46084572-46084572 on Build GRCh38
rs761593652 Chr.18: 46084528-46084528 on Build GRCh38
rs947498321 Chr.18: 46084397-46084397 on Build GRCh38
rs750150923 Chr.18: 46084461-46084461 on Build GRCh38
rs774959235 Chr.18: 46084753-46084753 on Build GRCh38
rs780466269 Chr.18: 46084624-46084624 on Build GRCh38
rs564184761 Chr.18: 46084761-46084761 on Build GRCh38
rs35738466 Chr.18: 46084833-46084833 on Build GRCh38
rs773079671 Chr.18: 46084536-46084536 on Build GRCh38
rs746260834 Chr.18: 46084615-46084615 on Build GRCh38
rs1051912421 Chr.18: 46084489-46084489 on Build GRCh38
rs771883737 Chr.18: 46084602-46084602 on Build GRCh38
rs948364506 Chr.18: 46084805-46084805 on Build GRCh38
rs192780099 Chr.18: 46084803-46084803 on Build GRCh38
rs748813517 Chr.18: 46084517-46084517 on Build GRCh38
rs75974428 Chr.18: 46084640-46084640 on Build GRCh38
rs773076596 Chr.18: 46084658-46084658 on Build GRCh38
rs1039110798 Chr.18: 46084455-46084455 on Build GRCh38
rs992949155 Chr.18: 46084798-46084798 on Build GRCh38
rs779803625 Chr.18: 46084474-46084474 on Build GRCh38
rs747609570 Chr.18: 46084675-46084675 on Build GRCh38
rs757450512 Chr.18: 46084495-46084495 on Build GRCh38
rs372574926 Chr.18: 46084466-46084466 on Build GRCh38
rs775805857 Chr.18: 46084513-46084513 on Build GRCh38
rs751194998 Chr.18: 46084411-46084411 on Build GRCh38
rs769468595 Chr.18: 46084665-46084665 on Build GRCh38
rs766227387 Chr.18: 46084835-46084836 on Build GRCh38
rs774025106 Chr.18: 46084666-46084666 on Build GRCh38
rs533341367 Chr.18: 46084483-46084483 on Build GRCh38
rs767033836 Chr.18: 46084531-46084531 on Build GRCh38
rs33919884 Chr.18: 46084837-46084838 on Build GRCh38
rs546302348 Chr.18: 46084806-46084806 on Build GRCh38
rs552943937 Chr.18: 46084486-46084488 on Build GRCh38
rs751608860 Chr.18: 46084610-46084610 on Build GRCh38
rs975110889 Chr.18: 46084846-46084846 on Build GRCh38
rs764206728 Chr.18: 46084582-46084582 on Build GRCh38
rs981927384 Chr.18: 46084655-46084655 on Build GRCh38
rs770356759 Chr.18: 46084512-46084512 on Build GRCh38
rs541416190 Chr.18: 46084459-46084459 on Build GRCh38
rs947419044 Chr.18: 46084843-46084844 on Build GRCh38
rs948393890 Chr.18: 46084451-46084451 on Build GRCh38
rs904128816 Chr.18: 46084480-46084480 on Build GRCh38
rs894170493 Chr.18: 46084423-46084423 on Build GRCh38
rs763855531 Chr.18: 46084408-46084408 on Build GRCh38
rs777825853 Chr.18: 46084488-46084488 on Build GRCh38
rs34463537 Chr.18: 46084837-46084837 on Build GRCh38
rs760562511 Chr.18: 46084667-46084669 on Build GRCh38
rs373170033 Chr.18: 46084456-46084456 on Build GRCh38
rs959812953 Chr.18: 46084766-46084766 on Build GRCh38
rs748865084 Chr.18: 46084782-46084782 on Build GRCh38

View Full Product Details