Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46094882-46095347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5A1
Entrez Gene ID
498
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
Gene Aliases
ATP5A, ATP5AL2, ATPM, COXPD22, HEL-S-123m, MC5DN4, MOM2, OMR, ORM, hATP1
UniGene
Hs.298280
Gene Chromosome Location
Chr.18: 46084144-46104233 on Build GRCh38
Transcripts
NM_001001935.2 NM_001001937.1 NM_001257334.1 NM_001257335.1 NM_004046.5 XM_017025789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607927 c.114C>A p.A38A Chr.18: 46095078-46095078 on Build GRCh38
6607923 c.84A>G p.S28S Chr.18: 46095108-46095108 on Build GRCh38
6303758 c.139+5G>T p.? Chr.18: 46095048-46095048 on Build GRCh38
6434083 c.97G>A p.A33T Chr.18: 46095095-46095095 on Build GRCh38
274093 c.117T>G p.S39S Chr.18: 46095075-46095075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550057616 Chr.18: 46095116-46095116 on Build GRCh38
rs752960534 Chr.18: 46095037-46095037 on Build GRCh38
rs771421663 Chr.18: 46095010-46095010 on Build GRCh38
rs780882068 Chr.18: 46095020-46095020 on Build GRCh38
rs755201202 Chr.18: 46094990-46094990 on Build GRCh38
rs376015700 Chr.18: 46095027-46095027 on Build GRCh38
rs1047256299 Chr.18: 46095228-46095228 on Build GRCh38
rs778123236 Chr.18: 46094948-46094948 on Build GRCh38
rs746697869 Chr.18: 46095018-46095019 on Build GRCh38
rs886494904 Chr.18: 46094965-46094965 on Build GRCh38
rs747519031 Chr.18: 46095006-46095006 on Build GRCh38
rs756663379 Chr.18: 46095148-46095148 on Build GRCh38
rs746364441 Chr.18: 46095065-46095065 on Build GRCh38
rs775590957 Chr.18: 46095109-46095109 on Build GRCh38
rs1017198112 Chr.18: 46095252-46095252 on Build GRCh38
rs773544077 Chr.18: 46094941-46094941 on Build GRCh38
rs763118169 Chr.18: 46095112-46095112 on Build GRCh38
rs371613044 Chr.18: 46095146-46095146 on Build GRCh38
rs752907367 Chr.18: 46094993-46094993 on Build GRCh38
rs779051427 Chr.18: 46094991-46094991 on Build GRCh38
rs184693047 Chr.18: 46095063-46095063 on Build GRCh38
rs991531091 Chr.18: 46094945-46094945 on Build GRCh38
rs1056940055 Chr.18: 46095307-46095307 on Build GRCh38
rs1049183975 Chr.18: 46095242-46095242 on Build GRCh38
rs758624976 Chr.18: 46095038-46095038 on Build GRCh38
rs1023972466 Chr.18: 46095258-46095258 on Build GRCh38
rs971170105 Chr.18: 46095267-46095267 on Build GRCh38
rs548271044 Chr.18: 46095060-46095060 on Build GRCh38
rs772973691 Chr.18: 46095179-46095179 on Build GRCh38
rs957245054 Chr.18: 46095314-46095314 on Build GRCh38
rs754587827 Chr.18: 46095157-46095157 on Build GRCh38
rs776507828 Chr.18: 46095175-46095175 on Build GRCh38
rs372098285 Chr.18: 46095026-46095026 on Build GRCh38
rs746972181 Chr.18: 46095183-46095183 on Build GRCh38
rs559516147 Chr.18: 46094988-46094988 on Build GRCh38
rs369713479 Chr.18: 46095025-46095025 on Build GRCh38
rs571634598 Chr.18: 46095160-46095160 on Build GRCh38
rs942731767 Chr.18: 46095137-46095137 on Build GRCh38
rs745308865 Chr.18: 46095024-46095024 on Build GRCh38
rs200765347 Chr.18: 46095028-46095028 on Build GRCh38
rs977177112 Chr.18: 46095291-46095291 on Build GRCh38
rs377317993 Chr.18: 46095007-46095007 on Build GRCh38
rs779675872 Chr.18: 46095076-46095076 on Build GRCh38
rs894704067 Chr.18: 46095253-46095253 on Build GRCh38
rs746509051 Chr.18: 46095113-46095113 on Build GRCh38
rs367903141 Chr.18: 46095102-46095102 on Build GRCh38
rs775438038 Chr.18: 46095192-46095192 on Build GRCh38
rs748789373 Chr.18: 46095156-46095156 on Build GRCh38
rs766491097 Chr.18: 46095029-46095029 on Build GRCh38
rs901290300 Chr.18: 46095202-46095202 on Build GRCh38
rs760578081 Chr.18: 46095104-46095104 on Build GRCh38
rs767989967 Chr.18: 46095136-46095136 on Build GRCh38
rs761614852 Chr.18: 46095092-46095092 on Build GRCh38
rs1005297381 Chr.18: 46094981-46094981 on Build GRCh38
rs771898815 Chr.18: 46095172-46095172 on Build GRCh38
rs189208584 Chr.18: 46095080-46095080 on Build GRCh38
rs147760997 Chr.18: 46095286-46095286 on Build GRCh38
rs940822684 Chr.18: 46095298-46095298 on Build GRCh38
rs757707814 Chr.18: 46095050-46095050 on Build GRCh38
rs908034570 Chr.18: 46095278-46095278 on Build GRCh38
rs777561696 Chr.18: 46094972-46094972 on Build GRCh38
rs778688373 Chr.18: 46095159-46095159 on Build GRCh38
rs770817462 Chr.18: 46095186-46095186 on Build GRCh38
rs990085782 Chr.18: 46095087-46095087 on Build GRCh38
rs896472960 Chr.18: 46095013-46095013 on Build GRCh38
rs1012605538 Chr.18: 46095256-46095257 on Build GRCh38
rs751913949 Chr.18: 46095049-46095049 on Build GRCh38
rs778169278 Chr.18: 46094997-46094997 on Build GRCh38
rs765301106 Chr.18: 46095034-46095034 on Build GRCh38
rs759568376 Chr.18: 46095251-46095251 on Build GRCh38
rs28699609 Chr.18: 46094913-46094913 on Build GRCh38
rs758760294 Chr.18: 46094996-46094996 on Build GRCh38
rs372392795 Chr.18: 46095070-46095070 on Build GRCh38
rs79011243 Chr.18: 46095098-46095098 on Build GRCh38
rs765350359 Chr.18: 46095107-46095107 on Build GRCh38
rs138315345 Chr.18: 46095078-46095078 on Build GRCh38
rs754259358 Chr.18: 46095195-46095198 on Build GRCh38
rs934076078 Chr.18: 46095215-46095215 on Build GRCh38
rs770827087 Chr.18: 46095151-46095151 on Build GRCh38
rs778023531 Chr.18: 46095045-46095045 on Build GRCh38
rs779728877 Chr.18: 46095149-46095149 on Build GRCh38
rs1049151072 Chr.18: 46094961-46094961 on Build GRCh38

View Full Product Details