Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63350746-63351289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692795 c.447C>T p.S149S Chr.18: 63351050-63351050 on Build GRCh38
4433830 c.543C>T p.F181F Chr.18: 63350954-63350954 on Build GRCh38
5694708 c.566C>T p.S189F Chr.18: 63350931-63350931 on Build GRCh38
4756251 c.449T>C p.V150A Chr.18: 63351048-63351048 on Build GRCh38
5564641 c.490C>T p.R164W Chr.18: 63351007-63351007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974606580 Chr.18: 63351259-63351259 on Build GRCh38
rs767427017 Chr.18: 63350865-63350865 on Build GRCh38
rs1000408845 Chr.18: 63351144-63351144 on Build GRCh38
rs533621939 Chr.18: 63350797-63350797 on Build GRCh38
rs996582288 Chr.18: 63351090-63351090 on Build GRCh38
rs747738448 Chr.18: 63351131-63351131 on Build GRCh38
rs781725078 Chr.18: 63350842-63350842 on Build GRCh38
rs190777855 Chr.18: 63351161-63351161 on Build GRCh38
rs764212085 Chr.18: 63351050-63351050 on Build GRCh38
rs924888281 Chr.18: 63350807-63350807 on Build GRCh38
rs558247035 Chr.18: 63351127-63351127 on Build GRCh38
rs770741711 Chr.18: 63351037-63351037 on Build GRCh38
rs78463512 Chr.18: 63351073-63351073 on Build GRCh38
rs754725800 Chr.18: 63351125-63351125 on Build GRCh38
rs766101689 Chr.18: 63350906-63350906 on Build GRCh38
rs969853309 Chr.18: 63351133-63351133 on Build GRCh38
rs535843222 Chr.18: 63351012-63351012 on Build GRCh38
rs563478891 Chr.18: 63351071-63351071 on Build GRCh38
rs773143504 Chr.18: 63351032-63351032 on Build GRCh38
rs764853231 Chr.18: 63351024-63351024 on Build GRCh38
rs76278802 Chr.18: 63350850-63350850 on Build GRCh38
rs747651555 Chr.18: 63351025-63351025 on Build GRCh38
rs978852768 Chr.18: 63350787-63350791 on Build GRCh38
rs773702860 Chr.18: 63350857-63350857 on Build GRCh38
rs1038547029 Chr.18: 63351031-63351031 on Build GRCh38
rs998482085 Chr.18: 63351170-63351170 on Build GRCh38
rs988920250 Chr.18: 63351153-63351153 on Build GRCh38
rs895522355 Chr.18: 63351087-63351087 on Build GRCh38
rs761989959 Chr.18: 63351057-63351057 on Build GRCh38
rs763288681 Chr.18: 63351049-63351049 on Build GRCh38
rs758114739 Chr.18: 63351058-63351059 on Build GRCh38
rs1032864613 Chr.18: 63351145-63351145 on Build GRCh38
rs939246657 Chr.18: 63350881-63350881 on Build GRCh38
rs911447766 Chr.18: 63351250-63351250 on Build GRCh38
rs531189319 Chr.18: 63350851-63350851 on Build GRCh38
rs199560861 Chr.18: 63350840-63350840 on Build GRCh38
rs750894162 Chr.18: 63350976-63350976 on Build GRCh38
rs558689966 Chr.18: 63351209-63351209 on Build GRCh38
rs778537413 Chr.18: 63350965-63350965 on Build GRCh38
rs941638527 Chr.18: 63351253-63351253 on Build GRCh38
rs776256009 Chr.18: 63351044-63351044 on Build GRCh38
rs990344604 Chr.18: 63350832-63350832 on Build GRCh38
rs550316154 Chr.18: 63350853-63350853 on Build GRCh38
rs372546740 Chr.18: 63351122-63351122 on Build GRCh38
rs546316503 Chr.18: 63351027-63351027 on Build GRCh38
rs1050044561 Chr.18: 63350981-63350981 on Build GRCh38
rs956279685 Chr.18: 63351148-63351148 on Build GRCh38
rs752982452 Chr.18: 63350920-63350920 on Build GRCh38
rs752444792 Chr.18: 63351000-63351000 on Build GRCh38
rs781764372 Chr.18: 63350961-63350961 on Build GRCh38
rs760553387 Chr.18: 63351033-63351033 on Build GRCh38
rs773055901 Chr.18: 63350878-63350878 on Build GRCh38
rs955382558 Chr.18: 63351220-63351220 on Build GRCh38
rs538101608 Chr.18: 63351265-63351265 on Build GRCh38
rs567294982 Chr.18: 63350947-63350947 on Build GRCh38
rs543545590 Chr.18: 63351006-63351006 on Build GRCh38
rs1063341 Chr.18: 63351041-63351041 on Build GRCh38
rs751732191 Chr.18: 63351053-63351053 on Build GRCh38
rs756650943 Chr.18: 63351074-63351074 on Build GRCh38
rs751571336 Chr.18: 63350953-63350953 on Build GRCh38
rs769274445 Chr.18: 63351007-63351007 on Build GRCh38
rs985696103 Chr.18: 63351234-63351234 on Build GRCh38
rs957734504 Chr.18: 63350808-63350808 on Build GRCh38
rs201388139 Chr.18: 63350864-63350864 on Build GRCh38
rs748905300 Chr.18: 63350852-63350852 on Build GRCh38
rs768170793 Chr.18: 63351010-63351010 on Build GRCh38
rs746885045 Chr.18: 63351110-63351110 on Build GRCh38
rs955083359 Chr.18: 63351141-63351141 on Build GRCh38
rs181653542 Chr.18: 63351260-63351260 on Build GRCh38
rs201014559 Chr.18: 63350985-63350985 on Build GRCh38
rs777615499 Chr.18: 63351059-63351060 on Build GRCh38
rs1032309527 Chr.18: 63351221-63351221 on Build GRCh38
rs757901419 Chr.18: 63350954-63350954 on Build GRCh38
rs370831192 Chr.18: 63350839-63350839 on Build GRCh38
rs760465413 Chr.18: 63350893-63350893 on Build GRCh38
rs767648186 Chr.18: 63351067-63351067 on Build GRCh38
rs780515282 Chr.18: 63351102-63351102 on Build GRCh38
rs1038226452 Chr.18: 63351036-63351036 on Build GRCh38
rs763042491 Chr.18: 63350933-63350933 on Build GRCh38

View Full Product Details