Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17075420-17075916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS8
Entrez Gene ID
93323
Gene Name
HAUS augmin like complex subunit 8
Gene Aliases
DGT4, HICE1, NY-SAR-48
UniGene
Hs.404088
Gene Chromosome Location
Chr.19: 17049761-17075533 on Build GRCh38
Transcripts
NM_001011699.1 NM_033417.1 XM_005260154.4 NM_001130065.1 NM_004145.3

Gene Symbol
MYO9B
Entrez Gene ID
4650
Gene Name
myosin IXB
Gene Aliases
CELIAC4, MYR5
UniGene
Hs.123198
Gene Chromosome Location
Chr.19: 17075781-17213295 on Build GRCh38
Transcripts
NM_001011699.1 NM_033417.1 XM_005260154.4 NM_001130065.1 NM_004145.3

SNP Detail

SNP ID Chromosome Location
rs1023163367 Chr.19: 17075483-17075483 on Build GRCh38
rs918200969 Chr.19: 17075705-17075705 on Build GRCh38
rs769799871 Chr.19: 17075473-17075473 on Build GRCh38
rs1056468307 Chr.19: 17075704-17075704 on Build GRCh38
rs1010935063 Chr.19: 17075545-17075545 on Build GRCh38
rs1047326157 Chr.19: 17075726-17075726 on Build GRCh38
rs898945136 Chr.19: 17075515-17075515 on Build GRCh38
rs532510936 Chr.19: 17075572-17075572 on Build GRCh38
rs1052205046 Chr.19: 17075673-17075673 on Build GRCh38
rs898636190 Chr.19: 17075715-17075715 on Build GRCh38
rs935586489 Chr.19: 17075609-17075609 on Build GRCh38
rs113610362 Chr.19: 17075710-17075710 on Build GRCh38
rs558055187 Chr.19: 17075784-17075785 on Build GRCh38
rs764120181 Chr.19: 17075466-17075466 on Build GRCh38
rs780531251 Chr.19: 17075458-17075480 on Build GRCh38
rs745397420 Chr.19: 17075464-17075464 on Build GRCh38
rs1012382920 Chr.19: 17075516-17075516 on Build GRCh38
rs996074994 Chr.19: 17075783-17075783 on Build GRCh38
rs572551678 Chr.19: 17075489-17075489 on Build GRCh38
rs966360324 Chr.19: 17075779-17075779 on Build GRCh38
rs753259177 Chr.19: 17075456-17075456 on Build GRCh38
rs1012861150 Chr.19: 17075628-17075628 on Build GRCh38
rs979246154 Chr.19: 17075796-17075796 on Build GRCh38
rs1043749789 Chr.19: 17075593-17075593 on Build GRCh38
rs772894478 Chr.19: 17075443-17075443 on Build GRCh38
rs905610589 Chr.19: 17075631-17075631 on Build GRCh38
rs955558978 Chr.19: 17075585-17075585 on Build GRCh38
rs200756286 Chr.19: 17075448-17075448 on Build GRCh38
rs527527011 Chr.19: 17075877-17075877 on Build GRCh38
rs548702103 Chr.19: 17075563-17075563 on Build GRCh38
rs920233267 Chr.19: 17075624-17075624 on Build GRCh38
rs950365196 Chr.19: 17075667-17075667 on Build GRCh38
rs765726889 Chr.19: 17075455-17075455 on Build GRCh38
rs896632358 Chr.19: 17075470-17075470 on Build GRCh38
rs982946174 Chr.19: 17075587-17075587 on Build GRCh38
rs886178784 Chr.19: 17075762-17075762 on Build GRCh38
rs764750452 Chr.19: 17075607-17075607 on Build GRCh38
rs530399877 Chr.19: 17075541-17075541 on Build GRCh38
rs956770651 Chr.19: 17075873-17075873 on Build GRCh38
rs922773249 Chr.19: 17075578-17075578 on Build GRCh38
rs749253261 Chr.19: 17075710-17075711 on Build GRCh38
rs969823442 Chr.19: 17075490-17075490 on Build GRCh38
rs921748383 Chr.19: 17075684-17075684 on Build GRCh38
rs973515696 Chr.19: 17075494-17075494 on Build GRCh38
rs908787255 Chr.19: 17075481-17075481 on Build GRCh38
rs528919813 Chr.19: 17075554-17075554 on Build GRCh38
rs1044896756 Chr.19: 17075528-17075528 on Build GRCh38
rs543863605 Chr.19: 17075547-17075547 on Build GRCh38
rs1048822230 Chr.19: 17075751-17075751 on Build GRCh38
rs757439431 Chr.19: 17075469-17075469 on Build GRCh38
rs973107457 Chr.19: 17075612-17075612 on Build GRCh38
rs555441209 Chr.19: 17075767-17075768 on Build GRCh38
rs766718576 Chr.19: 17075451-17075451 on Build GRCh38
rs935347653 Chr.19: 17075696-17075696 on Build GRCh38
rs975022309 Chr.19: 17075676-17075676 on Build GRCh38
rs1002932936 Chr.19: 17075786-17075786 on Build GRCh38
rs567251000 Chr.19: 17075894-17075894 on Build GRCh38
rs776874404 Chr.19: 17075452-17075452 on Build GRCh38
rs948289154 Chr.19: 17075709-17075709 on Build GRCh38
rs79900220 Chr.19: 17075560-17075560 on Build GRCh38
rs947834867 Chr.19: 17075625-17075625 on Build GRCh38
rs566040791 Chr.19: 17075580-17075580 on Build GRCh38
rs868574590 Chr.19: 17075722-17075722 on Build GRCh38
rs143850959 Chr.19: 17075535-17075535 on Build GRCh38
rs924055575 Chr.19: 17075801-17075801 on Build GRCh38
rs759971105 Chr.19: 17075453-17075453 on Build GRCh38
rs939209255 Chr.19: 17075664-17075664 on Build GRCh38
rs1004489289 Chr.19: 17075670-17075671 on Build GRCh38
rs541640564 Chr.19: 17075533-17075533 on Build GRCh38
rs907307400 Chr.19: 17075776-17075776 on Build GRCh38
rs547173873 Chr.19: 17075883-17075883 on Build GRCh38
rs1039509002 Chr.19: 17075444-17075444 on Build GRCh38
rs374138402 Chr.19: 17075442-17075442 on Build GRCh38
rs758449349 Chr.19: 17075463-17075463 on Build GRCh38
rs755967441 Chr.19: 17075617-17075617 on Build GRCh38
rs552596210 Chr.19: 17075579-17075579 on Build GRCh38
rs112344057 Chr.19: 17075670-17075670 on Build GRCh38
rs375959530 Chr.19: 17075589-17075589 on Build GRCh38
rs368359670 Chr.19: 17075467-17075467 on Build GRCh38
rs528019949 Chr.19: 17075680-17075681 on Build GRCh38
rs756052754 Chr.19: 17075687-17075687 on Build GRCh38
rs1037756861 Chr.19: 17075504-17075504 on Build GRCh38
rs906287444 Chr.19: 17075630-17075630 on Build GRCh38
rs1044761537 Chr.19: 17075629-17075629 on Build GRCh38
rs968549736 Chr.19: 17075686-17075686 on Build GRCh38
rs994510898 Chr.19: 17075637-17075637 on Build GRCh38
rs967929754 Chr.19: 17075788-17075788 on Build GRCh38
rs1030913657 Chr.19: 17075539-17075539 on Build GRCh38

View Full Product Details