Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44623622-44624128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF23
Entrez Gene ID
147710
Gene Name
immunoglobulin superfamily member 23
Gene Aliases
-
UniGene
Hs.512509
Gene Chromosome Location
Chr.19: 44612577-44636781 on Build GRCh38
Transcripts
NM_001205280.1 XM_017026317.1 XM_017026318.1 XM_017026319.1 XM_017026320.1 XM_017026321.1 XM_017026322.1 XM_017026323.1 XM_017026324.1 XM_017026325.1 XM_017026326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630979 c.153C>G p.F51L Chr.19: 44623734-44623734 on Build GRCh38
5343343 c.391+5G>A p.? Chr.19: 44623977-44623977 on Build GRCh38
6501957 c.167G>A p.R56Q Chr.19: 44623748-44623748 on Build GRCh38
6257564 c.391+3G>T p.? Chr.19: 44623975-44623975 on Build GRCh38
6223062 c.175A>G p.I59V Chr.19: 44623756-44623756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748489754 Chr.19: 44623713-44623713 on Build GRCh38
rs921836525 Chr.19: 44624095-44624095 on Build GRCh38
rs551193574 Chr.19: 44624082-44624082 on Build GRCh38
rs762744162 Chr.19: 44623732-44623732 on Build GRCh38
rs878919741 Chr.19: 44623982-44623982 on Build GRCh38
rs1004277872 Chr.19: 44624008-44624008 on Build GRCh38
rs745915226 Chr.19: 44623877-44623877 on Build GRCh38
rs953871371 Chr.19: 44623801-44623801 on Build GRCh38
rs568756159 Chr.19: 44623995-44623995 on Build GRCh38
rs977839532 Chr.19: 44623966-44623966 on Build GRCh38
rs760301623 Chr.19: 44623993-44623993 on Build GRCh38
rs531260204 Chr.19: 44624021-44624021 on Build GRCh38
rs888528229 Chr.19: 44623998-44623998 on Build GRCh38
rs1057356379 Chr.19: 44623894-44623894 on Build GRCh38
rs939017280 Chr.19: 44623893-44623893 on Build GRCh38
rs780608505 Chr.19: 44623908-44623908 on Build GRCh38
rs144450214 Chr.19: 44623679-44623679 on Build GRCh38
rs1051801938 Chr.19: 44623700-44623700 on Build GRCh38
rs957855365 Chr.19: 44624044-44624044 on Build GRCh38
rs532068914 Chr.19: 44623858-44623858 on Build GRCh38
rs573168129 Chr.19: 44623669-44623669 on Build GRCh38
rs545589900 Chr.19: 44623876-44623876 on Build GRCh38
rs543189270 Chr.19: 44623751-44623751 on Build GRCh38
rs771178844 Chr.19: 44623859-44623859 on Build GRCh38
rs965247530 Chr.19: 44623692-44623692 on Build GRCh38
rs540064665 Chr.19: 44623699-44623702 on Build GRCh38
rs574424666 Chr.19: 44623747-44623747 on Build GRCh38
rs548830386 Chr.19: 44623965-44623965 on Build GRCh38
rs749812874 Chr.19: 44623814-44623814 on Build GRCh38
rs1011864795 Chr.19: 44623936-44623936 on Build GRCh38
rs749947591 Chr.19: 44623935-44623936 on Build GRCh38
rs1049073266 Chr.19: 44623986-44623986 on Build GRCh38
rs916155565 Chr.19: 44623906-44623906 on Build GRCh38
rs757196933 Chr.19: 44623879-44623879 on Build GRCh38
rs763876937 Chr.19: 44623748-44623748 on Build GRCh38
rs1024620918 Chr.19: 44623954-44623954 on Build GRCh38
rs914992806 Chr.19: 44623827-44623827 on Build GRCh38
rs772216333 Chr.19: 44623880-44623880 on Build GRCh38
rs989724772 Chr.19: 44624064-44624064 on Build GRCh38
rs901154746 Chr.19: 44624070-44624070 on Build GRCh38
rs192565688 Chr.19: 44624105-44624105 on Build GRCh38
rs749982834 Chr.19: 44624099-44624099 on Build GRCh38
rs921846401 Chr.19: 44623682-44623682 on Build GRCh38
rs571099870 Chr.19: 44624086-44624086 on Build GRCh38
rs846857 Chr.19: 44623697-44623697 on Build GRCh38
rs113309881 Chr.19: 44623712-44623712 on Build GRCh38
rs565084949 Chr.19: 44623808-44623808 on Build GRCh38
rs563186985 Chr.19: 44623755-44623755 on Build GRCh38
rs989723348 Chr.19: 44623813-44623813 on Build GRCh38
rs770685202 Chr.19: 44623986-44623987 on Build GRCh38
rs1037096667 Chr.19: 44624018-44624018 on Build GRCh38
rs559254209 Chr.19: 44623891-44623891 on Build GRCh38
rs147996658 Chr.19: 44623916-44623916 on Build GRCh38
rs374947259 Chr.19: 44623842-44623842 on Build GRCh38

View Full Product Details