Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57491415-57491952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF419
Entrez Gene ID
79744
Gene Name
zinc finger protein 419
Gene Aliases
ZAPHIR, ZNF419A
UniGene
Hs.744339
Gene Chromosome Location
Chr.19: 57487711-57494680 on Build GRCh38
Transcripts
NM_001098491.1 NM_001098492.1 NM_001098493.1 NM_001098494.1 NM_001098495.1 NM_001098496.1 NM_001291743.1 NM_001291744.1 NM_001291745.1 NM_024691.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693122 c.95A>G p.D32G Chr.19: 57491490-57491490 on Build GRCh38
5654389 c.138G>C p.L46L Chr.19: 57491596-57491596 on Build GRCh38
272215 c.33T>C p.D11D Chr.19: 57491491-57491491 on Build GRCh38
4768420 c.96T>C p.D32D Chr.19: 57491491-57491491 on Build GRCh38
1751201 c.73G>C p.D25H Chr.19: 57491531-57491531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2074067 Chr.19: 57491773-57491773 on Build GRCh38
rs184185036 Chr.19: 57491597-57491597 on Build GRCh38
rs764875881 Chr.19: 57491801-57491802 on Build GRCh38
rs749052297 Chr.19: 57491720-57491720 on Build GRCh38
rs767116789 Chr.19: 57491624-57491624 on Build GRCh38
rs779736118 Chr.19: 57491706-57491706 on Build GRCh38
rs372960635 Chr.19: 57491526-57491526 on Build GRCh38
rs756486698 Chr.19: 57491567-57491567 on Build GRCh38
rs767776878 Chr.19: 57491788-57491789 on Build GRCh38
rs766228580 Chr.19: 57491447-57491447 on Build GRCh38
rs769597339 Chr.19: 57491551-57491551 on Build GRCh38
rs760965858 Chr.19: 57491791-57491792 on Build GRCh38
rs988179356 Chr.19: 57491691-57491691 on Build GRCh38
rs939958524 Chr.19: 57491906-57491906 on Build GRCh38
rs1135692 Chr.19: 57491596-57491596 on Build GRCh38
rs758838528 Chr.19: 57491920-57491920 on Build GRCh38
rs760254697 Chr.19: 57491781-57491781 on Build GRCh38
rs549839083 Chr.19: 57491697-57491697 on Build GRCh38
rs761452855 Chr.19: 57491615-57491615 on Build GRCh38
rs776890769 Chr.19: 57491793-57491793 on Build GRCh38
rs547962908 Chr.19: 57491634-57491634 on Build GRCh38
rs764690951 Chr.19: 57491458-57491458 on Build GRCh38
rs755493632 Chr.19: 57491667-57491667 on Build GRCh38
rs193920918 Chr.19: 57491471-57491471 on Build GRCh38
rs572166534 Chr.19: 57491487-57491487 on Build GRCh38
rs765032014 Chr.19: 57491470-57491470 on Build GRCh38
rs767243314 Chr.19: 57491566-57491566 on Build GRCh38
rs748087633 Chr.19: 57491739-57491739 on Build GRCh38
rs1050981562 Chr.19: 57491844-57491844 on Build GRCh38
rs768543774 Chr.19: 57491729-57491729 on Build GRCh38
rs765563232 Chr.19: 57491833-57491833 on Build GRCh38
rs562521826 Chr.19: 57491575-57491575 on Build GRCh38
rs62126198 Chr.19: 57491491-57491491 on Build GRCh38
rs762408298 Chr.19: 57491482-57491482 on Build GRCh38
rs772223834 Chr.19: 57491537-57491537 on Build GRCh38
rs975141067 Chr.19: 57491816-57491816 on Build GRCh38
rs760368177 Chr.19: 57491637-57491637 on Build GRCh38
rs748500211 Chr.19: 57491546-57491546 on Build GRCh38
rs201293789 Chr.19: 57491502-57491502 on Build GRCh38
rs555465551 Chr.19: 57491483-57491483 on Build GRCh38
rs762945731 Chr.19: 57491555-57491555 on Build GRCh38
rs759934019 Chr.19: 57491814-57491814 on Build GRCh38
rs774033386 Chr.19: 57491614-57491614 on Build GRCh38
rs776428398 Chr.19: 57491540-57491540 on Build GRCh38
rs533657559 Chr.19: 57491679-57491679 on Build GRCh38
rs376959722 Chr.19: 57491553-57491553 on Build GRCh38
rs772708793 Chr.19: 57491778-57491778 on Build GRCh38
rs780451672 Chr.19: 57491569-57491569 on Build GRCh38
rs369932920 Chr.19: 57491732-57491732 on Build GRCh38
rs751609554 Chr.19: 57491923-57491923 on Build GRCh38
rs569798581 Chr.19: 57491443-57491443 on Build GRCh38
rs777896213 Chr.19: 57491591-57491591 on Build GRCh38
rs540265152 Chr.19: 57491536-57491536 on Build GRCh38
rs777893291 Chr.19: 57491690-57491690 on Build GRCh38
rs72490761 Chr.19: 57491640-57491640 on Build GRCh38
rs200574247 Chr.19: 57491590-57491590 on Build GRCh38
rs928268005 Chr.19: 57491467-57491467 on Build GRCh38
rs921043251 Chr.19: 57491832-57491832 on Build GRCh38
rs753171000 Chr.19: 57491678-57491678 on Build GRCh38
rs374324322 Chr.19: 57491564-57491564 on Build GRCh38
rs759526615 Chr.19: 57491460-57491460 on Build GRCh38
rs769545991 Chr.19: 57491594-57491594 on Build GRCh38
rs754366919 Chr.19: 57491578-57491578 on Build GRCh38
rs933803458 Chr.19: 57491843-57491843 on Build GRCh38
rs752279577 Chr.19: 57491476-57491476 on Build GRCh38
rs886984812 Chr.19: 57491889-57491889 on Build GRCh38
rs762008661 Chr.19: 57491439-57491439 on Build GRCh38
rs373444846 Chr.19: 57491782-57491782 on Build GRCh38
rs899685233 Chr.19: 57491503-57491505 on Build GRCh38
rs768779348 Chr.19: 57491519-57491519 on Build GRCh38
rs761957077 Chr.19: 57491786-57491787 on Build GRCh38
rs758520819 Chr.19: 57491687-57491687 on Build GRCh38
rs754170043 Chr.19: 57491665-57491665 on Build GRCh38
rs549801975 Chr.19: 57491438-57491438 on Build GRCh38
rs778959056 Chr.19: 57491522-57491522 on Build GRCh38
rs1036741250 Chr.19: 57491909-57491909 on Build GRCh38
rs536176215 Chr.19: 57491483-57491483 on Build GRCh38
rs189517219 Chr.19: 57491787-57491787 on Build GRCh38
rs778973586 Chr.19: 57491582-57491582 on Build GRCh38
rs145680611 Chr.19: 57491556-57491556 on Build GRCh38
rs759208180 Chr.19: 57491542-57491542 on Build GRCh38
rs147596854 Chr.19: 57491917-57491917 on Build GRCh38
rs764097126 Chr.19: 57491922-57491922 on Build GRCh38
rs527891060 Chr.19: 57491603-57491603 on Build GRCh38
rs182023600 Chr.19: 57491495-57491495 on Build GRCh38
rs6510085 Chr.19: 57491451-57491451 on Build GRCh38
rs747230279 Chr.19: 57491592-57491592 on Build GRCh38
rs745366490 Chr.19: 57491671-57491671 on Build GRCh38
rs750705395 Chr.19: 57491492-57491492 on Build GRCh38
rs748176708 Chr.19: 57491588-57491588 on Build GRCh38
rs368422914 Chr.19: 57491580-57491580 on Build GRCh38
rs994893182 Chr.19: 57491590-57491591 on Build GRCh38
rs747202244 Chr.19: 57491695-57491695 on Build GRCh38

View Full Product Details