Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 12717796-12718303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIB2
Entrez Gene ID
28951
Gene Name
tribbles pseudokinase 2
Gene Aliases
C5FW, GS3955, TRB2
UniGene
Hs.467751
Gene Chromosome Location
Chr.2: 12716872-12742734 on Build GRCh38
Transcripts
NM_021643.3 NR_027303.1

SNP Detail

SNP ID Chromosome Location
rs77773048 Chr.2: 12717952-12717952 on Build GRCh38
rs1017697049 Chr.2: 12718179-12718179 on Build GRCh38
rs111282515 Chr.2: 12718086-12718086 on Build GRCh38
rs1049160932 Chr.2: 12718013-12718013 on Build GRCh38
rs1013672910 Chr.2: 12717866-12717866 on Build GRCh38
rs189518756 Chr.2: 12718154-12718154 on Build GRCh38
rs1034737249 Chr.2: 12718004-12718005 on Build GRCh38
rs568203374 Chr.2: 12717846-12717846 on Build GRCh38
rs7590994 Chr.2: 12718201-12718201 on Build GRCh38
rs976343484 Chr.2: 12718189-12718189 on Build GRCh38
rs917736689 Chr.2: 12718246-12718246 on Build GRCh38
rs1008532360 Chr.2: 12717905-12717905 on Build GRCh38
rs966877954 Chr.2: 12717906-12717906 on Build GRCh38
rs1049820512 Chr.2: 12717900-12717900 on Build GRCh38
rs560008833 Chr.2: 12718276-12718276 on Build GRCh38
rs896192759 Chr.2: 12717852-12717852 on Build GRCh38
rs191327196 Chr.2: 12718014-12718014 on Build GRCh38
rs902160044 Chr.2: 12718142-12718142 on Build GRCh38
rs957525183 Chr.2: 12717940-12717940 on Build GRCh38
rs931022261 Chr.2: 12718067-12718067 on Build GRCh38
rs966713139 Chr.2: 12718186-12718186 on Build GRCh38
rs966284889 Chr.2: 12717982-12717982 on Build GRCh38
rs535599608 Chr.2: 12717947-12717947 on Build GRCh38
rs1049384931 Chr.2: 12717827-12717827 on Build GRCh38
rs187916442 Chr.2: 12718009-12718009 on Build GRCh38
rs509761 Chr.2: 12718076-12718076 on Build GRCh38
rs1042296480 Chr.2: 12718280-12718280 on Build GRCh38
rs984913217 Chr.2: 12718219-12718219 on Build GRCh38
rs1033165327 Chr.2: 12717922-12717922 on Build GRCh38
rs1010077736 Chr.2: 12718149-12718149 on Build GRCh38
rs938901106 Chr.2: 12717837-12717837 on Build GRCh38
rs990667153 Chr.2: 12718242-12718242 on Build GRCh38
rs776545753 Chr.2: 12718273-12718273 on Build GRCh38
rs1049417297 Chr.2: 12718073-12718073 on Build GRCh38
rs1007687824 Chr.2: 12718164-12718164 on Build GRCh38
rs887590970 Chr.2: 12717838-12717838 on Build GRCh38
rs909299672 Chr.2: 12718226-12718226 on Build GRCh38
rs1002003093 Chr.2: 12718178-12718178 on Build GRCh38
rs763797449 Chr.2: 12717876-12717876 on Build GRCh38
rs889491288 Chr.2: 12718162-12718162 on Build GRCh38
rs764783048 Chr.2: 12718134-12718134 on Build GRCh38
rs950740263 Chr.2: 12718212-12718212 on Build GRCh38
rs993472754 Chr.2: 12718012-12718012 on Build GRCh38
rs937826309 Chr.2: 12718022-12718022 on Build GRCh38
rs898198394 Chr.2: 12717893-12717893 on Build GRCh38
rs772378978 Chr.2: 12718265-12718265 on Build GRCh38
rs1032047781 Chr.2: 12718157-12718157 on Build GRCh38
rs528057074 Chr.2: 12718092-12718092 on Build GRCh38
rs1000598877 Chr.2: 12717912-12717912 on Build GRCh38
rs919678042 Chr.2: 12718060-12718060 on Build GRCh38
rs1042468874 Chr.2: 12718141-12718141 on Build GRCh38
rs761863456 Chr.2: 12718279-12718282 on Build GRCh38
rs889717063 Chr.2: 12717902-12717902 on Build GRCh38
rs949304720 Chr.2: 12718130-12718130 on Build GRCh38
rs867525332 Chr.2: 12718000-12718000 on Build GRCh38
rs1010359343 Chr.2: 12717931-12717931 on Build GRCh38
rs938044166 Chr.2: 12718239-12718239 on Build GRCh38
rs929378744 Chr.2: 12717819-12717819 on Build GRCh38
rs764485871 Chr.2: 12718273-12718273 on Build GRCh38
rs930990404 Chr.2: 12717898-12717898 on Build GRCh38
rs1015125232 Chr.2: 12718218-12718218 on Build GRCh38
rs919241335 Chr.2: 12718004-12718004 on Build GRCh38
rs986384060 Chr.2: 12717951-12717951 on Build GRCh38
rs574564523 Chr.2: 12718190-12718190 on Build GRCh38
rs891673976 Chr.2: 12718114-12718114 on Build GRCh38
rs758393679 Chr.2: 12718272-12718272 on Build GRCh38
rs572437376 Chr.2: 12717989-12717989 on Build GRCh38
rs113192828 Chr.2: 12718051-12718051 on Build GRCh38
rs768612987 Chr.2: 12718272-12718273 on Build GRCh38
rs765225177 Chr.2: 12718281-12718282 on Build GRCh38
rs1055041139 Chr.2: 12717851-12717851 on Build GRCh38
rs183863370 Chr.2: 12718110-12718110 on Build GRCh38

View Full Product Details