Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30738311-30738818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
UniGene
Hs.660911
Gene Chromosome Location
Chr.2: 30610382-30809527 on Build GRCh38
Transcripts
NM_144575.2 XM_011533159.2 XM_011533160.2 XM_011533161.2 XM_017005264.1 XM_017005265.1 XM_017005266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1020053 c.1567G>T p.G523C Chr.2: 30738427-30738427 on Build GRCh38
229916 c.1582G>A p.E528K Chr.2: 30738412-30738412 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs930080432 Chr.2: 30738393-30738393 on Build GRCh38
rs757889230 Chr.2: 30738732-30738733 on Build GRCh38
rs996222907 Chr.2: 30738649-30738649 on Build GRCh38
rs761007641 Chr.2: 30738617-30738617 on Build GRCh38
rs1019437271 Chr.2: 30738333-30738333 on Build GRCh38
rs777938203 Chr.2: 30738599-30738599 on Build GRCh38
rs942834867 Chr.2: 30738667-30738667 on Build GRCh38
rs1054225475 Chr.2: 30738567-30738567 on Build GRCh38
rs1007090386 Chr.2: 30738454-30738454 on Build GRCh38
rs912729585 Chr.2: 30738661-30738661 on Build GRCh38
rs902544001 Chr.2: 30738611-30738611 on Build GRCh38
rs904073219 Chr.2: 30738696-30738696 on Build GRCh38
rs35032002 Chr.2: 30738535-30738535 on Build GRCh38
rs371040924 Chr.2: 30738452-30738452 on Build GRCh38
rs891325827 Chr.2: 30738441-30738441 on Build GRCh38
rs980040061 Chr.2: 30738542-30738542 on Build GRCh38
rs556436382 Chr.2: 30738653-30738653 on Build GRCh38
rs749356688 Chr.2: 30738484-30738484 on Build GRCh38
rs534938595 Chr.2: 30738513-30738513 on Build GRCh38
rs955748538 Chr.2: 30738511-30738511 on Build GRCh38
rs1009981524 Chr.2: 30738741-30738741 on Build GRCh38
rs751414260 Chr.2: 30738355-30738355 on Build GRCh38
rs756096271 Chr.2: 30738590-30738590 on Build GRCh38
rs554480520 Chr.2: 30738350-30738350 on Build GRCh38
rs531347683 Chr.2: 30738541-30738541 on Build GRCh38
rs754270796 Chr.2: 30738651-30738651 on Build GRCh38
rs184805064 Chr.2: 30738462-30738462 on Build GRCh38
rs768535327 Chr.2: 30738495-30738495 on Build GRCh38
rs536410301 Chr.2: 30738648-30738648 on Build GRCh38
rs771210181 Chr.2: 30738491-30738491 on Build GRCh38
rs752528181 Chr.2: 30738391-30738391 on Build GRCh38
rs541357339 Chr.2: 30738364-30738364 on Build GRCh38
rs927169301 Chr.2: 30738560-30738560 on Build GRCh38
rs866528018 Chr.2: 30738412-30738412 on Build GRCh38
rs769715786 Chr.2: 30738337-30738337 on Build GRCh38
rs567569504 Chr.2: 30738632-30738632 on Build GRCh38
rs776838680 Chr.2: 30738774-30738774 on Build GRCh38
rs773331932 Chr.2: 30738479-30738479 on Build GRCh38
rs952274501 Chr.2: 30738460-30738460 on Build GRCh38
rs763968593 Chr.2: 30738340-30738341 on Build GRCh38
rs777187044 Chr.2: 30738399-30738399 on Build GRCh38
rs767503429 Chr.2: 30738370-30738370 on Build GRCh38
rs78810531 Chr.2: 30738527-30738527 on Build GRCh38
rs527296312 Chr.2: 30738621-30738621 on Build GRCh38
rs908101896 Chr.2: 30738687-30738687 on Build GRCh38
rs1024099599 Chr.2: 30738777-30738777 on Build GRCh38
rs189681298 Chr.2: 30738628-30738628 on Build GRCh38
rs570130117 Chr.2: 30738722-30738722 on Build GRCh38
rs1019070785 Chr.2: 30738644-30738644 on Build GRCh38
rs937207738 Chr.2: 30738563-30738563 on Build GRCh38
rs759746062 Chr.2: 30738489-30738489 on Build GRCh38
rs767114836 Chr.2: 30738714-30738714 on Build GRCh38
rs1039879573 Chr.2: 30738669-30738669 on Build GRCh38
rs376207020 Chr.2: 30738706-30738706 on Build GRCh38
rs974907057 Chr.2: 30738341-30738341 on Build GRCh38
rs141316385 Chr.2: 30738568-30738568 on Build GRCh38
rs143789133 Chr.2: 30738753-30738753 on Build GRCh38
rs563680539 Chr.2: 30738794-30738794 on Build GRCh38
rs866457214 Chr.2: 30738476-30738476 on Build GRCh38
rs774498205 Chr.2: 30738492-30738492 on Build GRCh38
rs983730245 Chr.2: 30738685-30738685 on Build GRCh38
rs561264625 Chr.2: 30738395-30738395 on Build GRCh38
rs927031727 Chr.2: 30738538-30738538 on Build GRCh38
rs926944110 Chr.2: 30738778-30738778 on Build GRCh38
rs748567291 Chr.2: 30738475-30738475 on Build GRCh38
rs897202964 Chr.2: 30738357-30738357 on Build GRCh38
rs891495714 Chr.2: 30738740-30738740 on Build GRCh38
rs952160182 Chr.2: 30738655-30738655 on Build GRCh38
rs780084903 Chr.2: 30738411-30738411 on Build GRCh38
rs543614208 Chr.2: 30738510-30738510 on Build GRCh38
rs1032689564 Chr.2: 30738506-30738506 on Build GRCh38
rs2593439 Chr.2: 30738780-30738780 on Build GRCh38
rs904106293 Chr.2: 30738465-30738465 on Build GRCh38
rs372856121 Chr.2: 30738416-30738416 on Build GRCh38
rs370849221 Chr.2: 30738526-30738526 on Build GRCh38
rs72867127 Chr.2: 30738586-30738586 on Build GRCh38
rs747386499 Chr.2: 30738439-30738439 on Build GRCh38
rs753373607 Chr.2: 30738400-30738400 on Build GRCh38
rs770305481 Chr.2: 30738478-30738478 on Build GRCh38

View Full Product Details