Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 40114723-40115271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8A1
Entrez Gene ID
6546
Gene Name
solute carrier family 8 member A1
Gene Aliases
NCX1
UniGene
Hs.31961
Gene Chromosome Location
Chr.2: 40112146-40512452 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Gene Symbol
SLC8A1-AS1
Entrez Gene ID
100128590
Gene Name
SLC8A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.744988
Gene Chromosome Location
Chr.2: 39917634-40255209 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3372777 c.2800A>G p.I934V Chr.2: 40115267-40115267 on Build GRCh38
6209475 c.2914G>A p.G972S Chr.2: 40115261-40115261 on Build GRCh38
4786414 c.2796C>A p.C932* Chr.2: 40115271-40115271 on Build GRCh38
6509275 c.2807G>T p.G936V Chr.2: 40115260-40115260 on Build GRCh38
3372778 c.2800A>G p.I934V Chr.2: 40115267-40115267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72558069 Chr.2: 40115003-40115003 on Build GRCh38
rs993759817 Chr.2: 40114956-40114956 on Build GRCh38
rs752982570 Chr.2: 40115205-40115205 on Build GRCh38
rs572394483 Chr.2: 40115076-40115076 on Build GRCh38
rs550020233 Chr.2: 40115180-40115180 on Build GRCh38
rs753403545 Chr.2: 40115025-40115025 on Build GRCh38
rs939878515 Chr.2: 40115089-40115089 on Build GRCh38
rs983926352 Chr.2: 40115074-40115074 on Build GRCh38
rs778989167 Chr.2: 40115235-40115235 on Build GRCh38
rs887006795 Chr.2: 40114938-40114938 on Build GRCh38
rs72791077 Chr.2: 40115055-40115055 on Build GRCh38
rs755939167 Chr.2: 40115116-40115117 on Build GRCh38
rs888828123 Chr.2: 40114897-40114897 on Build GRCh38
rs1041970901 Chr.2: 40114834-40114834 on Build GRCh38
rs5539 Chr.2: 40114994-40114994 on Build GRCh38
rs199611406 Chr.2: 40115212-40115212 on Build GRCh38
rs901122362 Chr.2: 40115101-40115101 on Build GRCh38
rs745724859 Chr.2: 40115237-40115237 on Build GRCh38
rs1021109643 Chr.2: 40114998-40114998 on Build GRCh38
rs977576441 Chr.2: 40115207-40115207 on Build GRCh38
rs879303294 Chr.2: 40115041-40115041 on Build GRCh38
rs1042512946 Chr.2: 40115155-40115155 on Build GRCh38
rs868150182 Chr.2: 40115086-40115086 on Build GRCh38
rs908392787 Chr.2: 40115075-40115075 on Build GRCh38
rs923523296 Chr.2: 40115060-40115060 on Build GRCh38
rs938412112 Chr.2: 40114831-40114831 on Build GRCh38
rs915643520 Chr.2: 40115066-40115066 on Build GRCh38
rs901176234 Chr.2: 40114966-40114966 on Build GRCh38
rs531789156 Chr.2: 40115138-40115138 on Build GRCh38
rs1049773663 Chr.2: 40115116-40115116 on Build GRCh38
rs1024878191 Chr.2: 40114961-40114961 on Build GRCh38
rs991651897 Chr.2: 40114839-40114839 on Build GRCh38
rs775108252 Chr.2: 40115240-40115240 on Build GRCh38
rs758668689 Chr.2: 40115226-40115227 on Build GRCh38
rs993313897 Chr.2: 40115108-40115108 on Build GRCh38
rs902693917 Chr.2: 40115173-40115173 on Build GRCh38
rs959983315 Chr.2: 40115051-40115051 on Build GRCh38
rs986378442 Chr.2: 40115218-40115218 on Build GRCh38
rs1003718021 Chr.2: 40115179-40115179 on Build GRCh38
rs1005982344 Chr.2: 40114902-40114906 on Build GRCh38
rs946241627 Chr.2: 40114832-40114832 on Build GRCh38
rs749354098 Chr.2: 40115219-40115219 on Build GRCh38
rs977046493 Chr.2: 40114798-40114798 on Build GRCh38
rs752742485 Chr.2: 40115130-40115130 on Build GRCh38
rs915713460 Chr.2: 40114848-40114848 on Build GRCh38
rs1048264420 Chr.2: 40114877-40114877 on Build GRCh38
rs561829178 Chr.2: 40115211-40115211 on Build GRCh38
rs897596527 Chr.2: 40115103-40115103 on Build GRCh38
rs778072284 Chr.2: 40114823-40114823 on Build GRCh38
rs910828946 Chr.2: 40115219-40115222 on Build GRCh38
rs931101659 Chr.2: 40115072-40115072 on Build GRCh38
rs72558068 Chr.2: 40115093-40115093 on Build GRCh38
rs529067751 Chr.2: 40115225-40115225 on Build GRCh38
rs1011471642 Chr.2: 40114995-40114995 on Build GRCh38
rs190612619 Chr.2: 40115109-40115109 on Build GRCh38
rs1022658439 Chr.2: 40115229-40115229 on Build GRCh38
rs371847204 Chr.2: 40114807-40114807 on Build GRCh38
rs777814249 Chr.2: 40114907-40114909 on Build GRCh38
rs1028817074 Chr.2: 40114988-40114988 on Build GRCh38
rs746309791 Chr.2: 40115218-40115219 on Build GRCh38
rs990240737 Chr.2: 40114766-40114766 on Build GRCh38
rs35552627 Chr.2: 40115183-40115184 on Build GRCh38
rs982350038 Chr.2: 40115020-40115020 on Build GRCh38
rs771914092 Chr.2: 40115238-40115238 on Build GRCh38
rs538053706 Chr.2: 40114872-40114872 on Build GRCh38
rs376135853 Chr.2: 40114939-40114939 on Build GRCh38
rs749807832 Chr.2: 40114982-40114982 on Build GRCh38
rs1050105490 Chr.2: 40114883-40114884 on Build GRCh38
rs771367640 Chr.2: 40114847-40114847 on Build GRCh38
rs1021699930 Chr.2: 40115164-40115164 on Build GRCh38
rs990186532 Chr.2: 40115006-40115006 on Build GRCh38
rs958750521 Chr.2: 40115004-40115004 on Build GRCh38
rs914681373 Chr.2: 40115023-40115023 on Build GRCh38
rs1035281038 Chr.2: 40115050-40115050 on Build GRCh38
rs5540 Chr.2: 40114949-40114949 on Build GRCh38
rs939931954 Chr.2: 40114947-40114947 on Build GRCh38
rs888775823 Chr.2: 40115085-40115085 on Build GRCh38
rs937446507 Chr.2: 40114764-40114764 on Build GRCh38

View Full Product Details