Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 40115400-40115897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8A1
Entrez Gene ID
6546
Gene Name
solute carrier family 8 member A1
Gene Aliases
NCX1
UniGene
Hs.31961
Gene Chromosome Location
Chr.2: 40112146-40512452 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Gene Symbol
SLC8A1-AS1
Entrez Gene ID
100128590
Gene Name
SLC8A1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.744988
Gene Chromosome Location
Chr.2: 39917634-40255209 on Build GRCh38
Transcripts
NM_001112800.1 NM_001112801.1 NM_001112802.1 NM_001252624.1 NM_021097.2 XM_005264514.3 XM_006712081.2 XM_006712082.3 XM_006712083.3 XM_006712084.3 XM_006712085.3 XM_011533050.1 XM_011533054.2 XM_011533055.2 XM_011533056.1 XM_011533057.2 XM_017004745.1 XM_017004746.1 XM_017004747.1 XM_017004748.1 XM_017004749.1 XM_017004750.1 XM_017004751.1 XM_017004752.1 XM_017004753.1 XM_017004754.1 XM_017004755.1 XM_017004756.1 XM_017004757.1 XM_017004758.1 XM_017004759.1 XM_017004760.1 XM_017004761.1 XM_017004762.1 XM_017004763.1 NR_038441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4313378 c.2477A>T p.Y826F Chr.2: 40115590-40115590 on Build GRCh38
6091597 c.2658G>T p.V886V Chr.2: 40115409-40115409 on Build GRCh38
353131 c.2623G>T p.A875S Chr.2: 40115552-40115552 on Build GRCh38
6158224 c.2737C>A p.L913I Chr.2: 40115438-40115438 on Build GRCh38
387018 c.2607C>A p.N869K Chr.2: 40115568-40115568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777095687 Chr.2: 40115638-40115638 on Build GRCh38
rs183947542 Chr.2: 40115686-40115686 on Build GRCh38
rs915266590 Chr.2: 40115494-40115494 on Build GRCh38
rs888064345 Chr.2: 40115820-40115820 on Build GRCh38
rs961137973 Chr.2: 40115663-40115663 on Build GRCh38
rs867120963 Chr.2: 40115533-40115533 on Build GRCh38
rs112945578 Chr.2: 40115554-40115554 on Build GRCh38
rs775979049 Chr.2: 40115488-40115488 on Build GRCh38
rs201804871 Chr.2: 40115467-40115467 on Build GRCh38
rs767885076 Chr.2: 40115483-40115483 on Build GRCh38
rs370669554 Chr.2: 40115660-40115660 on Build GRCh38
rs531492197 Chr.2: 40115736-40115736 on Build GRCh38
rs576623727 Chr.2: 40115619-40115619 on Build GRCh38
rs747744815 Chr.2: 40115442-40115442 on Build GRCh38
rs187957282 Chr.2: 40115553-40115553 on Build GRCh38
rs550978029 Chr.2: 40115801-40115801 on Build GRCh38
rs752463757 Chr.2: 40115671-40115671 on Build GRCh38
rs192452025 Chr.2: 40115562-40115562 on Build GRCh38
rs937834073 Chr.2: 40115723-40115723 on Build GRCh38
rs375657524 Chr.2: 40115507-40115507 on Build GRCh38
rs939466291 Chr.2: 40115779-40115779 on Build GRCh38
rs1054898453 Chr.2: 40115729-40115729 on Build GRCh38
rs1003665406 Chr.2: 40115511-40115511 on Build GRCh38
rs575515901 Chr.2: 40115744-40115744 on Build GRCh38
rs780034980 Chr.2: 40115868-40115868 on Build GRCh38
rs748731223 Chr.2: 40115583-40115583 on Build GRCh38
rs762175920 Chr.2: 40115650-40115650 on Build GRCh38
rs998889868 Chr.2: 40115770-40115770 on Build GRCh38
rs545745311 Chr.2: 40115759-40115759 on Build GRCh38
rs368879351 Chr.2: 40115750-40115750 on Build GRCh38
rs766426625 Chr.2: 40115662-40115662 on Build GRCh38
rs141218115 Chr.2: 40115784-40115784 on Build GRCh38
rs765544343 Chr.2: 40115651-40115651 on Build GRCh38
rs932611785 Chr.2: 40115438-40115438 on Build GRCh38
rs878856540 Chr.2: 40115529-40115529 on Build GRCh38
rs963630801 Chr.2: 40115809-40115809 on Build GRCh38
rs564442817 Chr.2: 40115595-40115595 on Build GRCh38
rs986324456 Chr.2: 40115548-40115548 on Build GRCh38
rs1005202177 Chr.2: 40115826-40115826 on Build GRCh38
rs973055147 Chr.2: 40115568-40115568 on Build GRCh38
rs890556178 Chr.2: 40115781-40115781 on Build GRCh38
rs922472028 Chr.2: 40115730-40115730 on Build GRCh38
rs543965268 Chr.2: 40115636-40115636 on Build GRCh38
rs1015347619 Chr.2: 40115634-40115634 on Build GRCh38
rs531921299 Chr.2: 40115747-40115747 on Build GRCh38
rs141423996 Chr.2: 40115493-40115493 on Build GRCh38
rs35480686 Chr.2: 40115486-40115487 on Build GRCh38
rs776229256 Chr.2: 40115780-40115780 on Build GRCh38
rs760360226 Chr.2: 40115691-40115691 on Build GRCh38
rs560427129 Chr.2: 40115496-40115496 on Build GRCh38
rs370192379 Chr.2: 40115520-40115520 on Build GRCh38
rs781000410 Chr.2: 40115670-40115670 on Build GRCh38
rs769400672 Chr.2: 40115567-40115567 on Build GRCh38
rs779852676 Chr.2: 40115551-40115551 on Build GRCh38
rs765449185 Chr.2: 40115547-40115547 on Build GRCh38
rs755818311 Chr.2: 40115675-40115675 on Build GRCh38
rs772823388 Chr.2: 40115582-40115582 on Build GRCh38
rs536628361 Chr.2: 40115849-40115849 on Build GRCh38
rs1042409996 Chr.2: 40115762-40115762 on Build GRCh38
rs562162811 Chr.2: 40115654-40115654 on Build GRCh38
rs746900476 Chr.2: 40115552-40115552 on Build GRCh38
rs981771860 Chr.2: 40115866-40115866 on Build GRCh38
rs765156795 Chr.2: 40115813-40115813 on Build GRCh38
rs772700481 Chr.2: 40115591-40115591 on Build GRCh38
rs927741045 Chr.2: 40115719-40115719 on Build GRCh38
rs895318685 Chr.2: 40115806-40115806 on Build GRCh38
rs777755714 Chr.2: 40115563-40115563 on Build GRCh38
rs760942212 Chr.2: 40115495-40115495 on Build GRCh38
rs776273809 Chr.2: 40115433-40115433 on Build GRCh38
rs924010772 Chr.2: 40115776-40115776 on Build GRCh38
rs555029931 Chr.2: 40115459-40115459 on Build GRCh38
rs374472434 Chr.2: 40115535-40115535 on Build GRCh38
rs184171310 Chr.2: 40115550-40115550 on Build GRCh38
rs751478459 Chr.2: 40115664-40115664 on Build GRCh38
rs41280643 Chr.2: 40115818-40115818 on Build GRCh38
rs780419192 Chr.2: 40115647-40115648 on Build GRCh38
rs375826653 Chr.2: 40115580-40115580 on Build GRCh38
rs757506199 Chr.2: 40115544-40115544 on Build GRCh38
rs777283379 Chr.2: 40115681-40115681 on Build GRCh38
rs902641212 Chr.2: 40115498-40115498 on Build GRCh38
rs372922406 Chr.2: 40115457-40115457 on Build GRCh38
rs754873460 Chr.2: 40115666-40115666 on Build GRCh38
rs1030404932 Chr.2: 40115771-40115771 on Build GRCh38
rs138110651 Chr.2: 40115522-40115522 on Build GRCh38
rs148215685 Chr.2: 40115460-40115460 on Build GRCh38

View Full Product Details