Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73074826-73075335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP5
Entrez Gene ID
26056
Gene Name
RAB11 family interacting protein 5
Gene Aliases
GAF1, RIP11, pp75
UniGene
Hs.24557
Gene Chromosome Location
Chr.2: 73073382-73113018 on Build GRCh38
Transcripts
NM_015470.2 XM_005264251.3 XM_005264252.3 XM_006711985.3 XM_011532753.2 XM_017003788.1

SNP Detail

SNP ID Chromosome Location
rs968454803 Chr.2: 73075019-73075019 on Build GRCh38
rs1005688226 Chr.2: 73074859-73074859 on Build GRCh38
rs774287119 Chr.2: 73075264-73075264 on Build GRCh38
rs543775175 Chr.2: 73075279-73075279 on Build GRCh38
rs141828110 Chr.2: 73074910-73074910 on Build GRCh38
rs1020953509 Chr.2: 73074939-73074939 on Build GRCh38
rs775626569 Chr.2: 73074856-73074856 on Build GRCh38
rs934637332 Chr.2: 73074988-73074988 on Build GRCh38
rs889596406 Chr.2: 73074891-73074891 on Build GRCh38
rs574829832 Chr.2: 73075063-73075063 on Build GRCh38
rs750575654 Chr.2: 73074921-73074921 on Build GRCh38
rs1008084626 Chr.2: 73074893-73074893 on Build GRCh38
rs961144107 Chr.2: 73075081-73075081 on Build GRCh38
rs762741168 Chr.2: 73075207-73075207 on Build GRCh38
rs919569908 Chr.2: 73075130-73075130 on Build GRCh38
rs972932076 Chr.2: 73075112-73075112 on Build GRCh38
rs931213779 Chr.2: 73074849-73074849 on Build GRCh38
rs565143147 Chr.2: 73075305-73075305 on Build GRCh38
rs185880130 Chr.2: 73075179-73075179 on Build GRCh38
rs113543144 Chr.2: 73074909-73074909 on Build GRCh38
rs1001037689 Chr.2: 73075025-73075025 on Build GRCh38
rs1044135520 Chr.2: 73075044-73075044 on Build GRCh38
rs189450864 Chr.2: 73075065-73075065 on Build GRCh38
rs776778776 Chr.2: 73075198-73075198 on Build GRCh38
rs534664748 Chr.2: 73074994-73074994 on Build GRCh38
rs764150622 Chr.2: 73075254-73075254 on Build GRCh38
rs116257706 Chr.2: 73075055-73075055 on Build GRCh38
rs992266021 Chr.2: 73075050-73075050 on Build GRCh38
rs897307506 Chr.2: 73075152-73075152 on Build GRCh38
rs947222484 Chr.2: 73074956-73074956 on Build GRCh38
rs181379691 Chr.2: 73074901-73074901 on Build GRCh38
rs1033905570 Chr.2: 73075026-73075026 on Build GRCh38
rs978586154 Chr.2: 73074959-73074959 on Build GRCh38
rs945443448 Chr.2: 73075304-73075304 on Build GRCh38
rs977405347 Chr.2: 73074877-73074877 on Build GRCh38
rs1022811627 Chr.2: 73075286-73075286 on Build GRCh38
rs959507890 Chr.2: 73075049-73075049 on Build GRCh38
rs181498306 Chr.2: 73075137-73075137 on Build GRCh38
rs761742146 Chr.2: 73074876-73074876 on Build GRCh38
rs915305763 Chr.2: 73074926-73074926 on Build GRCh38
rs930981650 Chr.2: 73075170-73075170 on Build GRCh38
rs1006135237 Chr.2: 73075108-73075108 on Build GRCh38
rs891663790 Chr.2: 73075293-73075293 on Build GRCh38
rs570543396 Chr.2: 73074976-73074976 on Build GRCh38

View Full Product Details