Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84431370-84431879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCLG1
Entrez Gene ID
8802
Gene Name
succinate-CoA ligase alpha subunit
Gene Aliases
GALPHA, MTDPS9, SUCLA1
UniGene
Hs.270428
Gene Chromosome Location
Chr.2: 84423523-84459462 on Build GRCh38
Transcripts
NM_003849.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3001048 c.806T>C p.F269S Chr.2: 84431527-84431527 on Build GRCh38
4095993 c.735C>T p.N245N Chr.2: 84431598-84431598 on Build GRCh38
1194674 c.765T>G p.I255M Chr.2: 84431568-84431568 on Build GRCh38
197393 c.721G>A p.E241K Chr.2: 84431612-84431612 on Build GRCh38
1023233 c.753C>T p.G251G Chr.2: 84431580-84431580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745797257 Chr.2: 84431794-84431794 on Build GRCh38
rs778016037 Chr.2: 84431641-84431641 on Build GRCh38
rs748368534 Chr.2: 84431513-84431513 on Build GRCh38
rs779008093 Chr.2: 84431509-84431509 on Build GRCh38
rs373564399 Chr.2: 84431812-84431813 on Build GRCh38
rs747218566 Chr.2: 84431642-84431642 on Build GRCh38
rs949817531 Chr.2: 84431444-84431444 on Build GRCh38
rs1001422962 Chr.2: 84431479-84431479 on Build GRCh38
rs758417336 Chr.2: 84431635-84431635 on Build GRCh38
rs113220946 Chr.2: 84431542-84431542 on Build GRCh38
rs546579829 Chr.2: 84431481-84431481 on Build GRCh38
rs143229361 Chr.2: 84431490-84431490 on Build GRCh38
rs527586632 Chr.2: 84431396-84431396 on Build GRCh38
rs370376281 Chr.2: 84431687-84431687 on Build GRCh38
rs191563521 Chr.2: 84431807-84431807 on Build GRCh38
rs888648636 Chr.2: 84431819-84431819 on Build GRCh38
rs377483465 Chr.2: 84431598-84431598 on Build GRCh38
rs776767968 Chr.2: 84431785-84431785 on Build GRCh38
rs924067038 Chr.2: 84431569-84431569 on Build GRCh38
rs750388794 Chr.2: 84431507-84431507 on Build GRCh38
rs1057518365 Chr.2: 84431588-84431588 on Build GRCh38
rs1023891770 Chr.2: 84431848-84431848 on Build GRCh38
rs1012334359 Chr.2: 84431844-84431844 on Build GRCh38
rs762680540 Chr.2: 84431590-84431590 on Build GRCh38
rs549987155 Chr.2: 84431603-84431603 on Build GRCh38
rs1045526877 Chr.2: 84431836-84431836 on Build GRCh38
rs768822116 Chr.2: 84431423-84431423 on Build GRCh38
rs781113881 Chr.2: 84431494-84431494 on Build GRCh38
rs777875336 Chr.2: 84431516-84431516 on Build GRCh38
rs776153953 Chr.2: 84431698-84431698 on Build GRCh38
rs369370817 Chr.2: 84431515-84431515 on Build GRCh38
rs373814688 Chr.2: 84431530-84431530 on Build GRCh38
rs796052051 Chr.2: 84431557-84431557 on Build GRCh38
rs145106503 Chr.2: 84431612-84431612 on Build GRCh38
rs771230994 Chr.2: 84431533-84431533 on Build GRCh38
rs369964431 Chr.2: 84431806-84431806 on Build GRCh38
rs145973147 Chr.2: 84431613-84431613 on Build GRCh38
rs1024298437 Chr.2: 84431476-84431476 on Build GRCh38
rs764863544 Chr.2: 84431468-84431468 on Build GRCh38
rs1037149961 Chr.2: 84431412-84431412 on Build GRCh38
rs781720083 Chr.2: 84431666-84431666 on Build GRCh38
rs568592324 Chr.2: 84431622-84431622 on Build GRCh38
rs572730457 Chr.2: 84431811-84431811 on Build GRCh38
rs949765928 Chr.2: 84431812-84431812 on Build GRCh38
rs767234604 Chr.2: 84431607-84431607 on Build GRCh38
rs777009364 Chr.2: 84431461-84431461 on Build GRCh38
rs943721369 Chr.2: 84431639-84431639 on Build GRCh38
rs149171674 Chr.2: 84431448-84431448 on Build GRCh38
rs994315718 Chr.2: 84431399-84431399 on Build GRCh38
rs777230652 Chr.2: 84431548-84431548 on Build GRCh38
rs763743761 Chr.2: 84431597-84431597 on Build GRCh38
rs1010065778 Chr.2: 84431546-84431546 on Build GRCh38
rs760170442 Chr.2: 84431576-84431576 on Build GRCh38
rs775204941 Chr.2: 84431584-84431584 on Build GRCh38
rs536019894 Chr.2: 84431806-84431807 on Build GRCh38
rs759077313 Chr.2: 84431466-84431466 on Build GRCh38
rs781250063 Chr.2: 84431436-84431436 on Build GRCh38
rs111970833 Chr.2: 84431693-84431693 on Build GRCh38
rs756603330 Chr.2: 84431604-84431605 on Build GRCh38
rs956744733 Chr.2: 84431582-84431582 on Build GRCh38
rs754998980 Chr.2: 84431508-84431508 on Build GRCh38
rs539741704 Chr.2: 84431856-84431856 on Build GRCh38
rs763792155 Chr.2: 84431482-84431482 on Build GRCh38
rs140950916 Chr.2: 84431581-84431581 on Build GRCh38
rs770479768 Chr.2: 84431695-84431695 on Build GRCh38
rs373709522 Chr.2: 84431472-84431472 on Build GRCh38
rs757013081 Chr.2: 84431491-84431491 on Build GRCh38
rs535815226 Chr.2: 84431776-84431776 on Build GRCh38
rs770827084 Chr.2: 84431573-84431573 on Build GRCh38
rs1020570482 Chr.2: 84431549-84431549 on Build GRCh38
rs757620469 Chr.2: 84431654-84431654 on Build GRCh38

View Full Product Details