Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85368589-85369080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
UniGene
Hs.269990
Gene Chromosome Location
Chr.2: 85354394-85391752 on Build GRCh38
Transcripts
NM_001135021.1 NM_001135022.1 NM_001135023.1 NM_032213.4 XM_005264596.3 XM_005264597.3 XM_005264598.3 XM_017005078.1 XM_017005079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6885888 c.264G>A p.E88E Chr.2: 85368750-85368750 on Build GRCh38
7143082 c.262G>A p.E88K Chr.2: 85368748-85368748 on Build GRCh38
3758391 c.209C>T p.T70I Chr.2: 85368695-85368695 on Build GRCh38
7143083 c.262G>A p.E88K Chr.2: 85368748-85368748 on Build GRCh38
6653415 c.229G>C p.E77Q Chr.2: 85368715-85368715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955592 Chr.2: 85368695-85368695 on Build GRCh38
rs978055567 Chr.2: 85368759-85368759 on Build GRCh38
rs374281804 Chr.2: 85369045-85369045 on Build GRCh38
rs886081258 Chr.2: 85368867-85368867 on Build GRCh38
rs890299817 Chr.2: 85369022-85369022 on Build GRCh38
rs948728967 Chr.2: 85369003-85369003 on Build GRCh38
rs986771695 Chr.2: 85368997-85368997 on Build GRCh38
rs952647999 Chr.2: 85368949-85368949 on Build GRCh38
rs765214839 Chr.2: 85368702-85368702 on Build GRCh38
rs766620068 Chr.2: 85368737-85368740 on Build GRCh38
rs779335836 Chr.2: 85368765-85368765 on Build GRCh38
rs912398439 Chr.2: 85368612-85368612 on Build GRCh38
rs1051609333 Chr.2: 85368795-85368795 on Build GRCh38
rs72840080 Chr.2: 85368920-85368920 on Build GRCh38
rs567428858 Chr.2: 85368984-85368984 on Build GRCh38
rs750668721 Chr.2: 85368745-85368745 on Build GRCh38
rs1038869169 Chr.2: 85368815-85368815 on Build GRCh38
rs760927281 Chr.2: 85368761-85368761 on Build GRCh38
rs12104980 Chr.2: 85368903-85368903 on Build GRCh38
rs893402509 Chr.2: 85368853-85368853 on Build GRCh38
rs770691510 Chr.2: 85368692-85368692 on Build GRCh38
rs550119349 Chr.2: 85368619-85368619 on Build GRCh38
rs111946526 Chr.2: 85368968-85368968 on Build GRCh38
rs546384429 Chr.2: 85368981-85368981 on Build GRCh38
rs978170153 Chr.2: 85368934-85368934 on Build GRCh38
rs773874462 Chr.2: 85368704-85368705 on Build GRCh38
rs1003222500 Chr.2: 85368873-85368873 on Build GRCh38
rs200984088 Chr.2: 85368684-85368684 on Build GRCh38
rs7420282 Chr.2: 85368862-85368862 on Build GRCh38
rs1011798830 Chr.2: 85368854-85368854 on Build GRCh38
rs746693011 Chr.2: 85368787-85368787 on Build GRCh38
rs1049052744 Chr.2: 85368700-85368700 on Build GRCh38
rs761914219 Chr.2: 85368733-85368733 on Build GRCh38
rs7340196 Chr.2: 85368807-85368807 on Build GRCh38
rs778427720 Chr.2: 85368675-85368675 on Build GRCh38
rs183153345 Chr.2: 85368901-85368901 on Build GRCh38
rs1028425766 Chr.2: 85368883-85368883 on Build GRCh38
rs943341759 Chr.2: 85369017-85369017 on Build GRCh38
rs919720010 Chr.2: 85368931-85368931 on Build GRCh38
rs961538624 Chr.2: 85368906-85368906 on Build GRCh38
rs990666448 Chr.2: 85368962-85368962 on Build GRCh38
rs750804968 Chr.2: 85368647-85368647 on Build GRCh38
rs777327264 Chr.2: 85368783-85368783 on Build GRCh38
rs186339439 Chr.2: 85368957-85368957 on Build GRCh38
rs894451472 Chr.2: 85368723-85368723 on Build GRCh38
rs144657153 Chr.2: 85368904-85368904 on Build GRCh38
rs564544668 Chr.2: 85369044-85369044 on Build GRCh38
rs920469786 Chr.2: 85369025-85369025 on Build GRCh38
rs749841842 Chr.2: 85368656-85368656 on Build GRCh38
rs139481126 Chr.2: 85368740-85368740 on Build GRCh38
rs7340198 Chr.2: 85368798-85368798 on Build GRCh38
rs373496479 Chr.2: 85368679-85368679 on Build GRCh38
rs774583444 Chr.2: 85368707-85368707 on Build GRCh38
rs756502601 Chr.2: 85368653-85368653 on Build GRCh38
rs911759252 Chr.2: 85369014-85369014 on Build GRCh38
rs143127882 Chr.2: 85368654-85368654 on Build GRCh38
rs763563707 Chr.2: 85368705-85368706 on Build GRCh38
rs770381810 Chr.2: 85368702-85368703 on Build GRCh38
rs755448717 Chr.2: 85368658-85368658 on Build GRCh38
rs757916482 Chr.2: 85368769-85368769 on Build GRCh38
rs531982816 Chr.2: 85369057-85369057 on Build GRCh38
rs370500612 Chr.2: 85368764-85368764 on Build GRCh38
rs374114666 Chr.2: 85368627-85368627 on Build GRCh38
rs746673570 Chr.2: 85368688-85368688 on Build GRCh38
rs1053401356 Chr.2: 85368846-85368846 on Build GRCh38
rs570693706 Chr.2: 85368899-85368899 on Build GRCh38

View Full Product Details