Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85661318-85661816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFTPB
Entrez Gene ID
6439
Gene Name
surfactant protein B
Gene Aliases
PSP-B, SFTB3, SFTP3, SMDP1, SP-B
UniGene
Hs.512690
Gene Chromosome Location
Chr.2: 85657317-85668741 on Build GRCh38
Transcripts
NM_000542.3 NM_198843.2 XM_005264487.2 XM_005264488.3 XM_017004702.1 XM_017004703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5002416 c.1094T>G p.V365G Chr.2: 85661525-85661525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs993103162 Chr.2: 85661583-85661583 on Build GRCh38
rs1040302102 Chr.2: 85661538-85661538 on Build GRCh38
rs760486731 Chr.2: 85661570-85661570 on Build GRCh38
rs749530951 Chr.2: 85661639-85661639 on Build GRCh38
rs1022785876 Chr.2: 85661456-85661456 on Build GRCh38
rs968728232 Chr.2: 85661738-85661738 on Build GRCh38
rs561905742 Chr.2: 85661679-85661679 on Build GRCh38
rs1026204298 Chr.2: 85661584-85661584 on Build GRCh38
rs951027870 Chr.2: 85661596-85661596 on Build GRCh38
rs184713280 Chr.2: 85661348-85661348 on Build GRCh38
rs901512774 Chr.2: 85661418-85661418 on Build GRCh38
rs757642887 Chr.2: 85661522-85661522 on Build GRCh38
rs149825633 Chr.2: 85661668-85661668 on Build GRCh38
rs995774221 Chr.2: 85661549-85661549 on Build GRCh38
rs561499435 Chr.2: 85661350-85661350 on Build GRCh38
rs370785493 Chr.2: 85661481-85661481 on Build GRCh38
rs541068019 Chr.2: 85661757-85661757 on Build GRCh38
rs1018586876 Chr.2: 85661416-85661416 on Build GRCh38
rs189886757 Chr.2: 85661669-85661669 on Build GRCh38
rs374932425 Chr.2: 85661510-85661510 on Build GRCh38
rs373005606 Chr.2: 85661582-85661582 on Build GRCh38
rs781349851 Chr.2: 85661524-85661524 on Build GRCh38
rs368851995 Chr.2: 85661586-85661586 on Build GRCh38
rs755182538 Chr.2: 85661570-85661571 on Build GRCh38
rs768460155 Chr.2: 85661454-85661454 on Build GRCh38
rs576208643 Chr.2: 85661687-85661687 on Build GRCh38
rs368016212 Chr.2: 85661504-85661504 on Build GRCh38
rs540753929 Chr.2: 85661388-85661388 on Build GRCh38
rs144831319 Chr.2: 85661483-85661483 on Build GRCh38
rs935379565 Chr.2: 85661503-85661503 on Build GRCh38
rs980850393 Chr.2: 85661482-85661482 on Build GRCh38
rs769355789 Chr.2: 85661441-85661441 on Build GRCh38
rs774139517 Chr.2: 85661455-85661455 on Build GRCh38
rs756623575 Chr.2: 85661530-85661530 on Build GRCh38
rs45529633 Chr.2: 85661461-85661461 on Build GRCh38
rs35030790 Chr.2: 85661419-85661419 on Build GRCh38
rs946917663 Chr.2: 85661513-85661513 on Build GRCh38
rs377400299 Chr.2: 85661445-85661445 on Build GRCh38
rs551956264 Chr.2: 85661497-85661497 on Build GRCh38
rs773204473 Chr.2: 85661470-85661470 on Build GRCh38
rs943503407 Chr.2: 85661788-85661788 on Build GRCh38
rs573547914 Chr.2: 85661735-85661735 on Build GRCh38
rs964676516 Chr.2: 85661347-85661347 on Build GRCh38
rs764091759 Chr.2: 85661575-85661575 on Build GRCh38
rs751886630 Chr.2: 85661500-85661501 on Build GRCh38
rs555668835 Chr.2: 85661722-85661722 on Build GRCh38
rs772842721 Chr.2: 85661551-85661551 on Build GRCh38
rs747700146 Chr.2: 85661548-85661548 on Build GRCh38
rs567527986 Chr.2: 85661600-85661600 on Build GRCh38
rs770903345 Chr.2: 85661571-85661571 on Build GRCh38
rs546688676 Chr.2: 85661648-85661648 on Build GRCh38
rs1026151966 Chr.2: 85661426-85661426 on Build GRCh38
rs750958534 Chr.2: 85661529-85661529 on Build GRCh38
rs563880358 Chr.2: 85661541-85661541 on Build GRCh38
rs369092640 Chr.2: 85661431-85661431 on Build GRCh38
rs771704983 Chr.2: 85661464-85661464 on Build GRCh38
rs370528656 Chr.2: 85661506-85661506 on Build GRCh38
rs573610627 Chr.2: 85661368-85661368 on Build GRCh38
rs371367742 Chr.2: 85661432-85661432 on Build GRCh38
rs35384723 Chr.2: 85661427-85661428 on Build GRCh38
rs979703234 Chr.2: 85661745-85661745 on Build GRCh38
rs978652518 Chr.2: 85661697-85661697 on Build GRCh38
rs1035437389 Chr.2: 85661601-85661601 on Build GRCh38
rs371255380 Chr.2: 85661458-85661458 on Build GRCh38
rs771924404 Chr.2: 85661550-85661550 on Build GRCh38
rs988394080 Chr.2: 85661499-85661499 on Build GRCh38
rs3024832 Chr.2: 85661661-85661661 on Build GRCh38
rs559530265 Chr.2: 85661421-85661421 on Build GRCh38
rs780688071 Chr.2: 85661535-85661535 on Build GRCh38
rs776535857 Chr.2: 85661572-85661572 on Build GRCh38
rs1007295597 Chr.2: 85661392-85661392 on Build GRCh38
rs45602841 Chr.2: 85661647-85661647 on Build GRCh38
rs910743474 Chr.2: 85661779-85661779 on Build GRCh38
rs901522764 Chr.2: 85661558-85661558 on Build GRCh38
rs888832769 Chr.2: 85661367-85661367 on Build GRCh38
rs535050091 Chr.2: 85661629-85661629 on Build GRCh38
rs951081146 Chr.2: 85661395-85661395 on Build GRCh38
rs367642416 Chr.2: 85661544-85661544 on Build GRCh38
rs995154388 Chr.2: 85661360-85661360 on Build GRCh38
rs748780773 Chr.2: 85661448-85661448 on Build GRCh38
rs1035064024 Chr.2: 85661662-85661662 on Build GRCh38
rs778715336 Chr.2: 85661546-85661546 on Build GRCh38
rs536926847 Chr.2: 85661714-85661714 on Build GRCh38

View Full Product Details