Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95106331-95106826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS5
Entrez Gene ID
64969
Gene Name
mitochondrial ribosomal protein S5
Gene Aliases
MRP-S5, S5mt
UniGene
Hs.655259
Gene Chromosome Location
Chr.2: 95085391-95122027 on Build GRCh38
Transcripts
NM_001321995.1 NM_001321996.1 NM_001321997.1 NM_031902.4 NR_136152.1 NR_136153.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7194123 c.663G>T p.R221S Chr.2: 95106432-95106432 on Build GRCh38
5911690 c.638-8C>T p.? Chr.2: 95106465-95106465 on Build GRCh38
7156812 c.646G>C p.E216Q Chr.2: 95106449-95106449 on Build GRCh38
7288087 c.639A>G p.E213E Chr.2: 95106456-95106456 on Build GRCh38
3583968 c.672G>A p.E224E Chr.2: 95106423-95106423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774918202 Chr.2: 95106484-95106484 on Build GRCh38
rs1038216592 Chr.2: 95106787-95106787 on Build GRCh38
rs370602866 Chr.2: 95106463-95106463 on Build GRCh38
rs1022298048 Chr.2: 95106661-95106661 on Build GRCh38
rs373819000 Chr.2: 95106385-95106385 on Build GRCh38
rs966773856 Chr.2: 95106714-95106714 on Build GRCh38
rs759425834 Chr.2: 95106497-95106497 on Build GRCh38
rs191979634 Chr.2: 95106795-95106795 on Build GRCh38
rs534428445 Chr.2: 95106597-95106597 on Build GRCh38
rs368393219 Chr.2: 95106471-95106471 on Build GRCh38
rs74517367 Chr.2: 95106393-95106393 on Build GRCh38
rs561251957 Chr.2: 95106790-95106790 on Build GRCh38
rs978181793 Chr.2: 95106725-95106725 on Build GRCh38
rs567995898 Chr.2: 95106450-95106450 on Build GRCh38
rs200920483 Chr.2: 95106464-95106464 on Build GRCh38
rs780792043 Chr.2: 95106450-95106451 on Build GRCh38
rs577437335 Chr.2: 95106673-95106673 on Build GRCh38
rs573602917 Chr.2: 95106551-95106551 on Build GRCh38
rs922603857 Chr.2: 95106758-95106758 on Build GRCh38
rs750631445 Chr.2: 95106363-95106363 on Build GRCh38
rs941233838 Chr.2: 95106490-95106490 on Build GRCh38
rs764799726 Chr.2: 95106381-95106381 on Build GRCh38
rs763938067 Chr.2: 95106488-95106488 on Build GRCh38
rs748237624 Chr.2: 95106446-95106446 on Build GRCh38
rs375374542 Chr.2: 95106505-95106505 on Build GRCh38
rs751459942 Chr.2: 95106494-95106494 on Build GRCh38
rs746624146 Chr.2: 95106394-95106395 on Build GRCh38
rs755084785 Chr.2: 95106428-95106428 on Build GRCh38
rs537709591 Chr.2: 95106359-95106359 on Build GRCh38
rs993798161 Chr.2: 95106554-95106554 on Build GRCh38
rs768449968 Chr.2: 95106418-95106429 on Build GRCh38
rs557484218 Chr.2: 95106663-95106663 on Build GRCh38
rs115432097 Chr.2: 95106803-95106803 on Build GRCh38
rs1011091855 Chr.2: 95106639-95106640 on Build GRCh38
rs559494898 Chr.2: 95106802-95106802 on Build GRCh38
rs149256759 Chr.2: 95106354-95106354 on Build GRCh38
rs573898605 Chr.2: 95106791-95106791 on Build GRCh38
rs769015720 Chr.2: 95106650-95106651 on Build GRCh38
rs911156350 Chr.2: 95106768-95106768 on Build GRCh38
rs780272600 Chr.2: 95106422-95106422 on Build GRCh38
rs1038501747 Chr.2: 95106513-95106513 on Build GRCh38
rs199728959 Chr.2: 95106455-95106455 on Build GRCh38
rs754014336 Chr.2: 95106423-95106423 on Build GRCh38
rs758314654 Chr.2: 95106390-95106390 on Build GRCh38
rs751374387 Chr.2: 95106415-95106415 on Build GRCh38
rs933893872 Chr.2: 95106452-95106452 on Build GRCh38
rs747347250 Chr.2: 95106453-95106453 on Build GRCh38
rs765172060 Chr.2: 95106512-95106512 on Build GRCh38
rs747666733 Chr.2: 95106467-95106468 on Build GRCh38
rs766166351 Chr.2: 95106579-95106579 on Build GRCh38
rs754893283 Chr.2: 95106759-95106759 on Build GRCh38
rs200411385 Chr.2: 95106434-95106434 on Build GRCh38
rs78186627 Chr.2: 95106769-95106769 on Build GRCh38
rs377158608 Chr.2: 95106421-95106421 on Build GRCh38
rs747539642 Chr.2: 95106589-95106589 on Build GRCh38
rs572097846 Chr.2: 95106567-95106567 on Build GRCh38
rs778902489 Chr.2: 95106429-95106429 on Build GRCh38
rs535412773 Chr.2: 95106476-95106476 on Build GRCh38
rs778339007 Chr.2: 95106451-95106451 on Build GRCh38
rs985769104 Chr.2: 95106764-95106764 on Build GRCh38
rs941149954 Chr.2: 95106772-95106772 on Build GRCh38
rs929707084 Chr.2: 95106792-95106792 on Build GRCh38
rs751358252 Chr.2: 95106604-95106604 on Build GRCh38
rs959775122 Chr.2: 95106614-95106614 on Build GRCh38

View Full Product Details