Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162142785-162143282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCG
Entrez Gene ID
2641
Gene Name
glucagon
Gene Aliases
GLP1, GLP2, GRPP
UniGene
Hs.516494
Gene Chromosome Location
Chr.2: 162142869-162152404 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

Gene Symbol
LOC101929532
Entrez Gene ID
101929532
Gene Name
uncharacterized LOC101929532
Gene Aliases
-
UniGene
Hs.639883
Gene Chromosome Location
Chr.2: 162114441-162172739 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

SNP Detail

SNP ID Chromosome Location
rs937653356 Chr.2: 162142892-162142892 on Build GRCh38
rs973640383 Chr.2: 162142839-162142839 on Build GRCh38
rs538428277 Chr.2: 162142984-162142984 on Build GRCh38
rs116510735 Chr.2: 162143218-162143218 on Build GRCh38
rs929143625 Chr.2: 162142949-162142949 on Build GRCh38
rs11552332 Chr.2: 162142918-162142918 on Build GRCh38
rs963358444 Chr.2: 162142988-162142988 on Build GRCh38
rs560950841 Chr.2: 162143203-162143203 on Build GRCh38
rs912025376 Chr.2: 162143087-162143087 on Build GRCh38
rs912071397 Chr.2: 162142875-162142875 on Build GRCh38
rs1044742162 Chr.2: 162143150-162143150 on Build GRCh38
rs1017335173 Chr.2: 162142824-162142824 on Build GRCh38
rs542661715 Chr.2: 162143195-162143195 on Build GRCh38
rs766905798 Chr.2: 162143124-162143124 on Build GRCh38
rs979103389 Chr.2: 162142884-162142884 on Build GRCh38
rs142104367 Chr.2: 162142967-162142967 on Build GRCh38
rs188487658 Chr.2: 162143085-162143085 on Build GRCh38
rs149201782 Chr.2: 162142862-162142862 on Build GRCh38
rs773583775 Chr.2: 162142932-162142932 on Build GRCh38
rs560191939 Chr.2: 162143153-162143153 on Build GRCh38
rs922171542 Chr.2: 162143021-162143021 on Build GRCh38
rs11552335 Chr.2: 162143234-162143234 on Build GRCh38
rs374121111 Chr.2: 162143002-162143002 on Build GRCh38
rs555967139 Chr.2: 162142974-162142974 on Build GRCh38
rs11552336 Chr.2: 162143112-162143112 on Build GRCh38
rs11552339 Chr.2: 162143120-162143120 on Build GRCh38
rs533941455 Chr.2: 162142876-162142876 on Build GRCh38
rs1003367897 Chr.2: 162142931-162142931 on Build GRCh38
rs1009721040 Chr.2: 162143163-162143163 on Build GRCh38
rs915015992 Chr.2: 162143113-162143113 on Build GRCh38
rs778875424 Chr.2: 162143030-162143030 on Build GRCh38
rs368278814 Chr.2: 162143038-162143038 on Build GRCh38
rs964195336 Chr.2: 162142844-162142844 on Build GRCh38
rs571980614 Chr.2: 162143154-162143154 on Build GRCh38
rs893444035 Chr.2: 162142880-162142880 on Build GRCh38
rs935525522 Chr.2: 162143108-162143108 on Build GRCh38
rs754344538 Chr.2: 162143192-162143192 on Build GRCh38
rs980095348 Chr.2: 162143228-162143228 on Build GRCh38
rs183105157 Chr.2: 162142917-162142917 on Build GRCh38
rs11552331 Chr.2: 162143230-162143230 on Build GRCh38
rs548912528 Chr.2: 162142856-162142856 on Build GRCh38
rs543831356 Chr.2: 162143024-162143024 on Build GRCh38
rs78895539 Chr.2: 162143254-162143254 on Build GRCh38
rs917646890 Chr.2: 162142920-162142920 on Build GRCh38
rs953723048 Chr.2: 162143084-162143084 on Build GRCh38
rs1035230366 Chr.2: 162142934-162142934 on Build GRCh38
rs6732914 Chr.2: 162142878-162142878 on Build GRCh38
rs145343576 Chr.2: 162142855-162142855 on Build GRCh38
rs1021073364 Chr.2: 162143198-162143198 on Build GRCh38
rs1056400773 Chr.2: 162143241-162143241 on Build GRCh38
rs996822747 Chr.2: 162143092-162143092 on Build GRCh38
rs6717659 Chr.2: 162142845-162142845 on Build GRCh38
rs926281851 Chr.2: 162142891-162142891 on Build GRCh38
rs1029705764 Chr.2: 162142851-162142851 on Build GRCh38
rs891193500 Chr.2: 162143137-162143137 on Build GRCh38
rs1015046505 Chr.2: 162142983-162142983 on Build GRCh38

View Full Product Details