Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196763438-196763942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C3
Entrez Gene ID
9330
Gene Name
general transcription factor IIIC subunit 3
Gene Aliases
TFIIIC102, TFIIICgamma, TFiiiC2-102
UniGene
Hs.470996
Gene Chromosome Location
Chr.2: 196763032-196799768 on Build GRCh38
Transcripts
NM_012086.4 XM_005246965.4

SNP Detail

SNP ID Chromosome Location
rs1038178104 Chr.2: 196763655-196763655 on Build GRCh38
rs544057380 Chr.2: 196763494-196763494 on Build GRCh38
rs928618701 Chr.2: 196763662-196763662 on Build GRCh38
rs964498002 Chr.2: 196763562-196763562 on Build GRCh38
rs1006932788 Chr.2: 196763783-196763784 on Build GRCh38
rs376729316 Chr.2: 196763602-196763603 on Build GRCh38
rs974558450 Chr.2: 196763813-196763813 on Build GRCh38
rs753677793 Chr.2: 196763505-196763509 on Build GRCh38
rs559917580 Chr.2: 196763721-196763721 on Build GRCh38
rs898212490 Chr.2: 196763803-196763803 on Build GRCh38
rs914614473 Chr.2: 196763714-196763715 on Build GRCh38
rs923185449 Chr.2: 196763571-196763571 on Build GRCh38
rs191020400 Chr.2: 196763874-196763874 on Build GRCh38
rs933207822 Chr.2: 196763595-196763595 on Build GRCh38
rs185769020 Chr.2: 196763739-196763739 on Build GRCh38
rs995697060 Chr.2: 196763812-196763813 on Build GRCh38
rs974672420 Chr.2: 196763566-196763566 on Build GRCh38
rs577533798 Chr.2: 196763521-196763521 on Build GRCh38
rs1574424 Chr.2: 196763491-196763491 on Build GRCh38
rs1043522671 Chr.2: 196763724-196763724 on Build GRCh38
rs966766775 Chr.2: 196763795-196763795 on Build GRCh38
rs914686198 Chr.2: 196763899-196763899 on Build GRCh38
rs562262306 Chr.2: 196763814-196763814 on Build GRCh38
rs1036321277 Chr.2: 196763864-196763864 on Build GRCh38
rs369894879 Chr.2: 196763886-196763886 on Build GRCh38
rs549263835 Chr.2: 196763763-196763763 on Build GRCh38
rs1018034223 Chr.2: 196763792-196763792 on Build GRCh38
rs77205562 Chr.2: 196763540-196763540 on Build GRCh38
rs1010882306 Chr.2: 196763760-196763760 on Build GRCh38
rs1013244274 Chr.2: 196763477-196763477 on Build GRCh38
rs890800214 Chr.2: 196763780-196763780 on Build GRCh38
rs564051456 Chr.2: 196763507-196763507 on Build GRCh38
rs1018400802 Chr.2: 196763544-196763544 on Build GRCh38
rs907897708 Chr.2: 196763868-196763868 on Build GRCh38
rs113063229 Chr.2: 196763613-196763613 on Build GRCh38
rs964550555 Chr.2: 196763807-196763807 on Build GRCh38
rs555401286 Chr.2: 196763701-196763703 on Build GRCh38
rs758510291 Chr.2: 196763813-196763816 on Build GRCh38
rs778844711 Chr.2: 196763585-196763586 on Build GRCh38
rs962918417 Chr.2: 196763584-196763584 on Build GRCh38
rs369652558 Chr.2: 196763734-196763734 on Build GRCh38
rs951589018 Chr.2: 196763625-196763625 on Build GRCh38
rs974207146 Chr.2: 196763603-196763603 on Build GRCh38
rs960399657 Chr.2: 196763470-196763470 on Build GRCh38
rs781461448 Chr.2: 196763745-196763745 on Build GRCh38
rs907841872 Chr.2: 196763609-196763609 on Build GRCh38
rs528965482 Chr.2: 196763722-196763722 on Build GRCh38
rs1032051185 Chr.2: 196763802-196763802 on Build GRCh38
rs981467976 Chr.2: 196763659-196763659 on Build GRCh38
rs991875765 Chr.2: 196763892-196763892 on Build GRCh38
rs1000646201 Chr.2: 196763797-196763797 on Build GRCh38
rs906052939 Chr.2: 196763667-196763667 on Build GRCh38
rs954666200 Chr.2: 196763824-196763824 on Build GRCh38
rs1047723632 Chr.2: 196763607-196763607 on Build GRCh38
rs781450990 Chr.2: 196763784-196763787 on Build GRCh38
rs551254814 Chr.2: 196763848-196763848 on Build GRCh38
rs752006141 Chr.2: 196763679-196763679 on Build GRCh38
rs1050756587 Chr.2: 196763741-196763745 on Build GRCh38
rs758974119 Chr.2: 196763508-196763508 on Build GRCh38
rs534526730 Chr.2: 196763885-196763885 on Build GRCh38
rs535546629 Chr.2: 196763595-196763596 on Build GRCh38
rs531129212 Chr.2: 196763844-196763844 on Build GRCh38
rs146840113 Chr.2: 196763903-196763903 on Build GRCh38
rs371665030 Chr.2: 196763710-196763710 on Build GRCh38

View Full Product Details