Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196836768-196837267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGAP1
Entrez Gene ID
80055
Gene Name
post-GPI attachment to proteins 1
Gene Aliases
Bst1, ISPD3024, MRT42, SPG67
UniGene
Hs.229988
Gene Chromosome Location
Chr.2: 196833004-196926995 on Build GRCh38
Transcripts
NM_001321099.1 NM_001321100.1 NM_024989.3 XM_017004992.1 XM_017004993.1 XM_017004994.1

SNP Detail

SNP ID Chromosome Location
rs992769531 Chr.2: 196837046-196837046 on Build GRCh38
rs753124619 Chr.2: 196836898-196836898 on Build GRCh38
rs571836825 Chr.2: 196836928-196836928 on Build GRCh38
rs781332202 Chr.2: 196837051-196837051 on Build GRCh38
rs371357833 Chr.2: 196837005-196837005 on Build GRCh38
rs917000411 Chr.2: 196837100-196837100 on Build GRCh38
rs142688695 Chr.2: 196837065-196837065 on Build GRCh38
rs773104173 Chr.2: 196836865-196836865 on Build GRCh38
rs750991139 Chr.2: 196836794-196836794 on Build GRCh38
rs935892117 Chr.2: 196837202-196837202 on Build GRCh38
rs534345965 Chr.2: 196837134-196837134 on Build GRCh38
rs562710415 Chr.2: 196836900-196836900 on Build GRCh38
rs111643979 Chr.2: 196836947-196836947 on Build GRCh38
rs377025878 Chr.2: 196837091-196837091 on Build GRCh38
rs763372485 Chr.2: 196836814-196836814 on Build GRCh38
rs1041390574 Chr.2: 196837128-196837128 on Build GRCh38
rs945802129 Chr.2: 196837112-196837112 on Build GRCh38
rs548104756 Chr.2: 196837024-196837024 on Build GRCh38
rs991993301 Chr.2: 196837150-196837150 on Build GRCh38
rs951507218 Chr.2: 196836803-196836803 on Build GRCh38
rs975352187 Chr.2: 196836896-196836896 on Build GRCh38
rs1039624830 Chr.2: 196837220-196837220 on Build GRCh38
rs556546041 Chr.2: 196837116-196837116 on Build GRCh38
rs985097131 Chr.2: 196837002-196837002 on Build GRCh38
rs144541374 Chr.2: 196836813-196836813 on Build GRCh38
rs924922950 Chr.2: 196837243-196837243 on Build GRCh38
rs148493287 Chr.2: 196836855-196836855 on Build GRCh38
rs7602892 Chr.2: 196836980-196836980 on Build GRCh38
rs74851256 Chr.2: 196836923-196836923 on Build GRCh38
rs751787178 Chr.2: 196836977-196836977 on Build GRCh38
rs750028767 Chr.2: 196837064-196837064 on Build GRCh38
rs576378640 Chr.2: 196837181-196837181 on Build GRCh38
rs1019634221 Chr.2: 196836854-196836854 on Build GRCh38
rs947778425 Chr.2: 196837218-196837218 on Build GRCh38
rs536235654 Chr.2: 196837066-196837066 on Build GRCh38
rs752648790 Chr.2: 196837172-196837172 on Build GRCh38

View Full Product Details