Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199928162-199928660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199911256-199928273 on Build GRCh38
Transcripts
NM_153689.5

SNP Detail

SNP ID Chromosome Location
rs953149252 Chr.2: 199928273-199928273 on Build GRCh38
rs927360250 Chr.2: 199928195-199928195 on Build GRCh38
rs1025827793 Chr.2: 199928555-199928555 on Build GRCh38
rs151293001 Chr.2: 199928626-199928626 on Build GRCh38
rs994307828 Chr.2: 199928552-199928553 on Build GRCh38
rs566818423 Chr.2: 199928306-199928306 on Build GRCh38
rs10202253 Chr.2: 199928482-199928482 on Build GRCh38
rs907399929 Chr.2: 199928635-199928635 on Build GRCh38
rs577278200 Chr.2: 199928460-199928462 on Build GRCh38
rs952989197 Chr.2: 199928582-199928583 on Build GRCh38
rs993281367 Chr.2: 199928618-199928618 on Build GRCh38
rs530667397 Chr.2: 199928534-199928534 on Build GRCh38
rs538455126 Chr.2: 199928542-199928542 on Build GRCh38
rs548296882 Chr.2: 199928271-199928271 on Build GRCh38
rs536724183 Chr.2: 199928224-199928224 on Build GRCh38
rs369672187 Chr.2: 199928568-199928568 on Build GRCh38
rs537826842 Chr.2: 199928263-199928263 on Build GRCh38
rs1010594247 Chr.2: 199928213-199928213 on Build GRCh38
rs915976009 Chr.2: 199928517-199928517 on Build GRCh38
rs74511880 Chr.2: 199928503-199928503 on Build GRCh38
rs755259303 Chr.2: 199928574-199928574 on Build GRCh38
rs1045867471 Chr.2: 199928633-199928633 on Build GRCh38
rs534176247 Chr.2: 199928461-199928461 on Build GRCh38
rs919239296 Chr.2: 199928490-199928490 on Build GRCh38
rs148785517 Chr.2: 199928200-199928200 on Build GRCh38
rs1041215872 Chr.2: 199928495-199928495 on Build GRCh38
rs768772208 Chr.2: 199928413-199928413 on Build GRCh38
rs965981768 Chr.2: 199928394-199928394 on Build GRCh38
rs1034796179 Chr.2: 199928604-199928604 on Build GRCh38
rs1042296255 Chr.2: 199928373-199928373 on Build GRCh38
rs77160058 Chr.2: 199928601-199928601 on Build GRCh38
rs545261731 Chr.2: 199928391-199928391 on Build GRCh38
rs574719825 Chr.2: 199928517-199928560 on Build GRCh38
rs911721642 Chr.2: 199928302-199928302 on Build GRCh38
rs780651034 Chr.2: 199928399-199928399 on Build GRCh38
rs1049102615 Chr.2: 199928512-199928512 on Build GRCh38
rs778164598 Chr.2: 199928617-199928617 on Build GRCh38
rs949252286 Chr.2: 199928367-199928367 on Build GRCh38
rs917781585 Chr.2: 199928246-199928246 on Build GRCh38
rs569271080 Chr.2: 199928215-199928215 on Build GRCh38
rs1053473162 Chr.2: 199928461-199928462 on Build GRCh38
rs140210719 Chr.2: 199928618-199928620 on Build GRCh38
rs985941364 Chr.2: 199928293-199928293 on Build GRCh38

View Full Product Details