Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75787415-75787920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTB
Entrez Gene ID
5876
Gene Name
Rab geranylgeranyltransferase beta subunit
Gene Aliases
GGTB
UniGene
Hs.78948
Gene Chromosome Location
Chr.1: 75786194-75795090 on Build GRCh38
Transcripts
NM_004582.3 NR_073562.1 NR_002749.1

Gene Symbol
SNORD45A
Entrez Gene ID
26805
Gene Name
small nucleolar RNA, C/D box 45A
Gene Aliases
RNU45A, U45a
UniGene
Gene Chromosome Location
Chr.1: 75787889-75787972 on Build GRCh38
Transcripts
NM_004582.3 NR_073562.1 NR_002749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6127634 c.11C>G p.P4R Chr.1: 75787504-75787504 on Build GRCh38
5473930 c.71C>T p.A24V Chr.1: 75787564-75787564 on Build GRCh38
6064331 c.5G>T p.G2V Chr.1: 75787498-75787498 on Build GRCh38
3985219 c.25A>G p.I9V Chr.1: 75787518-75787518 on Build GRCh38
5436934 c.27T>C p.I9I Chr.1: 75787520-75787520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936011964 Chr.1: 75787709-75787709 on Build GRCh38
rs531992019 Chr.1: 75787629-75787629 on Build GRCh38
rs778279313 Chr.1: 75787641-75787643 on Build GRCh38
rs761499578 Chr.1: 75787522-75787522 on Build GRCh38
rs1026860500 Chr.1: 75787807-75787807 on Build GRCh38
rs778965079 Chr.1: 75787878-75787878 on Build GRCh38
rs1035763835 Chr.1: 75787781-75787781 on Build GRCh38
rs952596205 Chr.1: 75787823-75787823 on Build GRCh38
rs764694118 Chr.1: 75787501-75787502 on Build GRCh38
rs948818140 Chr.1: 75787731-75787731 on Build GRCh38
rs765751681 Chr.1: 75787628-75787628 on Build GRCh38
rs535778257 Chr.1: 75787720-75787720 on Build GRCh38
rs548043269 Chr.1: 75787761-75787761 on Build GRCh38
rs977351950 Chr.1: 75787699-75787699 on Build GRCh38
rs777256818 Chr.1: 75787532-75787532 on Build GRCh38
rs757349050 Chr.1: 75787569-75787569 on Build GRCh38
rs749246034 Chr.1: 75787518-75787518 on Build GRCh38
rs768491868 Chr.1: 75787582-75787582 on Build GRCh38
rs751831663 Chr.1: 75787566-75787566 on Build GRCh38
rs770958961 Chr.1: 75787507-75787507 on Build GRCh38
rs758251095 Chr.1: 75787894-75787894 on Build GRCh38
rs3737573 Chr.1: 75787544-75787544 on Build GRCh38
rs771875525 Chr.1: 75787524-75787524 on Build GRCh38
rs778000054 Chr.1: 75787563-75787563 on Build GRCh38
rs375650983 Chr.1: 75787574-75787574 on Build GRCh38
rs1045854469 Chr.1: 75787743-75787743 on Build GRCh38
rs751803539 Chr.1: 75787655-75787655 on Build GRCh38
rs550389179 Chr.1: 75787662-75787662 on Build GRCh38
rs111457474 Chr.1: 75787453-75787453 on Build GRCh38
rs747989521 Chr.1: 75787585-75787585 on Build GRCh38
rs780979051 Chr.1: 75787791-75787791 on Build GRCh38
rs756642610 Chr.1: 75787455-75787455 on Build GRCh38
rs915961500 Chr.1: 75787726-75787726 on Build GRCh38
rs200688483 Chr.1: 75787467-75787467 on Build GRCh38
rs754340395 Chr.1: 75787468-75787468 on Build GRCh38
rs755346089 Chr.1: 75787477-75787477 on Build GRCh38
rs908307300 Chr.1: 75787530-75787530 on Build GRCh38
rs750495825 Chr.1: 75787857-75787857 on Build GRCh38
rs748364806 Chr.1: 75787482-75787482 on Build GRCh38
rs753070718 Chr.1: 75787550-75787550 on Build GRCh38
rs772035160 Chr.1: 75787591-75787591 on Build GRCh38
rs958991712 Chr.1: 75787790-75787790 on Build GRCh38
rs760301683 Chr.1: 75787613-75787613 on Build GRCh38
rs774293237 Chr.1: 75787521-75787521 on Build GRCh38
rs749186509 Chr.1: 75787575-75787575 on Build GRCh38
rs781280076 Chr.1: 75787853-75787853 on Build GRCh38
rs778803339 Chr.1: 75787584-75787584 on Build GRCh38
rs11548708 Chr.1: 75787523-75787523 on Build GRCh38
rs758563844 Chr.1: 75787551-75787551 on Build GRCh38
rs763283861 Chr.1: 75787642-75787642 on Build GRCh38
rs991445068 Chr.1: 75787803-75787803 on Build GRCh38
rs760200189 Chr.1: 75787534-75787534 on Build GRCh38
rs764502836 Chr.1: 75787654-75787654 on Build GRCh38
rs754907536 Chr.1: 75787876-75787876 on Build GRCh38
rs542064322 Chr.1: 75787502-75787502 on Build GRCh38
rs779203303 Chr.1: 75787480-75787480 on Build GRCh38
rs559894863 Chr.1: 75787505-75787505 on Build GRCh38
rs1020432695 Chr.1: 75787693-75787693 on Build GRCh38
rs376211827 Chr.1: 75787890-75787890 on Build GRCh38
rs746628957 Chr.1: 75787600-75787600 on Build GRCh38
rs565954469 Chr.1: 75787765-75787765 on Build GRCh38
rs1003391502 Chr.1: 75787707-75787707 on Build GRCh38
rs371859962 Chr.1: 75787622-75787622 on Build GRCh38
rs770650468 Chr.1: 75787669-75787669 on Build GRCh38
rs770430958 Chr.1: 75787614-75787615 on Build GRCh38
rs768497645 Chr.1: 75787520-75787520 on Build GRCh38
rs368380945 Chr.1: 75787597-75787597 on Build GRCh38
rs1050700948 Chr.1: 75787653-75787653 on Build GRCh38
rs775478491 Chr.1: 75787511-75787511 on Build GRCh38
rs374947966 Chr.1: 75787710-75787710 on Build GRCh38
rs903283297 Chr.1: 75787695-75787695 on Build GRCh38
rs552337803 Chr.1: 75787537-75787537 on Build GRCh38
rs757425334 Chr.1: 75787852-75787852 on Build GRCh38
rs766943423 Chr.1: 75787462-75787462 on Build GRCh38
rs151093467 Chr.1: 75787696-75787696 on Build GRCh38
rs765855046 Chr.1: 75787536-75787536 on Build GRCh38
rs183981518 Chr.1: 75787533-75787533 on Build GRCh38
rs770470786 Chr.1: 75787621-75787621 on Build GRCh38
rs931185212 Chr.1: 75787463-75787463 on Build GRCh38
rs760027784 Chr.1: 75787543-75787543 on Build GRCh38
rs745588647 Chr.1: 75787642-75787644 on Build GRCh38
rs777774244 Chr.1: 75787895-75787895 on Build GRCh38
rs372853318 Chr.1: 75787484-75787484 on Build GRCh38
rs994945395 Chr.1: 75787579-75787579 on Build GRCh38

View Full Product Details