Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5184617-5185099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS2
Entrez Gene ID
8760
Gene Name
CDP-diacylglycerol synthase 2
Gene Aliases
-
UniGene
Hs.126857
Gene Chromosome Location
Chr.20: 5126761-5197887 on Build GRCh38
Transcripts
NM_003818.3 XM_006723660.1 XM_011529390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467754 c.735C>G p.F245L Chr.20: 5184921-5184921 on Build GRCh38
1483763 c.753C>A p.L251L Chr.20: 5184939-5184939 on Build GRCh38
2763366 c.673T>C p.F225L Chr.20: 5184859-5184859 on Build GRCh38
1200628 c.742C>T p.R248W Chr.20: 5184928-5184928 on Build GRCh38
3547829 c.756C>T p.I252I Chr.20: 5184942-5184942 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752517851 Chr.20: 5184963-5184963 on Build GRCh38
rs143175887 Chr.20: 5184929-5184929 on Build GRCh38
rs373552904 Chr.20: 5184846-5184846 on Build GRCh38
rs541733288 Chr.20: 5184646-5184646 on Build GRCh38
rs746073089 Chr.20: 5184817-5184817 on Build GRCh38
rs763517359 Chr.20: 5184680-5184680 on Build GRCh38
rs961610340 Chr.20: 5184802-5184802 on Build GRCh38
rs563196559 Chr.20: 5184761-5184761 on Build GRCh38
rs112055575 Chr.20: 5184874-5184874 on Build GRCh38
rs368700421 Chr.20: 5184933-5184933 on Build GRCh38
rs780405994 Chr.20: 5184815-5184815 on Build GRCh38
rs750135259 Chr.20: 5184930-5184930 on Build GRCh38
rs761393933 Chr.20: 5184917-5184917 on Build GRCh38
rs752048640 Chr.20: 5184681-5184681 on Build GRCh38
rs375314789 Chr.20: 5184785-5184785 on Build GRCh38
rs541950186 Chr.20: 5184747-5184747 on Build GRCh38
rs369736484 Chr.20: 5185041-5185041 on Build GRCh38
rs891167906 Chr.20: 5184731-5184731 on Build GRCh38
rs201506899 Chr.20: 5184879-5184879 on Build GRCh38
rs959630049 Chr.20: 5184899-5184899 on Build GRCh38
rs914456370 Chr.20: 5185066-5185066 on Build GRCh38
rs759139026 Chr.20: 5184981-5184982 on Build GRCh38
rs767953137 Chr.20: 5184708-5184708 on Build GRCh38
rs148097249 Chr.20: 5185042-5185042 on Build GRCh38
rs995490554 Chr.20: 5184789-5184789 on Build GRCh38
rs542885743 Chr.20: 5184642-5184642 on Build GRCh38
rs754070730 Chr.20: 5184989-5184989 on Build GRCh38
rs769231474 Chr.20: 5184900-5184900 on Build GRCh38
rs200494742 Chr.20: 5184861-5184861 on Build GRCh38
rs373780357 Chr.20: 5184950-5184950 on Build GRCh38
rs1027169749 Chr.20: 5184896-5184896 on Build GRCh38
rs201236627 Chr.20: 5185021-5185024 on Build GRCh38
rs55695350 Chr.20: 5185001-5185001 on Build GRCh38
rs900898281 Chr.20: 5184711-5184711 on Build GRCh38
rs770346753 Chr.20: 5184850-5184850 on Build GRCh38
rs6116681 Chr.20: 5184686-5184686 on Build GRCh38
rs745821974 Chr.20: 5184716-5184717 on Build GRCh38
rs183825567 Chr.20: 5184740-5184740 on Build GRCh38
rs997895486 Chr.20: 5184711-5184714 on Build GRCh38
rs149118554 Chr.20: 5184915-5184915 on Build GRCh38
rs757413433 Chr.20: 5184973-5184973 on Build GRCh38
rs6116682 Chr.20: 5184857-5184857 on Build GRCh38
rs989826613 Chr.20: 5185056-5185056 on Build GRCh38
rs763423288 Chr.20: 5184898-5184898 on Build GRCh38
rs1016834783 Chr.20: 5184752-5184752 on Build GRCh38
rs370921980 Chr.20: 5184958-5184958 on Build GRCh38
rs953875841 Chr.20: 5184743-5184743 on Build GRCh38
rs757542094 Chr.20: 5184694-5184694 on Build GRCh38
rs753376234 Chr.20: 5184904-5184905 on Build GRCh38
rs894070843 Chr.20: 5184672-5184672 on Build GRCh38
rs775961953 Chr.20: 5184856-5184856 on Build GRCh38
rs147647726 Chr.20: 5184696-5184696 on Build GRCh38
rs915464945 Chr.20: 5185070-5185070 on Build GRCh38
rs781413715 Chr.20: 5184983-5184983 on Build GRCh38
rs1009210720 Chr.20: 5184676-5184676 on Build GRCh38
rs1054353343 Chr.20: 5184671-5184671 on Build GRCh38
rs778913142 Chr.20: 5184959-5184959 on Build GRCh38

View Full Product Details