Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33055341-33055837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB3
Entrez Gene ID
359710
Gene Name
BPI fold containing family B member 3
Gene Aliases
C20orf185, LPLUNC3, RYA3
UniGene
Hs.472442
Gene Chromosome Location
Chr.20: 33055331-33073637 on Build GRCh38
Transcripts
NM_182658.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751390 c.67G>C p.E23Q Chr.20: 33055490-33055490 on Build GRCh38
7115503 c.42C>G p.L14L Chr.20: 33055465-33055465 on Build GRCh38
3911112 c.60A>G p.P20P Chr.20: 33055483-33055483 on Build GRCh38
7080097 c.58C>G p.P20A Chr.20: 33055481-33055481 on Build GRCh38
4752146 c.61T>C p.C21R Chr.20: 33055484-33055484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958116535 Chr.20: 33055691-33055691 on Build GRCh38
rs748321687 Chr.20: 33055445-33055445 on Build GRCh38
rs756756019 Chr.20: 33055410-33055410 on Build GRCh38
rs373640490 Chr.20: 33055400-33055400 on Build GRCh38
rs906889764 Chr.20: 33055585-33055585 on Build GRCh38
rs777365752 Chr.20: 33055402-33055402 on Build GRCh38
rs146768085 Chr.20: 33055635-33055635 on Build GRCh38
rs55823624 Chr.20: 33055549-33055549 on Build GRCh38
rs73262145 Chr.20: 33055364-33055364 on Build GRCh38
rs765063499 Chr.20: 33055481-33055481 on Build GRCh38
rs755503095 Chr.20: 33055508-33055508 on Build GRCh38
rs578055505 Chr.20: 33055403-33055403 on Build GRCh38
rs760971856 Chr.20: 33055644-33055644 on Build GRCh38
rs144150108 Chr.20: 33055516-33055516 on Build GRCh38
rs906832050 Chr.20: 33055784-33055784 on Build GRCh38
rs749815380 Chr.20: 33055434-33055434 on Build GRCh38
rs779209065 Chr.20: 33055512-33055512 on Build GRCh38
rs953419167 Chr.20: 33055712-33055712 on Build GRCh38
rs1013515136 Chr.20: 33055615-33055615 on Build GRCh38
rs138919082 Chr.20: 33055517-33055517 on Build GRCh38
rs762653879 Chr.20: 33055484-33055484 on Build GRCh38
rs542805953 Chr.20: 33055520-33055520 on Build GRCh38
rs760435672 Chr.20: 33055385-33055387 on Build GRCh38
rs985746190 Chr.20: 33055730-33055730 on Build GRCh38
rs375833464 Chr.20: 33055453-33055453 on Build GRCh38
rs759495083 Chr.20: 33055529-33055529 on Build GRCh38
rs766429713 Chr.20: 33055470-33055470 on Build GRCh38
rs969765030 Chr.20: 33055479-33055479 on Build GRCh38
rs778006952 Chr.20: 33055593-33055593 on Build GRCh38
rs765188284 Chr.20: 33055373-33055373 on Build GRCh38
rs929707268 Chr.20: 33055774-33055774 on Build GRCh38
rs756777068 Chr.20: 33055503-33055503 on Build GRCh38
rs550261954 Chr.20: 33055548-33055548 on Build GRCh38
rs147346398 Chr.20: 33055531-33055531 on Build GRCh38
rs942385272 Chr.20: 33055525-33055525 on Build GRCh38
rs967442930 Chr.20: 33055682-33055682 on Build GRCh38
rs138045241 Chr.20: 33055437-33055437 on Build GRCh38
rs140231383 Chr.20: 33055760-33055760 on Build GRCh38
rs985511045 Chr.20: 33055674-33055674 on Build GRCh38
rs752327527 Chr.20: 33055381-33055381 on Build GRCh38
rs763851993 Chr.20: 33055540-33055540 on Build GRCh38
rs202172895 Chr.20: 33055436-33055436 on Build GRCh38
rs751218887 Chr.20: 33055486-33055486 on Build GRCh38
rs191042825 Chr.20: 33055372-33055372 on Build GRCh38
rs771077411 Chr.20: 33055597-33055597 on Build GRCh38
rs751218546 Chr.20: 33055541-33055541 on Build GRCh38
rs774712864 Chr.20: 33055444-33055444 on Build GRCh38
rs540581621 Chr.20: 33055768-33055768 on Build GRCh38
rs11907401 Chr.20: 33055594-33055594 on Build GRCh38
rs6057713 Chr.20: 33055415-33055415 on Build GRCh38
rs995185170 Chr.20: 33055562-33055562 on Build GRCh38
rs149497264 Chr.20: 33055504-33055504 on Build GRCh38
rs1034617011 Chr.20: 33055601-33055601 on Build GRCh38
rs384430 Chr.20: 33055705-33055705 on Build GRCh38
rs143492136 Chr.20: 33055625-33055625 on Build GRCh38
rs758760950 Chr.20: 33055586-33055586 on Build GRCh38
rs773351282 Chr.20: 33055449-33055449 on Build GRCh38
rs1045473326 Chr.20: 33055782-33055782 on Build GRCh38
rs560586783 Chr.20: 33055483-33055483 on Build GRCh38
rs779374751 Chr.20: 33055580-33055580 on Build GRCh38
rs755662552 Chr.20: 33055572-33055572 on Build GRCh38
rs911143131 Chr.20: 33055749-33055749 on Build GRCh38
rs749743501 Chr.20: 33055458-33055458 on Build GRCh38
rs749967390 Chr.20: 33055561-33055561 on Build GRCh38
rs867524573 Chr.20: 33055771-33055771 on Build GRCh38
rs538332283 Chr.20: 33055368-33055368 on Build GRCh38
rs780586792 Chr.20: 33055418-33055418 on Build GRCh38
rs368960556 Chr.20: 33055533-33055533 on Build GRCh38
rs879488719 Chr.20: 33055706-33055706 on Build GRCh38
rs748485240 Chr.20: 33055513-33055513 on Build GRCh38
rs766849047 Chr.20: 33055388-33055389 on Build GRCh38
rs561454593 Chr.20: 33055521-33055521 on Build GRCh38
rs1024991233 Chr.20: 33055471-33055471 on Build GRCh38
rs190793150 Chr.20: 33055584-33055584 on Build GRCh38
rs910793540 Chr.20: 33055524-33055524 on Build GRCh38
rs369950926 Chr.20: 33055485-33055485 on Build GRCh38
rs759548237 Chr.20: 33055477-33055477 on Build GRCh38
rs776624763 Chr.20: 33055472-33055472 on Build GRCh38
rs764969187 Chr.20: 33055660-33055660 on Build GRCh38
rs976188920 Chr.20: 33055700-33055700 on Build GRCh38
rs567463622 Chr.20: 33055731-33055731 on Build GRCh38

View Full Product Details