Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49987766-49988259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAI1
Entrez Gene ID
6615
Gene Name
snail family transcriptional repressor 1
Gene Aliases
SLUGH2, SNA, SNAH, SNAIL, SNAIL1, dJ710H13.1
UniGene
Hs.48029
Gene Chromosome Location
Chr.20: 49982976-49988886 on Build GRCh38
Transcripts
NM_005985.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5598800 c.737C>T p.S246F Chr.20: 49987998-49987998 on Build GRCh38
4970704 c.652G>T p.A218S Chr.20: 49987913-49987913 on Build GRCh38
294839 c.727C>T p.R243W Chr.20: 49987988-49987988 on Build GRCh38
4890159 c.686C>A p.T229N Chr.20: 49987947-49987947 on Build GRCh38
1565724 c.613G>A p.E205K Chr.20: 49987874-49987874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949900582 Chr.20: 49987793-49987793 on Build GRCh38
rs766201407 Chr.20: 49987935-49987935 on Build GRCh38
rs140860809 Chr.20: 49988051-49988051 on Build GRCh38
rs768825298 Chr.20: 49987905-49987905 on Build GRCh38
rs573703613 Chr.20: 49987960-49987960 on Build GRCh38
rs751497172 Chr.20: 49987864-49987865 on Build GRCh38
rs763916344 Chr.20: 49987851-49987851 on Build GRCh38
rs567405771 Chr.20: 49988149-49988149 on Build GRCh38
rs566097649 Chr.20: 49988215-49988215 on Build GRCh38
rs752241335 Chr.20: 49987972-49987972 on Build GRCh38
rs756823401 Chr.20: 49988011-49988011 on Build GRCh38
rs377622092 Chr.20: 49987980-49987980 on Build GRCh38
rs142763927 Chr.20: 49987981-49987981 on Build GRCh38
rs144872502 Chr.20: 49988058-49988058 on Build GRCh38
rs780507847 Chr.20: 49988098-49988098 on Build GRCh38
rs750886158 Chr.20: 49988088-49988088 on Build GRCh38
rs751527363 Chr.20: 49987835-49987835 on Build GRCh38
rs747141400 Chr.20: 49988045-49988045 on Build GRCh38
rs919594313 Chr.20: 49988203-49988205 on Build GRCh38
rs761792854 Chr.20: 49987882-49987884 on Build GRCh38
rs201293282 Chr.20: 49987866-49987866 on Build GRCh38
rs776897898 Chr.20: 49987912-49987912 on Build GRCh38
rs926033898 Chr.20: 49987800-49987800 on Build GRCh38
rs778678794 Chr.20: 49988086-49988086 on Build GRCh38
rs375631988 Chr.20: 49987865-49987866 on Build GRCh38
rs200570900 Chr.20: 49987873-49987873 on Build GRCh38
rs151036730 Chr.20: 49988033-49988033 on Build GRCh38
rs374439062 Chr.20: 49988062-49988062 on Build GRCh38
rs375610941 Chr.20: 49987942-49987942 on Build GRCh38
rs539872877 Chr.20: 49988235-49988235 on Build GRCh38
rs939787304 Chr.20: 49988158-49988158 on Build GRCh38
rs865912088 Chr.20: 49987887-49987887 on Build GRCh38
rs902610951 Chr.20: 49988052-49988052 on Build GRCh38
rs767077334 Chr.20: 49988061-49988062 on Build GRCh38
rs749868936 Chr.20: 49988063-49988064 on Build GRCh38
rs558459348 Chr.20: 49987828-49987828 on Build GRCh38
rs111475890 Chr.20: 49987894-49987894 on Build GRCh38
rs372650135 Chr.20: 49987827-49987827 on Build GRCh38
rs775763372 Chr.20: 49987886-49987886 on Build GRCh38
rs1038912279 Chr.20: 49988049-49988049 on Build GRCh38
rs763694879 Chr.20: 49988083-49988083 on Build GRCh38
rs755809892 Chr.20: 49987847-49987847 on Build GRCh38
rs753536560 Chr.20: 49988001-49988001 on Build GRCh38
rs747604296 Chr.20: 49987838-49987838 on Build GRCh38
rs1049948768 Chr.20: 49988233-49988233 on Build GRCh38
rs767506905 Chr.20: 49988076-49988076 on Build GRCh38
rs891291611 Chr.20: 49987913-49987913 on Build GRCh38
rs753239373 Chr.20: 49988084-49988084 on Build GRCh38
rs779357818 Chr.20: 49988025-49988025 on Build GRCh38
rs974111593 Chr.20: 49988187-49988187 on Build GRCh38
rs41283590 Chr.20: 49988214-49988214 on Build GRCh38
rs371056563 Chr.20: 49987858-49987858 on Build GRCh38
rs752647667 Chr.20: 49988078-49988078 on Build GRCh38
rs375525704 Chr.20: 49987821-49987821 on Build GRCh38
rs932356999 Chr.20: 49988221-49988221 on Build GRCh38
rs759292518 Chr.20: 49988070-49988070 on Build GRCh38
rs761883004 Chr.20: 49987928-49987928 on Build GRCh38
rs759488579 Chr.20: 49987944-49987944 on Build GRCh38
rs760465365 Chr.20: 49988082-49988082 on Build GRCh38
rs762618438 Chr.20: 49987821-49987822 on Build GRCh38
rs138561934 Chr.20: 49987819-49987819 on Build GRCh38
rs750819893 Chr.20: 49987792-49987792 on Build GRCh38
rs530704361 Chr.20: 49988093-49988093 on Build GRCh38
rs771343871 Chr.20: 49987864-49987864 on Build GRCh38
rs200686434 Chr.20: 49987869-49987869 on Build GRCh38
rs902390520 Chr.20: 49988090-49988090 on Build GRCh38
rs112146891 Chr.20: 49988059-49988059 on Build GRCh38
rs548951484 Chr.20: 49988111-49988111 on Build GRCh38
rs747521619 Chr.20: 49988102-49988102 on Build GRCh38
rs529375612 Chr.20: 49988154-49988154 on Build GRCh38
rs756983850 Chr.20: 49988085-49988085 on Build GRCh38
rs757770152 Chr.20: 49988173-49988173 on Build GRCh38
rs752688108 Chr.20: 49988213-49988213 on Build GRCh38
rs751034214 Chr.20: 49987863-49987863 on Build GRCh38
rs760808201 Chr.20: 49987904-49987904 on Build GRCh38
rs746213399 Chr.20: 49988032-49988032 on Build GRCh38
rs371503562 Chr.20: 49988061-49988061 on Build GRCh38
rs780704231 Chr.20: 49988044-49988044 on Build GRCh38
rs376908483 Chr.20: 49987859-49987859 on Build GRCh38
rs755775431 Chr.20: 49988075-49988102 on Build GRCh38
rs369890190 Chr.20: 49987837-49987837 on Build GRCh38

View Full Product Details