Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 53994653-53995149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS1
Entrez Gene ID
8537
Gene Name
breast carcinoma amplified sequence 1
Gene Aliases
AIBC1, NABC1
UniGene
Hs.400556
Gene Chromosome Location
Chr.20: 53943538-54070765 on Build GRCh38
Transcripts
NM_001316361.2 NM_001323347.1 NM_003657.3 XM_005260587.2 XM_005260589.2 XM_005260590.2 XM_005260591.2 XM_005260593.2 XM_005260594.2 XM_005260595.2 XM_011529090.2 XM_011529091.2 XM_017028110.1 XM_017028111.1 XM_017028112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972615 c.911A>G p.K304R Chr.20: 53995028-53995028 on Build GRCh38
6610454 c.914A>G p.D305G Chr.20: 53995025-53995025 on Build GRCh38
5057429 c.927G>A p.T309T Chr.20: 53995012-53995012 on Build GRCh38
3972616 c.911A>G p.K304R Chr.20: 53995028-53995028 on Build GRCh38
6610455 c.914A>G p.D305G Chr.20: 53995025-53995025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780347718 Chr.20: 53994896-53994897 on Build GRCh38
rs973172107 Chr.20: 53995012-53995012 on Build GRCh38
rs972691991 Chr.20: 53994791-53994791 on Build GRCh38
rs904136067 Chr.20: 53994920-53994920 on Build GRCh38
rs1001030816 Chr.20: 53994931-53994931 on Build GRCh38
rs925351050 Chr.20: 53994913-53994918 on Build GRCh38
rs959481855 Chr.20: 53994722-53994722 on Build GRCh38
rs2276494 Chr.20: 53994683-53994683 on Build GRCh38
rs542484047 Chr.20: 53995064-53995064 on Build GRCh38
rs878879819 Chr.20: 53994889-53994890 on Build GRCh38
rs765710552 Chr.20: 53994916-53994917 on Build GRCh38
rs75411871 Chr.20: 53994853-53994853 on Build GRCh38
rs146234679 Chr.20: 53994889-53994896 on Build GRCh38
rs73124075 Chr.20: 53994918-53994918 on Build GRCh38
rs992559772 Chr.20: 53994746-53994746 on Build GRCh38
rs753229902 Chr.20: 53995106-53995106 on Build GRCh38
rs1000924696 Chr.20: 53994708-53994708 on Build GRCh38
rs368686287 Chr.20: 53994971-53994971 on Build GRCh38
rs776038525 Chr.20: 53994982-53994985 on Build GRCh38
rs11471603 Chr.20: 53994888-53994889 on Build GRCh38
rs772485460 Chr.20: 53994893-53994894 on Build GRCh38
rs781615053 Chr.20: 53994966-53994966 on Build GRCh38
rs762830957 Chr.20: 53995065-53995065 on Build GRCh38
rs963112703 Chr.20: 53994764-53994764 on Build GRCh38
rs943826212 Chr.20: 53994884-53994884 on Build GRCh38
rs753820049 Chr.20: 53994911-53994918 on Build GRCh38
rs2276495 Chr.20: 53994677-53994677 on Build GRCh38
rs1033745330 Chr.20: 53994718-53994718 on Build GRCh38
rs768578063 Chr.20: 53994915-53994916 on Build GRCh38
rs796762581 Chr.20: 53994917-53994918 on Build GRCh38
rs560642219 Chr.20: 53994760-53994760 on Build GRCh38
rs6127047 Chr.20: 53994692-53994692 on Build GRCh38
rs769609337 Chr.20: 53995061-53995061 on Build GRCh38
rs532952471 Chr.20: 53994901-53994902 on Build GRCh38
rs772280595 Chr.20: 53995007-53995007 on Build GRCh38
rs368323547 Chr.20: 53994915-53994918 on Build GRCh38
rs778052336 Chr.20: 53994990-53994990 on Build GRCh38
rs917932929 Chr.20: 53994777-53994777 on Build GRCh38
rs759045282 Chr.20: 53995100-53995100 on Build GRCh38
rs208381 Chr.20: 53995001-53995001 on Build GRCh38
rs45513598 Chr.20: 53994894-53994894 on Build GRCh38
rs1028915842 Chr.20: 53994789-53994789 on Build GRCh38
rs985252577 Chr.20: 53995056-53995056 on Build GRCh38
rs745457917 Chr.20: 53994778-53994778 on Build GRCh38
rs771234134 Chr.20: 53995000-53995000 on Build GRCh38
rs765960431 Chr.20: 53995083-53995083 on Build GRCh38
rs757785721 Chr.20: 53994807-53994807 on Build GRCh38
rs952671072 Chr.20: 53995054-53995054 on Build GRCh38
rs553948567 Chr.20: 53994928-53994928 on Build GRCh38
rs60878965 Chr.20: 53995013-53995013 on Build GRCh38
rs756615304 Chr.20: 53995107-53995107 on Build GRCh38
rs531594669 Chr.20: 53994868-53994868 on Build GRCh38
rs780340194 Chr.20: 53994948-53994949 on Build GRCh38
rs745883165 Chr.20: 53995002-53995002 on Build GRCh38
rs377522767 Chr.20: 53994972-53994972 on Build GRCh38
rs987314418 Chr.20: 53994844-53994844 on Build GRCh38
rs936693397 Chr.20: 53994913-53994914 on Build GRCh38
rs943749051 Chr.20: 53995111-53995111 on Build GRCh38
rs751220117 Chr.20: 53995090-53995090 on Build GRCh38
rs761713493 Chr.20: 53995047-53995047 on Build GRCh38
rs548229851 Chr.20: 53994882-53994882 on Build GRCh38
rs767075550 Chr.20: 53995103-53995103 on Build GRCh38
rs147415946 Chr.20: 53994733-53994733 on Build GRCh38
rs867852297 Chr.20: 53995052-53995052 on Build GRCh38
rs1010211479 Chr.20: 53994917-53994917 on Build GRCh38
rs968226635 Chr.20: 53994699-53994699 on Build GRCh38
rs78899533 Chr.20: 53994910-53994917 on Build GRCh38
rs539646642 Chr.20: 53994995-53994995 on Build GRCh38
rs1033881190 Chr.20: 53994961-53994961 on Build GRCh38
rs552346730 Chr.20: 53994835-53994835 on Build GRCh38
rs891463732 Chr.20: 53994906-53994906 on Build GRCh38
rs919729857 Chr.20: 53994827-53994827 on Build GRCh38

View Full Product Details