Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32316374-32316875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
URB1
Entrez Gene ID
9875
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Aliases
C21orf108, NPA1
UniGene
Hs.473611
Gene Chromosome Location
Chr.21: 32311019-32393003 on Build GRCh38
Transcripts
NM_014825.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030121 c.6632A>C p.Q2211P Chr.21: 32316468-32316468 on Build GRCh38
4792952 c.6550C>A p.Q2184K Chr.21: 32316550-32316550 on Build GRCh38
4624375 c.6509C>T p.A2170V Chr.21: 32316591-32316591 on Build GRCh38
6217010 c.6243C>T p.D2081D Chr.21: 32316857-32316857 on Build GRCh38
165418 c.6622G>A p.E2208K Chr.21: 32316478-32316478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929524155 Chr.21: 32316447-32316447 on Build GRCh38
rs763847056 Chr.21: 32316784-32316784 on Build GRCh38
rs867791584 Chr.21: 32316448-32316448 on Build GRCh38
rs910571772 Chr.21: 32316416-32316416 on Build GRCh38
rs553719917 Chr.21: 32316460-32316460 on Build GRCh38
rs758730332 Chr.21: 32316544-32316544 on Build GRCh38
rs530950328 Chr.21: 32316833-32316833 on Build GRCh38
rs372120737 Chr.21: 32316667-32316667 on Build GRCh38
rs868386199 Chr.21: 32316630-32316630 on Build GRCh38
rs1049224731 Chr.21: 32316469-32316469 on Build GRCh38
rs34860762 Chr.21: 32316827-32316828 on Build GRCh38
rs907891893 Chr.21: 32316400-32316400 on Build GRCh38
rs369395272 Chr.21: 32316656-32316656 on Build GRCh38
rs575644075 Chr.21: 32316568-32316568 on Build GRCh38
rs899603825 Chr.21: 32316587-32316587 on Build GRCh38
rs996584693 Chr.21: 32316612-32316612 on Build GRCh38
rs917914908 Chr.21: 32316780-32316780 on Build GRCh38
rs752063346 Chr.21: 32316816-32316816 on Build GRCh38
rs563331852 Chr.21: 32316779-32316779 on Build GRCh38
rs139492775 Chr.21: 32316845-32316845 on Build GRCh38
rs766723889 Chr.21: 32316812-32316812 on Build GRCh38
rs559741205 Chr.21: 32316668-32316668 on Build GRCh38
rs755734830 Chr.21: 32316521-32316521 on Build GRCh38
rs899467340 Chr.21: 32316840-32316840 on Build GRCh38
rs773063394 Chr.21: 32316694-32316694 on Build GRCh38
rs980799581 Chr.21: 32316792-32316792 on Build GRCh38
rs766324467 Chr.21: 32316803-32316803 on Build GRCh38
rs201544976 Chr.21: 32316420-32316420 on Build GRCh38
rs892139619 Chr.21: 32316553-32316553 on Build GRCh38
rs976115996 Chr.21: 32316579-32316579 on Build GRCh38
rs1034656760 Chr.21: 32316534-32316534 on Build GRCh38
rs909485549 Chr.21: 32316832-32316832 on Build GRCh38
rs956617629 Chr.21: 32316541-32316541 on Build GRCh38
rs887945690 Chr.21: 32316482-32316482 on Build GRCh38
rs1044906788 Chr.21: 32316522-32316522 on Build GRCh38
rs748328338 Chr.21: 32316586-32316586 on Build GRCh38
rs184986771 Chr.21: 32316559-32316559 on Build GRCh38
rs903668222 Chr.21: 32316532-32316532 on Build GRCh38
rs563626021 Chr.21: 32316811-32316811 on Build GRCh38
rs189966185 Chr.21: 32316838-32316838 on Build GRCh38
rs556682663 Chr.21: 32316823-32316823 on Build GRCh38
rs765542162 Chr.21: 32316524-32316524 on Build GRCh38
rs1029176778 Chr.21: 32316629-32316629 on Build GRCh38
rs538843308 Chr.21: 32316444-32316444 on Build GRCh38
rs866238648 Chr.21: 32316745-32316745 on Build GRCh38
rs565249859 Chr.21: 32316635-32316635 on Build GRCh38
rs1009515849 Chr.21: 32316695-32316695 on Build GRCh38
rs966155542 Chr.21: 32316781-32316781 on Build GRCh38
rs554136940 Chr.21: 32316560-32316560 on Build GRCh38
rs750760039 Chr.21: 32316543-32316543 on Build GRCh38
rs943950604 Chr.21: 32316837-32316837 on Build GRCh38
rs377686425 Chr.21: 32316637-32316637 on Build GRCh38
rs538038531 Chr.21: 32316764-32316764 on Build GRCh38
rs35894715 Chr.21: 32316786-32316787 on Build GRCh38
rs759177606 Chr.21: 32316440-32316440 on Build GRCh38
rs775017773 Chr.21: 32316773-32316773 on Build GRCh38
rs770839452 Chr.21: 32316651-32316651 on Build GRCh38
rs543664623 Chr.21: 32316575-32316575 on Build GRCh38
rs962318342 Chr.21: 32316747-32316747 on Build GRCh38
rs1022202792 Chr.21: 32316767-32316767 on Build GRCh38
rs548709746 Chr.21: 32316783-32316783 on Build GRCh38
rs1003228917 Chr.21: 32316533-32316533 on Build GRCh38
rs940646903 Chr.21: 32316415-32316415 on Build GRCh38
rs1005749164 Chr.21: 32316511-32316511 on Build GRCh38
rs948146022 Chr.21: 32316517-32316517 on Build GRCh38
rs7279896 Chr.21: 32316554-32316554 on Build GRCh38
rs574955611 Chr.21: 32316844-32316844 on Build GRCh38
rs763365015 Chr.21: 32316804-32316804 on Build GRCh38
rs1049294888 Chr.21: 32316606-32316606 on Build GRCh38
rs749147614 Chr.21: 32316642-32316642 on Build GRCh38
rs952165137 Chr.21: 32316636-32316636 on Build GRCh38
rs929601068 Chr.21: 32316588-32316588 on Build GRCh38
rs1040751016 Chr.21: 32316441-32316441 on Build GRCh38

View Full Product Details