Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29561084-29561578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPSNAP1
Entrez Gene ID
8508
Gene Name
nipsnap homolog 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.173878
Gene Chromosome Location
Chr.22: 29554808-29581337 on Build GRCh38
Transcripts
NM_001202502.1 NM_003634.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4994254 c.580-3C>T p.? Chr.22: 29561205-29561205 on Build GRCh38
4169190 c.512G>T p.S171I Chr.22: 29561573-29561573 on Build GRCh38
1308049 c.576C>G p.L192L Chr.22: 29561509-29561509 on Build GRCh38
4900862 c.568T>C p.Y190H Chr.22: 29561517-29561517 on Build GRCh38
1741742 c.536G>C p.R179T Chr.22: 29561549-29561549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761239603 Chr.22: 29561543-29561543 on Build GRCh38
rs1027543345 Chr.22: 29561449-29561461 on Build GRCh38
rs940586220 Chr.22: 29561509-29561509 on Build GRCh38
rs202154680 Chr.22: 29561128-29561128 on Build GRCh38
rs748445665 Chr.22: 29561472-29561472 on Build GRCh38
rs768712798 Chr.22: 29561239-29561239 on Build GRCh38
rs755360682 Chr.22: 29561146-29561146 on Build GRCh38
rs775731439 Chr.22: 29561228-29561228 on Build GRCh38
rs770711246 Chr.22: 29561224-29561224 on Build GRCh38
rs981662211 Chr.22: 29561361-29561361 on Build GRCh38
rs181346731 Chr.22: 29561372-29561372 on Build GRCh38
rs747559643 Chr.22: 29561209-29561209 on Build GRCh38
rs763141738 Chr.22: 29561234-29561234 on Build GRCh38
rs750952666 Chr.22: 29561458-29561458 on Build GRCh38
rs921564306 Chr.22: 29561294-29561294 on Build GRCh38
rs189514803 Chr.22: 29561293-29561293 on Build GRCh38
rs963110096 Chr.22: 29561142-29561142 on Build GRCh38
rs947171379 Chr.22: 29561423-29561423 on Build GRCh38
rs746868297 Chr.22: 29561220-29561220 on Build GRCh38
rs748816964 Chr.22: 29561360-29561360 on Build GRCh38
rs553684499 Chr.22: 29561350-29561350 on Build GRCh38
rs772073593 Chr.22: 29561211-29561211 on Build GRCh38
rs369466677 Chr.22: 29561465-29561465 on Build GRCh38
rs1033662399 Chr.22: 29561108-29561108 on Build GRCh38
rs372765883 Chr.22: 29561146-29561146 on Build GRCh38
rs6361 Chr.22: 29561332-29561332 on Build GRCh38
rs914202668 Chr.22: 29561399-29561399 on Build GRCh38
rs760823284 Chr.22: 29561481-29561481 on Build GRCh38
rs372170807 Chr.22: 29561494-29561494 on Build GRCh38
rs891140571 Chr.22: 29561323-29561323 on Build GRCh38
rs373232726 Chr.22: 29561484-29561484 on Build GRCh38
rs574268263 Chr.22: 29561433-29561433 on Build GRCh38
rs556022665 Chr.22: 29561397-29561397 on Build GRCh38
rs907048118 Chr.22: 29561435-29561435 on Build GRCh38
rs947120846 Chr.22: 29561289-29561289 on Build GRCh38
rs753592033 Chr.22: 29561469-29561469 on Build GRCh38
rs770021721 Chr.22: 29561126-29561126 on Build GRCh38
rs900457067 Chr.22: 29561116-29561116 on Build GRCh38
rs928321784 Chr.22: 29561218-29561218 on Build GRCh38
rs930320808 Chr.22: 29561295-29561295 on Build GRCh38
rs778385643 Chr.22: 29561472-29561472 on Build GRCh38
rs746921546 Chr.22: 29561480-29561480 on Build GRCh38
rs199705087 Chr.22: 29561535-29561535 on Build GRCh38
rs371622704 Chr.22: 29561134-29561134 on Build GRCh38
rs754678599 Chr.22: 29561470-29561470 on Build GRCh38
rs748317025 Chr.22: 29561507-29561507 on Build GRCh38
rs956479311 Chr.22: 29561374-29561374 on Build GRCh38
rs763788072 Chr.22: 29561405-29561405 on Build GRCh38
rs1016008565 Chr.22: 29561344-29561344 on Build GRCh38
rs778461433 Chr.22: 29561159-29561159 on Build GRCh38
rs777517826 Chr.22: 29561219-29561219 on Build GRCh38
rs761732100 Chr.22: 29561254-29561254 on Build GRCh38
rs201758280 Chr.22: 29561149-29561150 on Build GRCh38
rs768082496 Chr.22: 29561455-29561455 on Build GRCh38
rs377209759 Chr.22: 29561122-29561122 on Build GRCh38
rs754483576 Chr.22: 29561156-29561156 on Build GRCh38
rs376419305 Chr.22: 29561493-29561493 on Build GRCh38
rs201701870 Chr.22: 29561139-29561139 on Build GRCh38
rs200152347 Chr.22: 29561205-29561205 on Build GRCh38
rs374769736 Chr.22: 29561135-29561135 on Build GRCh38
rs375842229 Chr.22: 29561338-29561338 on Build GRCh38
rs898605246 Chr.22: 29561345-29561345 on Build GRCh38
rs185034873 Chr.22: 29561473-29561473 on Build GRCh38
rs189739703 Chr.22: 29561485-29561485 on Build GRCh38
rs988971609 Chr.22: 29561253-29561253 on Build GRCh38
rs572059046 Chr.22: 29561354-29561354 on Build GRCh38
rs544960193 Chr.22: 29561434-29561434 on Build GRCh38
rs768928750 Chr.22: 29561496-29561496 on Build GRCh38
rs373926410 Chr.22: 29561527-29561527 on Build GRCh38
rs578161472 Chr.22: 29561478-29561478 on Build GRCh38
rs750712904 Chr.22: 29561466-29561466 on Build GRCh38
rs527746841 Chr.22: 29561154-29561154 on Build GRCh38
rs933245845 Chr.22: 29561298-29561298 on Build GRCh38
rs5997494 Chr.22: 29561151-29561151 on Build GRCh38
rs537784152 Chr.22: 29561250-29561250 on Build GRCh38
rs914360668 Chr.22: 29561281-29561281 on Build GRCh38
rs571270739 Chr.22: 29561313-29561313 on Build GRCh38
rs774379609 Chr.22: 29561497-29561497 on Build GRCh38
rs548788416 Chr.22: 29561185-29561185 on Build GRCh38

View Full Product Details