Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10291219-10291714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
UniGene
Hs.590080
Gene Chromosome Location
Chr.3: 10285750-10292947 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

SNP Detail

SNP ID Chromosome Location
rs969258173 Chr.3: 10291308-10291308 on Build GRCh38
rs756279159 Chr.3: 10291503-10291503 on Build GRCh38
rs753925077 Chr.3: 10291324-10291324 on Build GRCh38
rs997230125 Chr.3: 10291617-10291617 on Build GRCh38
rs993044956 Chr.3: 10291512-10291512 on Build GRCh38
rs781328213 Chr.3: 10291432-10291432 on Build GRCh38
rs887531750 Chr.3: 10291363-10291363 on Build GRCh38
rs26311 Chr.3: 10291242-10291242 on Build GRCh38
rs930519789 Chr.3: 10291551-10291551 on Build GRCh38
rs560224641 Chr.3: 10291276-10291276 on Build GRCh38
rs568141766 Chr.3: 10291446-10291446 on Build GRCh38
rs969164603 Chr.3: 10291253-10291253 on Build GRCh38
rs1048011098 Chr.3: 10291562-10291562 on Build GRCh38
rs929076248 Chr.3: 10291359-10291359 on Build GRCh38
rs182919329 Chr.3: 10291564-10291564 on Build GRCh38
rs1024035686 Chr.3: 10291682-10291682 on Build GRCh38
rs1010070521 Chr.3: 10291429-10291429 on Build GRCh38
rs975594048 Chr.3: 10291292-10291292 on Build GRCh38
rs551761749 Chr.3: 10291376-10291376 on Build GRCh38
rs924332571 Chr.3: 10291360-10291360 on Build GRCh38
rs965594026 Chr.3: 10291280-10291280 on Build GRCh38
rs751785251 Chr.3: 10291266-10291266 on Build GRCh38
rs979321712 Chr.3: 10291471-10291471 on Build GRCh38
rs940113737 Chr.3: 10291569-10291569 on Build GRCh38
rs1031449061 Chr.3: 10291473-10291473 on Build GRCh38
rs527288959 Chr.3: 10291299-10291299 on Build GRCh38
rs997064408 Chr.3: 10291451-10291451 on Build GRCh38
rs934338932 Chr.3: 10291362-10291362 on Build GRCh38
rs766727388 Chr.3: 10291286-10291286 on Build GRCh38
rs948620128 Chr.3: 10291397-10291397 on Build GRCh38
rs908984456 Chr.3: 10291540-10291541 on Build GRCh38
rs1038662433 Chr.3: 10291389-10291389 on Build GRCh38
rs955771013 Chr.3: 10291479-10291479 on Build GRCh38
rs1028495585 Chr.3: 10291294-10291294 on Build GRCh38
rs139629059 Chr.3: 10291613-10291613 on Build GRCh38
rs956158838 Chr.3: 10291677-10291677 on Build GRCh38
rs887177559 Chr.3: 10291440-10291440 on Build GRCh38
rs553701283 Chr.3: 10291565-10291565 on Build GRCh38
rs1031845584 Chr.3: 10291652-10291652 on Build GRCh38
rs145984984 Chr.3: 10291419-10291419 on Build GRCh38
rs187926655 Chr.3: 10291637-10291637 on Build GRCh38
rs1022163116 Chr.3: 10291444-10291444 on Build GRCh38
rs1004205530 Chr.3: 10291445-10291445 on Build GRCh38
rs948338968 Chr.3: 10291515-10291515 on Build GRCh38
rs901143363 Chr.3: 10291449-10291449 on Build GRCh38
rs1044670926 Chr.3: 10291403-10291403 on Build GRCh38
rs1027454526 Chr.3: 10291254-10291254 on Build GRCh38
rs920488138 Chr.3: 10291533-10291533 on Build GRCh38
rs951430227 Chr.3: 10291269-10291269 on Build GRCh38
rs1000408195 Chr.3: 10291645-10291645 on Build GRCh38
rs1041075738 Chr.3: 10291572-10291572 on Build GRCh38
rs959988149 Chr.3: 10291685-10291685 on Build GRCh38
rs374074206 Chr.3: 10291513-10291513 on Build GRCh38
rs370470221 Chr.3: 10291632-10291632 on Build GRCh38
rs763825440 Chr.3: 10291244-10291244 on Build GRCh38
rs190663799 Chr.3: 10291480-10291480 on Build GRCh38
rs933242460 Chr.3: 10291591-10291591 on Build GRCh38
rs900143879 Chr.3: 10291573-10291573 on Build GRCh38
rs3755777 Chr.3: 10291680-10291680 on Build GRCh38
rs1028597200 Chr.3: 10291627-10291627 on Build GRCh38
rs918720417 Chr.3: 10291341-10291341 on Build GRCh38
rs570375479 Chr.3: 10291411-10291411 on Build GRCh38
rs549538472 Chr.3: 10291423-10291423 on Build GRCh38
rs1019851252 Chr.3: 10291271-10291271 on Build GRCh38
rs978548674 Chr.3: 10291315-10291315 on Build GRCh38
rs563169020 Chr.3: 10291548-10291548 on Build GRCh38
rs753247013 Chr.3: 10291531-10291531 on Build GRCh38
rs535675510 Chr.3: 10291455-10291455 on Build GRCh38
rs1057337433 Chr.3: 10291337-10291337 on Build GRCh38
rs968933088 Chr.3: 10291470-10291470 on Build GRCh38
rs187898498 Chr.3: 10291245-10291245 on Build GRCh38
rs765784867 Chr.3: 10291516-10291516 on Build GRCh38
rs1004148392 Chr.3: 10291270-10291270 on Build GRCh38
rs992485697 Chr.3: 10291371-10291371 on Build GRCh38
rs914581459 Chr.3: 10291251-10291251 on Build GRCh38

View Full Product Details