Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33813471-33813972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD6IP
Entrez Gene ID
10015
Gene Name
programmed cell death 6 interacting protein
Gene Aliases
AIP1, ALIX, DRIP4, HP95
UniGene
Hs.475896
Gene Chromosome Location
Chr.3: 33798571-33869707 on Build GRCh38
Transcripts
NM_001162429.2 NM_001256192.1 NM_013374.5 XM_011533252.1 XM_011533253.1 XM_017005488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825075 c.323C>G p.S108C Chr.3: 33813617-33813617 on Build GRCh38
446383 c.323C>T p.S108F Chr.3: 33813617-33813617 on Build GRCh38
14020 c.293C>T p.A98V Chr.3: 33813587-33813587 on Build GRCh38
480038 c.265-1G>A p.? Chr.3: 33813558-33813558 on Build GRCh38
7156928 c.267C>G p.I89M Chr.3: 33813561-33813561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34629079 Chr.3: 33813728-33813729 on Build GRCh38
rs766639769 Chr.3: 33813619-33813619 on Build GRCh38
rs753048318 Chr.3: 33813727-33813727 on Build GRCh38
rs186482804 Chr.3: 33813948-33813948 on Build GRCh38
rs1021422046 Chr.3: 33813725-33813725 on Build GRCh38
rs189596121 Chr.3: 33813848-33813848 on Build GRCh38
rs960336151 Chr.3: 33813812-33813812 on Build GRCh38
rs551721182 Chr.3: 33813901-33813901 on Build GRCh38
rs770243824 Chr.3: 33813600-33813600 on Build GRCh38
rs1019442681 Chr.3: 33813749-33813749 on Build GRCh38
rs965218508 Chr.3: 33813832-33813832 on Build GRCh38
rs755450823 Chr.3: 33813550-33813550 on Build GRCh38
rs370717543 Chr.3: 33813564-33813564 on Build GRCh38
rs746439916 Chr.3: 33813551-33813551 on Build GRCh38
rs1015309373 Chr.3: 33813834-33813834 on Build GRCh38
rs550202885 Chr.3: 33813926-33813926 on Build GRCh38
rs758633467 Chr.3: 33813746-33813746 on Build GRCh38
rs1003122818 Chr.3: 33813656-33813656 on Build GRCh38
rs759484252 Chr.3: 33813660-33813660 on Build GRCh38
rs149504311 Chr.3: 33813591-33813591 on Build GRCh38
rs1034256062 Chr.3: 33813810-33813810 on Build GRCh38
rs778644410 Chr.3: 33813523-33813523 on Build GRCh38
rs377044540 Chr.3: 33813806-33813806 on Build GRCh38
rs111655514 Chr.3: 33813678-33813678 on Build GRCh38
rs748666912 Chr.3: 33813592-33813592 on Build GRCh38
rs577172001 Chr.3: 33813519-33813519 on Build GRCh38
rs745726615 Chr.3: 33813530-33813530 on Build GRCh38
rs181862755 Chr.3: 33813867-33813867 on Build GRCh38
rs750157711 Chr.3: 33813679-33813679 on Build GRCh38
rs774743108 Chr.3: 33813646-33813646 on Build GRCh38
rs758029828 Chr.3: 33813548-33813548 on Build GRCh38
rs753835894 Chr.3: 33813509-33813509 on Build GRCh38
rs893239105 Chr.3: 33813510-33813510 on Build GRCh38
rs538187955 Chr.3: 33813628-33813628 on Build GRCh38
rs370110920 Chr.3: 33813661-33813661 on Build GRCh38
rs373997652 Chr.3: 33813664-33813664 on Build GRCh38
rs757210877 Chr.3: 33813667-33813667 on Build GRCh38
rs992859745 Chr.3: 33813813-33813813 on Build GRCh38
rs962127022 Chr.3: 33813845-33813845 on Build GRCh38
rs773708920 Chr.3: 33813613-33813613 on Build GRCh38
rs562655595 Chr.3: 33813730-33813730 on Build GRCh38
rs921202999 Chr.3: 33813873-33813873 on Build GRCh38
rs559845236 Chr.3: 33813851-33813851 on Build GRCh38
rs574590549 Chr.3: 33813761-33813761 on Build GRCh38
rs780903739 Chr.3: 33813890-33813890 on Build GRCh38
rs148116117 Chr.3: 33813618-33813618 on Build GRCh38
rs774948019 Chr.3: 33813634-33813634 on Build GRCh38
rs1054450456 Chr.3: 33813507-33813507 on Build GRCh38

View Full Product Details