Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43056491-43057001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM198A
Entrez Gene ID
729085
Gene Name
family with sequence similarity 198 member A
Gene Aliases
C3orf41
UniGene
Hs.729533
Gene Chromosome Location
Chr.3: 42979267-43057715 on Build GRCh38
Transcripts
NM_001129908.2

SNP Detail

SNP ID Chromosome Location
rs938371095 Chr.3: 43056865-43056865 on Build GRCh38
rs530331079 Chr.3: 43056667-43056667 on Build GRCh38
rs568918566 Chr.3: 43056532-43056532 on Build GRCh38
rs114786723 Chr.3: 43056541-43056541 on Build GRCh38
rs1052917829 Chr.3: 43056623-43056623 on Build GRCh38
rs749428678 Chr.3: 43056563-43056563 on Build GRCh38
rs651899 Chr.3: 43056734-43056734 on Build GRCh38
rs989644551 Chr.3: 43056595-43056595 on Build GRCh38
rs952986090 Chr.3: 43056735-43056735 on Build GRCh38
rs562509608 Chr.3: 43056585-43056585 on Build GRCh38
rs897433055 Chr.3: 43056908-43056908 on Build GRCh38
rs896919034 Chr.3: 43056915-43056915 on Build GRCh38
rs1056688756 Chr.3: 43056910-43056910 on Build GRCh38
rs1035186255 Chr.3: 43056702-43056702 on Build GRCh38
rs552102489 Chr.3: 43056527-43056527 on Build GRCh38
rs561598223 Chr.3: 43056774-43056774 on Build GRCh38
rs1011526485 Chr.3: 43056666-43056666 on Build GRCh38
rs941712432 Chr.3: 43056889-43056889 on Build GRCh38
rs914450643 Chr.3: 43056803-43056803 on Build GRCh38
rs572119221 Chr.3: 43056784-43056784 on Build GRCh38
rs923048851 Chr.3: 43056560-43056560 on Build GRCh38
rs934499428 Chr.3: 43056618-43056618 on Build GRCh38
rs6769625 Chr.3: 43056726-43056726 on Build GRCh38
rs137924368 Chr.3: 43056534-43056534 on Build GRCh38
rs148284912 Chr.3: 43056969-43056969 on Build GRCh38
rs979438228 Chr.3: 43056765-43056765 on Build GRCh38
rs576398205 Chr.3: 43056647-43056648 on Build GRCh38
rs75588702 Chr.3: 43056762-43056762 on Build GRCh38
rs1005864091 Chr.3: 43056614-43056614 on Build GRCh38
rs1036014041 Chr.3: 43056892-43056892 on Build GRCh38
rs896888082 Chr.3: 43056714-43056714 on Build GRCh38
rs923883374 Chr.3: 43056824-43056824 on Build GRCh38
rs149094641 Chr.3: 43056577-43056577 on Build GRCh38
rs916577972 Chr.3: 43056517-43056517 on Build GRCh38
rs1035412734 Chr.3: 43056703-43056703 on Build GRCh38
rs935368789 Chr.3: 43056770-43056770 on Build GRCh38
rs531894666 Chr.3: 43056539-43056539 on Build GRCh38
rs1053847 Chr.3: 43056704-43056704 on Build GRCh38
rs553942829 Chr.3: 43056652-43056652 on Build GRCh38
rs6772252 Chr.3: 43056962-43056962 on Build GRCh38
rs965399908 Chr.3: 43056757-43056757 on Build GRCh38
rs893050249 Chr.3: 43056662-43056662 on Build GRCh38
rs797018156 Chr.3: 43056746-43056746 on Build GRCh38
rs116610240 Chr.3: 43056557-43056557 on Build GRCh38
rs760381308 Chr.3: 43056916-43056916 on Build GRCh38
rs989962709 Chr.3: 43056776-43056776 on Build GRCh38
rs113216258 Chr.3: 43056736-43056736 on Build GRCh38
rs1002478724 Chr.3: 43056691-43056691 on Build GRCh38
rs1057078214 Chr.3: 43056957-43056957 on Build GRCh38
rs976361830 Chr.3: 43056840-43056840 on Build GRCh38
rs976721737 Chr.3: 43056760-43056760 on Build GRCh38
rs905757835 Chr.3: 43056855-43056855 on Build GRCh38
rs946679995 Chr.3: 43056518-43056518 on Build GRCh38
rs17469719 Chr.3: 43056838-43056838 on Build GRCh38
rs143126010 Chr.3: 43056854-43056854 on Build GRCh38
rs905559397 Chr.3: 43056682-43056682 on Build GRCh38

View Full Product Details