Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44660126-44660621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF35
Entrez Gene ID
7584
Gene Name
zinc finger protein 35
Gene Aliases
HF.10, HF10, Zfp105
UniGene
Hs.288658
Gene Chromosome Location
Chr.3: 44648723-44660791 on Build GRCh38
Transcripts
NM_003420.3 XM_017007129.1 XM_017007130.1 XM_017007131.1

SNP Detail

SNP ID Chromosome Location
rs951400582 Chr.3: 44660424-44660424 on Build GRCh38
rs76288781 Chr.3: 44660259-44660259 on Build GRCh38
rs958741155 Chr.3: 44660575-44660575 on Build GRCh38
rs777749084 Chr.3: 44660175-44660175 on Build GRCh38
rs927070593 Chr.3: 44660533-44660533 on Build GRCh38
rs576859834 Chr.3: 44660244-44660244 on Build GRCh38
rs180799438 Chr.3: 44660357-44660357 on Build GRCh38
rs1014866422 Chr.3: 44660484-44660484 on Build GRCh38
rs1030109205 Chr.3: 44660423-44660423 on Build GRCh38
rs145636061 Chr.3: 44660239-44660239 on Build GRCh38
rs1005231949 Chr.3: 44660438-44660438 on Build GRCh38
rs992584496 Chr.3: 44660589-44660589 on Build GRCh38
rs574898577 Chr.3: 44660329-44660329 on Build GRCh38
rs746109904 Chr.3: 44660287-44660287 on Build GRCh38
rs761042733 Chr.3: 44660432-44660432 on Build GRCh38
rs542416697 Chr.3: 44660346-44660346 on Build GRCh38
rs552946609 Chr.3: 44660382-44660382 on Build GRCh38
rs532175824 Chr.3: 44660594-44660594 on Build GRCh38
rs115539999 Chr.3: 44660150-44660150 on Build GRCh38
rs973208430 Chr.3: 44660248-44660248 on Build GRCh38
rs749394451 Chr.3: 44660215-44660215 on Build GRCh38
rs879789651 Chr.3: 44660284-44660285 on Build GRCh38
rs1402753 Chr.3: 44660246-44660246 on Build GRCh38
rs983379703 Chr.3: 44660191-44660191 on Build GRCh38
rs973240021 Chr.3: 44660531-44660531 on Build GRCh38
rs560763808 Chr.3: 44660303-44660303 on Build GRCh38
rs963259725 Chr.3: 44660504-44660504 on Build GRCh38
rs563031614 Chr.3: 44660386-44660386 on Build GRCh38
rs998277791 Chr.3: 44660401-44660401 on Build GRCh38
rs937071336 Chr.3: 44660268-44660268 on Build GRCh38
rs1041962717 Chr.3: 44660355-44660355 on Build GRCh38
rs954827476 Chr.3: 44660157-44660157 on Build GRCh38
rs531234253 Chr.3: 44660367-44660367 on Build GRCh38
rs899936315 Chr.3: 44660400-44660400 on Build GRCh38
rs927040535 Chr.3: 44660262-44660262 on Build GRCh38
rs946327320 Chr.3: 44660347-44660347 on Build GRCh38
rs188842958 Chr.3: 44660298-44660298 on Build GRCh38

View Full Product Details