Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48688304-48688799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_016291.3 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741860 c.1019G>A p.R340Q Chr.3: 48688535-48688535 on Build GRCh38
7284934 c.991C>T p.L331L Chr.3: 48688563-48688563 on Build GRCh38
3595117 c.990C>T p.S330S Chr.3: 48688564-48688564 on Build GRCh38
5419056 c.1186A>G p.T396A Chr.3: 48688368-48688368 on Build GRCh38
5514933 c.1024G>A p.E342K Chr.3: 48688530-48688530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141135563 Chr.3: 48688366-48688366 on Build GRCh38
rs200712199 Chr.3: 48688434-48688434 on Build GRCh38
rs747728098 Chr.3: 48688746-48688746 on Build GRCh38
rs750586432 Chr.3: 48688681-48688681 on Build GRCh38
rs941572891 Chr.3: 48688401-48688401 on Build GRCh38
rs571299199 Chr.3: 48688377-48688377 on Build GRCh38
rs997321233 Chr.3: 48688612-48688612 on Build GRCh38
rs760155028 Chr.3: 48688368-48688368 on Build GRCh38
rs772045943 Chr.3: 48688493-48688493 on Build GRCh38
rs773308953 Chr.3: 48688396-48688396 on Build GRCh38
rs150963193 Chr.3: 48688410-48688410 on Build GRCh38
rs779200655 Chr.3: 48688674-48688674 on Build GRCh38
rs1046234047 Chr.3: 48688460-48688460 on Build GRCh38
rs778980331 Chr.3: 48688588-48688588 on Build GRCh38
rs775420494 Chr.3: 48688623-48688623 on Build GRCh38
rs760183457 Chr.3: 48688500-48688500 on Build GRCh38
rs199770977 Chr.3: 48688708-48688708 on Build GRCh38
rs966293134 Chr.3: 48688606-48688606 on Build GRCh38
rs150229636 Chr.3: 48688737-48688737 on Build GRCh38
rs905913166 Chr.3: 48688474-48688474 on Build GRCh38
rs950221667 Chr.3: 48688438-48688438 on Build GRCh38
rs747008986 Chr.3: 48688625-48688625 on Build GRCh38
rs1016338010 Chr.3: 48688579-48688579 on Build GRCh38
rs375983230 Chr.3: 48688619-48688619 on Build GRCh38
rs759575244 Chr.3: 48688362-48688362 on Build GRCh38
rs756274683 Chr.3: 48688536-48688536 on Build GRCh38
rs904382016 Chr.3: 48688360-48688360 on Build GRCh38
rs768166028 Chr.3: 48688531-48688531 on Build GRCh38
rs945679630 Chr.3: 48688726-48688726 on Build GRCh38
rs1043229533 Chr.3: 48688729-48688729 on Build GRCh38
rs771044717 Chr.3: 48688445-48688445 on Build GRCh38
rs538381890 Chr.3: 48688423-48688423 on Build GRCh38
rs775367465 Chr.3: 48688494-48688494 on Build GRCh38
rs763538514 Chr.3: 48688503-48688503 on Build GRCh38
rs768475478 Chr.3: 48688628-48688628 on Build GRCh38
rs764767959 Chr.3: 48688525-48688525 on Build GRCh38
rs774268067 Chr.3: 48688466-48688466 on Build GRCh38
rs771127659 Chr.3: 48688665-48688665 on Build GRCh38
rs749323565 Chr.3: 48688543-48688543 on Build GRCh38
rs753183941 Chr.3: 48688535-48688535 on Build GRCh38
rs143054185 Chr.3: 48688526-48688526 on Build GRCh38
rs753998544 Chr.3: 48688669-48688669 on Build GRCh38
rs150451049 Chr.3: 48688412-48688412 on Build GRCh38
rs776248772 Chr.3: 48688630-48688630 on Build GRCh38
rs772703699 Chr.3: 48688657-48688657 on Build GRCh38
rs927416196 Chr.3: 48688651-48688651 on Build GRCh38
rs780955935 Chr.3: 48688771-48688771 on Build GRCh38
rs745632118 Chr.3: 48688492-48688492 on Build GRCh38
rs757538475 Chr.3: 48688549-48688549 on Build GRCh38
rs769513321 Chr.3: 48688653-48688653 on Build GRCh38
rs1020131272 Chr.3: 48688580-48688580 on Build GRCh38
rs753464760 Chr.3: 48688395-48688395 on Build GRCh38
rs893079246 Chr.3: 48688534-48688534 on Build GRCh38
rs368740167 Chr.3: 48688652-48688652 on Build GRCh38
rs762647213 Chr.3: 48688334-48688335 on Build GRCh38
rs745685310 Chr.3: 48688595-48688595 on Build GRCh38
rs141913549 Chr.3: 48688747-48688747 on Build GRCh38
rs376450910 Chr.3: 48688437-48688437 on Build GRCh38
rs755585369 Chr.3: 48688415-48688415 on Build GRCh38
rs375477556 Chr.3: 48688375-48688375 on Build GRCh38
rs761596550 Chr.3: 48688518-48688518 on Build GRCh38
rs771788541 Chr.3: 48688620-48688620 on Build GRCh38
rs768689853 Chr.3: 48688728-48688728 on Build GRCh38
rs144566634 Chr.3: 48688336-48688336 on Build GRCh38
rs755349495 Chr.3: 48688530-48688530 on Build GRCh38
rs376702328 Chr.3: 48688776-48688776 on Build GRCh38
rs777964687 Chr.3: 48688537-48688537 on Build GRCh38
rs775993668 Chr.3: 48688507-48688507 on Build GRCh38
rs767435418 Chr.3: 48688365-48688365 on Build GRCh38
rs138795713 Chr.3: 48688663-48688663 on Build GRCh38
rs372399821 Chr.3: 48688671-48688671 on Build GRCh38
rs759980196 Chr.3: 48688659-48688659 on Build GRCh38
rs761329817 Chr.3: 48688664-48688664 on Build GRCh38
rs746778515 Chr.3: 48688711-48688711 on Build GRCh38
rs781760039 Chr.3: 48688420-48688420 on Build GRCh38
rs758607679 Chr.3: 48688684-48688684 on Build GRCh38

View Full Product Details