Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108834355-108834851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
UniGene
Hs.729441
Gene Chromosome Location
Chr.1: 108815898-108858242 on Build GRCh38
Transcripts
NM_152763.4 NR_049760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4785060 c.1711G>T p.D571Y Chr.1: 108834482-108834482 on Build GRCh38
6241284 c.1726A>T p.R576W Chr.1: 108834467-108834467 on Build GRCh38
4785061 c.832G>T p.D278Y Chr.1: 108834482-108834482 on Build GRCh38
4217277 c.1665-8T>A p.? Chr.1: 108834536-108834536 on Build GRCh38
4770828 c.1674C>T p.N558N Chr.1: 108834519-108834519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749500729 Chr.1: 108834579-108834579 on Build GRCh38
rs780674477 Chr.1: 108834532-108834533 on Build GRCh38
rs764740118 Chr.1: 108834485-108834485 on Build GRCh38
rs557854470 Chr.1: 108834806-108834806 on Build GRCh38
rs909091995 Chr.1: 108834763-108834763 on Build GRCh38
rs750831106 Chr.1: 108834398-108834399 on Build GRCh38
rs758342132 Chr.1: 108834657-108834657 on Build GRCh38
rs923589697 Chr.1: 108834573-108834573 on Build GRCh38
rs546689243 Chr.1: 108834674-108834674 on Build GRCh38
rs775590361 Chr.1: 108834442-108834442 on Build GRCh38
rs76759362 Chr.1: 108834546-108834548 on Build GRCh38
rs375472058 Chr.1: 108834511-108834511 on Build GRCh38
rs997171143 Chr.1: 108834643-108834643 on Build GRCh38
rs80043780 Chr.1: 108834722-108834722 on Build GRCh38
rs935614898 Chr.1: 108834649-108834649 on Build GRCh38
rs560633616 Chr.1: 108834537-108834538 on Build GRCh38
rs12060255 Chr.1: 108834448-108834448 on Build GRCh38
rs778710149 Chr.1: 108834425-108834425 on Build GRCh38
rs1005975056 Chr.1: 108834622-108834622 on Build GRCh38
rs972630568 Chr.1: 108834496-108834496 on Build GRCh38
rs879087099 Chr.1: 108834536-108834537 on Build GRCh38
rs754432020 Chr.1: 108834480-108834480 on Build GRCh38
rs757149678 Chr.1: 108834506-108834507 on Build GRCh38
rs188844055 Chr.1: 108834433-108834433 on Build GRCh38
rs751416332 Chr.1: 108834545-108834545 on Build GRCh38
rs753375494 Chr.1: 108834404-108834404 on Build GRCh38
rs900256836 Chr.1: 108834636-108834636 on Build GRCh38
rs200066791 Chr.1: 108834537-108834537 on Build GRCh38
rs112859436 Chr.1: 108834535-108834536 on Build GRCh38
rs12061417 Chr.1: 108834712-108834712 on Build GRCh38
rs777482761 Chr.1: 108834499-108834499 on Build GRCh38
rs577829834 Chr.1: 108834536-108834537 on Build GRCh38
rs146520059 Chr.1: 108834468-108834468 on Build GRCh38
rs780895785 Chr.1: 108834566-108834566 on Build GRCh38
rs5776951 Chr.1: 108834548-108834548 on Build GRCh38
rs747472161 Chr.1: 108834533-108834533 on Build GRCh38
rs955670455 Chr.1: 108834673-108834673 on Build GRCh38
rs760452529 Chr.1: 108834475-108834475 on Build GRCh38
rs757060796 Chr.1: 108834552-108834552 on Build GRCh38
rs1017310200 Chr.1: 108834624-108834624 on Build GRCh38
rs961305569 Chr.1: 108834495-108834495 on Build GRCh38
rs879015356 Chr.1: 108834602-108834602 on Build GRCh38
rs371831293 Chr.1: 108834428-108834428 on Build GRCh38
rs753463639 Chr.1: 108834478-108834478 on Build GRCh38
rs769985256 Chr.1: 108834436-108834436 on Build GRCh38
rs746778594 Chr.1: 108834505-108834505 on Build GRCh38
rs936162410 Chr.1: 108834470-108834470 on Build GRCh38
rs755004150 Chr.1: 108834411-108834411 on Build GRCh38
rs140022518 Chr.1: 108834518-108834518 on Build GRCh38
rs759462726 Chr.1: 108834541-108834542 on Build GRCh38
rs758307146 Chr.1: 108834493-108834493 on Build GRCh38
rs564945489 Chr.1: 108834547-108834548 on Build GRCh38
rs764658166 Chr.1: 108834469-108834469 on Build GRCh38
rs916038070 Chr.1: 108834702-108834702 on Build GRCh38
rs990248299 Chr.1: 108834471-108834471 on Build GRCh38
rs201859332 Chr.1: 108834536-108834536 on Build GRCh38
rs952148757 Chr.1: 108834446-108834446 on Build GRCh38
rs750430883 Chr.1: 108834597-108834597 on Build GRCh38
rs563187777 Chr.1: 108834450-108834450 on Build GRCh38
rs762311421 Chr.1: 108834457-108834457 on Build GRCh38
rs566599289 Chr.1: 108834689-108834689 on Build GRCh38
rs990279548 Chr.1: 108834698-108834698 on Build GRCh38
rs778708401 Chr.1: 108834508-108834508 on Build GRCh38
rs538612836 Chr.1: 108834701-108834701 on Build GRCh38
rs574267198 Chr.1: 108834808-108834808 on Build GRCh38
rs12063387 Chr.1: 108834661-108834661 on Build GRCh38
rs540382671 Chr.1: 108834537-108834537 on Build GRCh38
rs752230337 Chr.1: 108834538-108834538 on Build GRCh38
rs752608075 Chr.1: 108834487-108834487 on Build GRCh38
rs957535319 Chr.1: 108834690-108834690 on Build GRCh38
rs773751707 Chr.1: 108834530-108834530 on Build GRCh38
rs929293831 Chr.1: 108834723-108834723 on Build GRCh38
rs981494567 Chr.1: 108834718-108834718 on Build GRCh38
rs5776950 Chr.1: 108834536-108834536 on Build GRCh38
rs946389768 Chr.1: 108834539-108834540 on Build GRCh38
rs1006698769 Chr.1: 108834699-108834699 on Build GRCh38
rs756398873 Chr.1: 108834507-108834507 on Build GRCh38
rs537700313 Chr.1: 108834732-108834732 on Build GRCh38
rs887620677 Chr.1: 108834548-108834548 on Build GRCh38
rs1044379773 Chr.1: 108834389-108834392 on Build GRCh38
rs41278492 Chr.1: 108834814-108834814 on Build GRCh38
rs58624207 Chr.1: 108834749-108834749 on Build GRCh38
rs1034894716 Chr.1: 108834744-108834744 on Build GRCh38
rs902738197 Chr.1: 108834392-108834392 on Build GRCh38
rs761115020 Chr.1: 108834535-108834535 on Build GRCh38
rs369579016 Chr.1: 108834710-108834710 on Build GRCh38
rs201355205 Chr.1: 108834526-108834526 on Build GRCh38
rs1048113956 Chr.1: 108834761-108834761 on Build GRCh38
rs36028304 Chr.1: 108834542-108834543 on Build GRCh38
rs758310920 Chr.1: 108834539-108834539 on Build GRCh38
rs764018252 Chr.1: 108834540-108834540 on Build GRCh38
rs1040596830 Chr.1: 108834658-108834658 on Build GRCh38
rs946242 Chr.1: 108834431-108834431 on Build GRCh38
rs376435620 Chr.1: 108834409-108834409 on Build GRCh38
rs999667303 Chr.1: 108834410-108834412 on Build GRCh38
rs1031897130 Chr.1: 108834686-108834686 on Build GRCh38
rs996265780 Chr.1: 108834678-108834678 on Build GRCh38
rs769266154 Chr.1: 108834449-108834449 on Build GRCh38
rs774124965 Chr.1: 108834536-108834538 on Build GRCh38
rs758746413 Chr.1: 108834446-108834446 on Build GRCh38
rs144829009 Chr.1: 108834519-108834519 on Build GRCh38
rs749452393 Chr.1: 108834516-108834516 on Build GRCh38

View Full Product Details