Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 73060931-73061430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R2
Entrez Gene ID
151987
Gene Name
protein phosphatase 4 regulatory subunit 2
Gene Aliases
PP4R2
UniGene
Hs.431092
Gene Chromosome Location
Chr.3: 72996743-73069201 on Build GRCh38
Transcripts
NM_001318025.1 NM_001318026.1 NM_001318027.1 NM_174907.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6754511 c.393T>A p.V131V Chr.3: 73061034-73061034 on Build GRCh38
4421074 c.419+4A>G p.? Chr.3: 73061064-73061064 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs944316277 Chr.3: 73061220-73061220 on Build GRCh38
rs200109244 Chr.3: 73061080-73061080 on Build GRCh38
rs937391437 Chr.3: 73061121-73061121 on Build GRCh38
rs747207630 Chr.3: 73061192-73061199 on Build GRCh38
rs868680227 Chr.3: 73061374-73061374 on Build GRCh38
rs984597566 Chr.3: 73061077-73061077 on Build GRCh38
rs757531254 Chr.3: 73061008-73061008 on Build GRCh38
rs995810939 Chr.3: 73061263-73061263 on Build GRCh38
rs200948193 Chr.3: 73060990-73060990 on Build GRCh38
rs574151407 Chr.3: 73060973-73060973 on Build GRCh38
rs373009608 Chr.3: 73061057-73061057 on Build GRCh38
rs760269770 Chr.3: 73061106-73061106 on Build GRCh38
rs1042316345 Chr.3: 73061208-73061208 on Build GRCh38
rs914675358 Chr.3: 73061142-73061142 on Build GRCh38
rs968657261 Chr.3: 73061342-73061342 on Build GRCh38
rs778949580 Chr.3: 73061051-73061053 on Build GRCh38
rs1005021018 Chr.3: 73061149-73061152 on Build GRCh38
rs970512235 Chr.3: 73060987-73060987 on Build GRCh38
rs773125285 Chr.3: 73061105-73061105 on Build GRCh38
rs940879971 Chr.3: 73061390-73061390 on Build GRCh38
rs974719648 Chr.3: 73061351-73061351 on Build GRCh38
rs765781686 Chr.3: 73061107-73061107 on Build GRCh38
rs771976827 Chr.3: 73061088-73061088 on Build GRCh38
rs922196422 Chr.3: 73061115-73061115 on Build GRCh38
rs981942474 Chr.3: 73060998-73060998 on Build GRCh38
rs901491823 Chr.3: 73061260-73061260 on Build GRCh38
rs577979361 Chr.3: 73061078-73061078 on Build GRCh38
rs114941617 Chr.3: 73061015-73061015 on Build GRCh38
rs758238319 Chr.3: 73061241-73061242 on Build GRCh38
rs75777231 Chr.3: 73061366-73061366 on Build GRCh38
rs377500209 Chr.3: 73061002-73061002 on Build GRCh38
rs187033558 Chr.3: 73061269-73061269 on Build GRCh38
rs371174123 Chr.3: 73061016-73061016 on Build GRCh38
rs747884056 Chr.3: 73061095-73061095 on Build GRCh38
rs903426010 Chr.3: 73061209-73061209 on Build GRCh38
rs779440172 Chr.3: 73061017-73061017 on Build GRCh38
rs947495900 Chr.3: 73061187-73061187 on Build GRCh38
rs934735192 Chr.3: 73061047-73061047 on Build GRCh38
rs113215422 Chr.3: 73061134-73061134 on Build GRCh38
rs887531276 Chr.3: 73061348-73061348 on Build GRCh38
rs532999536 Chr.3: 73061394-73061394 on Build GRCh38
rs2637751 Chr.3: 73061372-73061372 on Build GRCh38
rs555688235 Chr.3: 73060972-73060972 on Build GRCh38
rs560252192 Chr.3: 73061096-73061096 on Build GRCh38
rs561754502 Chr.3: 73061317-73061317 on Build GRCh38
rs749208683 Chr.3: 73060995-73060995 on Build GRCh38
rs1011925020 Chr.3: 73061211-73061211 on Build GRCh38
rs758979002 Chr.3: 73061051-73061051 on Build GRCh38
rs770021261 Chr.3: 73061085-73061085 on Build GRCh38
rs1037473641 Chr.3: 73061202-73061202 on Build GRCh38
rs995905491 Chr.3: 73060996-73060996 on Build GRCh38
rs1052046503 Chr.3: 73061368-73061368 on Build GRCh38
rs569129912 Chr.3: 73061389-73061389 on Build GRCh38
rs951853328 Chr.3: 73061028-73061028 on Build GRCh38
rs527787353 Chr.3: 73061117-73061117 on Build GRCh38
rs962310529 Chr.3: 73061090-73061090 on Build GRCh38
rs761102350 Chr.3: 73061239-73061239 on Build GRCh38
rs1006051239 Chr.3: 73061355-73061355 on Build GRCh38
rs371972160 Chr.3: 73061092-73061092 on Build GRCh38
rs1035627699 Chr.3: 73061405-73061405 on Build GRCh38
rs749944884 Chr.3: 73060978-73060978 on Build GRCh38
rs61754958 Chr.3: 73061030-73061030 on Build GRCh38
rs777040538 Chr.3: 73061308-73061309 on Build GRCh38
rs536841252 Chr.3: 73061236-73061237 on Build GRCh38
rs775448037 Chr.3: 73061158-73061158 on Build GRCh38
rs746129974 Chr.3: 73061070-73061070 on Build GRCh38
rs1023516120 Chr.3: 73061232-73061235 on Build GRCh38
rs746102605 Chr.3: 73061084-73061084 on Build GRCh38
rs1031240558 Chr.3: 73061256-73061256 on Build GRCh38
rs753071231 Chr.3: 73061045-73061045 on Build GRCh38
rs1040021134 Chr.3: 73061254-73061254 on Build GRCh38

View Full Product Details