Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121586401-121586899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARGFX
Entrez Gene ID
503582
Gene Name
arginine-fifty homeobox
Gene Aliases
-
UniGene
Hs.224976
Gene Chromosome Location
Chr.3: 121567931-121590622 on Build GRCh38
Transcripts
NM_001012659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1717110 c.781G>C p.V261L Chr.3: 121586433-121586433 on Build GRCh38
3585961 c.797C>T p.S266F Chr.3: 121586449-121586449 on Build GRCh38
4188800 c.800C>G p.S267C Chr.3: 121586452-121586452 on Build GRCh38
3585962 c.938G>A p.G313E Chr.3: 121586590-121586590 on Build GRCh38
6306805 c.752A>G p.Y251C Chr.3: 121586404-121586404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199782325 Chr.3: 121586524-121586524 on Build GRCh38
rs79657977 Chr.3: 121586515-121586515 on Build GRCh38
rs772768849 Chr.3: 121586519-121586520 on Build GRCh38
rs1047924761 Chr.3: 121586829-121586829 on Build GRCh38
rs1032004515 Chr.3: 121586720-121586720 on Build GRCh38
rs762861064 Chr.3: 121586591-121586591 on Build GRCh38
rs531424959 Chr.3: 121586601-121586601 on Build GRCh38
rs562116526 Chr.3: 121586693-121586693 on Build GRCh38
rs753253312 Chr.3: 121586535-121586535 on Build GRCh38
rs762775007 Chr.3: 121586492-121586492 on Build GRCh38
rs753978095 Chr.3: 121586625-121586625 on Build GRCh38
rs964359498 Chr.3: 121586766-121586766 on Build GRCh38
rs115045570 Chr.3: 121586780-121586780 on Build GRCh38
rs774655554 Chr.3: 121586445-121586445 on Build GRCh38
rs199657126 Chr.3: 121586579-121586579 on Build GRCh38
rs148566270 Chr.3: 121586577-121586577 on Build GRCh38
rs777639306 Chr.3: 121586462-121586462 on Build GRCh38
rs116595666 Chr.3: 121586798-121586798 on Build GRCh38
rs749768536 Chr.3: 121586433-121586433 on Build GRCh38
rs760825265 Chr.3: 121586519-121586519 on Build GRCh38
rs1025078444 Chr.3: 121586655-121586655 on Build GRCh38
rs971605406 Chr.3: 121586657-121586657 on Build GRCh38
rs770578060 Chr.3: 121586472-121586472 on Build GRCh38
rs746650733 Chr.3: 121586468-121586468 on Build GRCh38
rs923025305 Chr.3: 121586789-121586789 on Build GRCh38
rs769724857 Chr.3: 121586485-121586485 on Build GRCh38
rs759424644 Chr.3: 121586478-121586478 on Build GRCh38
rs796073040 Chr.3: 121586481-121586481 on Build GRCh38
rs748506354 Chr.3: 121586492-121586492 on Build GRCh38
rs779173062 Chr.3: 121586443-121586443 on Build GRCh38
rs780165477 Chr.3: 121586425-121586425 on Build GRCh38
rs768453486 Chr.3: 121586595-121586595 on Build GRCh38
rs758890905 Chr.3: 121586536-121586536 on Build GRCh38
rs192920748 Chr.3: 121586488-121586488 on Build GRCh38
rs373740963 Chr.3: 121586529-121586529 on Build GRCh38
rs769152357 Chr.3: 121586581-121586581 on Build GRCh38
rs541978803 Chr.3: 121586602-121586602 on Build GRCh38
rs35191130 Chr.3: 121586828-121586829 on Build GRCh38
rs989761011 Chr.3: 121586764-121586764 on Build GRCh38
rs779516277 Chr.3: 121586533-121586533 on Build GRCh38
rs762546821 Chr.3: 121586600-121586600 on Build GRCh38
rs755443818 Chr.3: 121586440-121586440 on Build GRCh38
rs759730195 Chr.3: 121586650-121586650 on Build GRCh38
rs527905713 Chr.3: 121586761-121586761 on Build GRCh38
rs868809341 Chr.3: 121586434-121586434 on Build GRCh38
rs550926826 Chr.3: 121586842-121586842 on Build GRCh38
rs763871087 Chr.3: 121586513-121586513 on Build GRCh38
rs776140648 Chr.3: 121586475-121586475 on Build GRCh38
rs1012315731 Chr.3: 121586629-121586629 on Build GRCh38
rs778225176 Chr.3: 121586546-121586546 on Build GRCh38
rs895216104 Chr.3: 121586606-121586606 on Build GRCh38
rs576722757 Chr.3: 121586569-121586569 on Build GRCh38
rs914214970 Chr.3: 121586765-121586765 on Build GRCh38
rs770160566 Chr.3: 121586505-121586506 on Build GRCh38
rs956017457 Chr.3: 121586750-121586750 on Build GRCh38
rs772536561 Chr.3: 121586452-121586452 on Build GRCh38

View Full Product Details