Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114678250-114678749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843140 c.1186C>T p.H396Y Chr.1: 114678338-114678338 on Build GRCh38
1294760 c.1175C>G p.S392C Chr.1: 114678349-114678349 on Build GRCh38
4414742 c.996T>C p.Y332Y Chr.1: 114678429-114678429 on Build GRCh38
2177378 c.1126G>A p.E376K Chr.1: 114678398-114678398 on Build GRCh38
6202195 c.986G>T p.R329I Chr.1: 114678439-114678439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759481062 Chr.1: 114678520-114678520 on Build GRCh38
rs376193713 Chr.1: 114678467-114678467 on Build GRCh38
rs762195221 Chr.1: 114678483-114678483 on Build GRCh38
rs933618475 Chr.1: 114678295-114678295 on Build GRCh38
rs757009837 Chr.1: 114678710-114678710 on Build GRCh38
rs781304274 Chr.1: 114678284-114678284 on Build GRCh38
rs1013897127 Chr.1: 114678519-114678519 on Build GRCh38
rs766029049 Chr.1: 114678574-114678575 on Build GRCh38
rs894870923 Chr.1: 114678618-114678619 on Build GRCh38
rs910805762 Chr.1: 114678700-114678700 on Build GRCh38
rs754868231 Chr.1: 114678314-114678314 on Build GRCh38
rs756011257 Chr.1: 114678302-114678302 on Build GRCh38
rs377513152 Chr.1: 114678395-114678395 on Build GRCh38
rs750474668 Chr.1: 114678414-114678414 on Build GRCh38
rs368452649 Chr.1: 114678579-114678579 on Build GRCh38
rs766427528 Chr.1: 114678513-114678513 on Build GRCh38
rs763666390 Chr.1: 114678605-114678605 on Build GRCh38
rs375516946 Chr.1: 114678517-114678517 on Build GRCh38
rs374283776 Chr.1: 114678320-114678320 on Build GRCh38
rs548608037 Chr.1: 114678476-114678476 on Build GRCh38
rs763275867 Chr.1: 114678363-114678363 on Build GRCh38
rs779885195 Chr.1: 114678304-114678304 on Build GRCh38
rs933337855 Chr.1: 114678668-114678668 on Build GRCh38
rs75349593 Chr.1: 114678724-114678724 on Build GRCh38
rs775914313 Chr.1: 114678361-114678361 on Build GRCh38
rs753758389 Chr.1: 114678430-114678430 on Build GRCh38
rs1052461075 Chr.1: 114678303-114678303 on Build GRCh38
rs778793235 Chr.1: 114678436-114678436 on Build GRCh38
rs764505296 Chr.1: 114678365-114678365 on Build GRCh38
rs76816611 Chr.1: 114678715-114678715 on Build GRCh38
rs201878293 Chr.1: 114678461-114678461 on Build GRCh38
rs549830692 Chr.1: 114678555-114678555 on Build GRCh38
rs1037355598 Chr.1: 114678504-114678504 on Build GRCh38
rs767727681 Chr.1: 114678403-114678403 on Build GRCh38
rs571348179 Chr.1: 114678671-114678671 on Build GRCh38
rs754808704 Chr.1: 114678435-114678435 on Build GRCh38
rs753640012 Chr.1: 114678616-114678616 on Build GRCh38
rs978625847 Chr.1: 114678556-114678556 on Build GRCh38
rs777315755 Chr.1: 114678448-114678448 on Build GRCh38
rs142123340 Chr.1: 114678475-114678475 on Build GRCh38
rs150190849 Chr.1: 114678454-114678454 on Build GRCh38
rs774539078 Chr.1: 114678373-114678373 on Build GRCh38
rs772981078 Chr.1: 114678352-114678358 on Build GRCh38
rs751241583 Chr.1: 114678560-114678560 on Build GRCh38
rs769108366 Chr.1: 114678576-114678576 on Build GRCh38
rs61752478 Chr.1: 114678495-114678495 on Build GRCh38
rs745383528 Chr.1: 114678572-114678572 on Build GRCh38
rs913960213 Chr.1: 114678617-114678617 on Build GRCh38
rs988212331 Chr.1: 114678609-114678609 on Build GRCh38
rs1025106955 Chr.1: 114678637-114678637 on Build GRCh38
rs370007374 Chr.1: 114678411-114678411 on Build GRCh38
rs537859575 Chr.1: 114678545-114678545 on Build GRCh38
rs750479609 Chr.1: 114678287-114678287 on Build GRCh38
rs762951803 Chr.1: 114678550-114678550 on Build GRCh38
rs765298679 Chr.1: 114678533-114678533 on Build GRCh38
rs969966808 Chr.1: 114678551-114678551 on Build GRCh38
rs756169459 Chr.1: 114678425-114678425 on Build GRCh38
rs758320481 Chr.1: 114678552-114678552 on Build GRCh38
rs778097856 Chr.1: 114678410-114678410 on Build GRCh38
rs772712902 Chr.1: 114678346-114678346 on Build GRCh38
rs373484604 Chr.1: 114678515-114678515 on Build GRCh38
rs373649754 Chr.1: 114678308-114678308 on Build GRCh38
rs771867356 Chr.1: 114678341-114678341 on Build GRCh38
rs746535692 Chr.1: 114678349-114678349 on Build GRCh38
rs758087316 Chr.1: 114678440-114678440 on Build GRCh38
rs547963174 Chr.1: 114678397-114678397 on Build GRCh38
rs766318137 Chr.1: 114678428-114678428 on Build GRCh38
rs770289186 Chr.1: 114678358-114678358 on Build GRCh38
rs773353828 Chr.1: 114678503-114678503 on Build GRCh38
rs370534764 Chr.1: 114678567-114678567 on Build GRCh38
rs749848289 Chr.1: 114678473-114678473 on Build GRCh38
rs532506042 Chr.1: 114678321-114678321 on Build GRCh38
rs760777657 Chr.1: 114678507-114678507 on Build GRCh38
rs1013353030 Chr.1: 114678630-114678630 on Build GRCh38

View Full Product Details