Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195578970-195579472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOD
Entrez Gene ID
347
Gene Name
apolipoprotein D
Gene Aliases
-
UniGene
Hs.522555
Gene Chromosome Location
Chr.3: 195568702-195584205 on Build GRCh38
Transcripts
NM_001647.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7257218 c.51G>T p.A17A Chr.3: 195579411-195579411 on Build GRCh38
159134 c.74T>C p.L25P Chr.3: 195579388-195579388 on Build GRCh38
4116245 c.52G>A p.A18T Chr.3: 195579410-195579410 on Build GRCh38
4483425 c.26C>T p.S9F Chr.3: 195579436-195579436 on Build GRCh38
4662529 c.29C>T p.A10V Chr.3: 195579433-195579433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749791885 Chr.3: 195579289-195579289 on Build GRCh38
rs574397627 Chr.3: 195579027-195579027 on Build GRCh38
rs944489999 Chr.3: 195579411-195579411 on Build GRCh38
rs5953 Chr.3: 195579348-195579348 on Build GRCh38
rs935310037 Chr.3: 195579126-195579126 on Build GRCh38
rs867618083 Chr.3: 195579079-195579079 on Build GRCh38
rs754512493 Chr.3: 195579141-195579141 on Build GRCh38
rs372722113 Chr.3: 195579322-195579322 on Build GRCh38
rs764190179 Chr.3: 195579412-195579412 on Build GRCh38
rs766193256 Chr.3: 195579399-195579399 on Build GRCh38
rs995099687 Chr.3: 195579075-195579075 on Build GRCh38
rs1042159848 Chr.3: 195579001-195579002 on Build GRCh38
rs767683972 Chr.3: 195579347-195579347 on Build GRCh38
rs866779593 Chr.3: 195579074-195579074 on Build GRCh38
rs776217968 Chr.3: 195579391-195579391 on Build GRCh38
rs776763868 Chr.3: 195579326-195579326 on Build GRCh38
rs979718071 Chr.3: 195579118-195579118 on Build GRCh38
rs543234548 Chr.3: 195579291-195579291 on Build GRCh38
rs746264167 Chr.3: 195579372-195579372 on Build GRCh38
rs753864007 Chr.3: 195579416-195579416 on Build GRCh38
rs559280753 Chr.3: 195579388-195579388 on Build GRCh38
rs553324936 Chr.3: 195579065-195579065 on Build GRCh38
rs771574116 Chr.3: 195579306-195579306 on Build GRCh38
rs150194441 Chr.3: 195579398-195579398 on Build GRCh38
rs757719233 Chr.3: 195579367-195579367 on Build GRCh38
rs769571884 Chr.3: 195579396-195579396 on Build GRCh38
rs754347210 Chr.3: 195579363-195579363 on Build GRCh38
rs747640177 Chr.3: 195579313-195579316 on Build GRCh38
rs775183506 Chr.3: 195579321-195579321 on Build GRCh38
rs140922131 Chr.3: 195579393-195579393 on Build GRCh38
rs780404719 Chr.3: 195579423-195579423 on Build GRCh38
rs1015133601 Chr.3: 195579028-195579028 on Build GRCh38
rs760819270 Chr.3: 195579406-195579406 on Build GRCh38
rs948106178 Chr.3: 195579218-195579218 on Build GRCh38
rs900644173 Chr.3: 195579064-195579064 on Build GRCh38
rs4677691 Chr.3: 195579184-195579184 on Build GRCh38
rs779635482 Chr.3: 195579381-195579381 on Build GRCh38
rs751264654 Chr.3: 195579434-195579434 on Build GRCh38
rs1015123733 Chr.3: 195579320-195579320 on Build GRCh38
rs569788660 Chr.3: 195579438-195579438 on Build GRCh38
rs769227949 Chr.3: 195579383-195579384 on Build GRCh38
rs946586925 Chr.3: 195579091-195579091 on Build GRCh38
rs574392206 Chr.3: 195579047-195579047 on Build GRCh38
rs747712978 Chr.3: 195579451-195579451 on Build GRCh38
rs775747728 Chr.3: 195579405-195579405 on Build GRCh38
rs142890022 Chr.3: 195579373-195579373 on Build GRCh38
rs199897110 Chr.3: 195579371-195579371 on Build GRCh38
rs183848580 Chr.3: 195579177-195579177 on Build GRCh38
rs2280520 Chr.3: 195579012-195579012 on Build GRCh38
rs764102054 Chr.3: 195579361-195579361 on Build GRCh38
rs534671718 Chr.3: 195579049-195579049 on Build GRCh38
rs750935303 Chr.3: 195579420-195579420 on Build GRCh38
rs186067694 Chr.3: 195579178-195579178 on Build GRCh38
rs1024426753 Chr.3: 195579170-195579170 on Build GRCh38
rs34860188 Chr.3: 195579094-195579095 on Build GRCh38
rs911015227 Chr.3: 195579088-195579088 on Build GRCh38
rs780060652 Chr.3: 195579145-195579145 on Build GRCh38
rs1045173450 Chr.3: 195579269-195579269 on Build GRCh38
rs923850853 Chr.3: 195579435-195579435 on Build GRCh38
rs183224694 Chr.3: 195579377-195579377 on Build GRCh38
rs745564078 Chr.3: 195579304-195579304 on Build GRCh38
rs56062729 Chr.3: 195579123-195579123 on Build GRCh38
rs778897431 Chr.3: 195579360-195579360 on Build GRCh38
rs5952 Chr.3: 195579418-195579418 on Build GRCh38
rs959434181 Chr.3: 195579259-195579259 on Build GRCh38
rs199684294 Chr.3: 195579450-195579450 on Build GRCh38
rs151068042 Chr.3: 195579387-195579387 on Build GRCh38
rs960995926 Chr.3: 195579045-195579045 on Build GRCh38
rs1006862223 Chr.3: 195579026-195579026 on Build GRCh38
rs191080620 Chr.3: 195579329-195579329 on Build GRCh38
rs1049741777 Chr.3: 195579117-195579117 on Build GRCh38
rs759829230 Chr.3: 195579345-195579345 on Build GRCh38
rs757303997 Chr.3: 195579417-195579417 on Build GRCh38
rs572152869 Chr.3: 195579327-195579327 on Build GRCh38
rs993778126 Chr.3: 195579331-195579331 on Build GRCh38
rs779080160 Chr.3: 195579303-195579303 on Build GRCh38
rs1033523285 Chr.3: 195579240-195579240 on Build GRCh38

View Full Product Details