Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39287777-39288285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC1
Entrez Gene ID
5981
Gene Name
replication factor C subunit 1
Gene Aliases
A1, MHCBFB, PO-GA, RECC1, RFC, RFC140
UniGene
Hs.507475
Gene Chromosome Location
Chr.4: 39287449-39366381 on Build GRCh38
Transcripts
NM_001204747.1 NM_002913.4 XM_011513730.1 XM_011513731.1 XM_017008528.1

SNP Detail

SNP ID Chromosome Location
rs761587788 Chr.4: 39288029-39288029 on Build GRCh38
rs533283593 Chr.4: 39288039-39288039 on Build GRCh38
rs941622580 Chr.4: 39287932-39287932 on Build GRCh38
rs953260115 Chr.4: 39288030-39288030 on Build GRCh38
rs893680896 Chr.4: 39288025-39288025 on Build GRCh38
rs1046412831 Chr.4: 39288205-39288205 on Build GRCh38
rs948045250 Chr.4: 39287879-39287879 on Build GRCh38
rs56152916 Chr.4: 39287901-39287902 on Build GRCh38
rs916642511 Chr.4: 39288149-39288149 on Build GRCh38
rs972993715 Chr.4: 39288083-39288083 on Build GRCh38
rs760214621 Chr.4: 39288146-39288149 on Build GRCh38
rs932687826 Chr.4: 39288012-39288012 on Build GRCh38
rs79591431 Chr.4: 39288245-39288245 on Build GRCh38
rs55702380 Chr.4: 39288071-39288071 on Build GRCh38
rs147194631 Chr.4: 39287801-39287801 on Build GRCh38
rs906425052 Chr.4: 39287947-39287947 on Build GRCh38
rs545014576 Chr.4: 39287981-39287981 on Build GRCh38
rs983421034 Chr.4: 39287926-39287926 on Build GRCh38
rs774402968 Chr.4: 39288028-39288028 on Build GRCh38
rs573774337 Chr.4: 39288219-39288219 on Build GRCh38
rs1051214465 Chr.4: 39288013-39288013 on Build GRCh38
rs546460828 Chr.4: 39288054-39288054 on Build GRCh38
rs962954061 Chr.4: 39288230-39288230 on Build GRCh38
rs891116152 Chr.4: 39288024-39288024 on Build GRCh38
rs908889816 Chr.4: 39288196-39288199 on Build GRCh38
rs779389583 Chr.4: 39287973-39287973 on Build GRCh38
rs932844705 Chr.4: 39288253-39288253 on Build GRCh38
rs531558013 Chr.4: 39287957-39287957 on Build GRCh38
rs560008120 Chr.4: 39288116-39288116 on Build GRCh38
rs983110273 Chr.4: 39288183-39288183 on Build GRCh38
rs948154585 Chr.4: 39288166-39288166 on Build GRCh38
rs748909027 Chr.4: 39288148-39288149 on Build GRCh38
rs1052256511 Chr.4: 39288000-39288000 on Build GRCh38
rs762817585 Chr.4: 39288222-39288222 on Build GRCh38
rs1057807 Chr.4: 39287853-39287853 on Build GRCh38
rs927980851 Chr.4: 39288238-39288238 on Build GRCh38
rs769592597 Chr.4: 39287950-39287950 on Build GRCh38
rs771784269 Chr.4: 39288095-39288095 on Build GRCh38
rs918752705 Chr.4: 39288089-39288089 on Build GRCh38
rs564008623 Chr.4: 39288032-39288032 on Build GRCh38
rs969742034 Chr.4: 39287834-39287834 on Build GRCh38
rs1046447285 Chr.4: 39287889-39287889 on Build GRCh38
rs773008566 Chr.4: 39288132-39288132 on Build GRCh38
rs529083722 Chr.4: 39288094-39288094 on Build GRCh38
rs375534742 Chr.4: 39288184-39288184 on Build GRCh38
rs1048511559 Chr.4: 39287852-39287852 on Build GRCh38
rs931379886 Chr.4: 39287836-39287836 on Build GRCh38
rs748868932 Chr.4: 39287975-39287975 on Build GRCh38

View Full Product Details