Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44447411-44447904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD8
Entrez Gene ID
386617
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
UniGene
Hs.479644
Gene Chromosome Location
Chr.4: 44173903-44448885 on Build GRCh38
Transcripts
NM_198353.2 XM_011513690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4124842 c.768C>T p.R256R Chr.4: 44447756-44447756 on Build GRCh38
6692133 c.678C>T p.D226D Chr.4: 44447846-44447846 on Build GRCh38
7373312 c.713G>T p.C238F Chr.4: 44447811-44447811 on Build GRCh38
4798484 c.625C>T p.L209F Chr.4: 44447899-44447899 on Build GRCh38
6226525 c.916C>T p.R306C Chr.4: 44447608-44447608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148977351 Chr.4: 44447727-44447727 on Build GRCh38
rs972839315 Chr.4: 44447648-44447648 on Build GRCh38
rs757865317 Chr.4: 44447767-44447767 on Build GRCh38
rs773048313 Chr.4: 44447722-44447722 on Build GRCh38
rs752010504 Chr.4: 44447770-44447770 on Build GRCh38
rs754595486 Chr.4: 44447579-44447579 on Build GRCh38
rs764817862 Chr.4: 44447866-44447866 on Build GRCh38
rs939740294 Chr.4: 44447736-44447736 on Build GRCh38
rs1031846669 Chr.4: 44447870-44447870 on Build GRCh38
rs200013511 Chr.4: 44447764-44447764 on Build GRCh38
rs935082580 Chr.4: 44447476-44447476 on Build GRCh38
rs771159710 Chr.4: 44447526-44447526 on Build GRCh38
rs778380899 Chr.4: 44447609-44447609 on Build GRCh38
rs921513700 Chr.4: 44447634-44447634 on Build GRCh38
rs769540960 Chr.4: 44447495-44447495 on Build GRCh38
rs750203965 Chr.4: 44447585-44447585 on Build GRCh38
rs779825338 Chr.4: 44447532-44447532 on Build GRCh38
rs761800922 Chr.4: 44447565-44447565 on Build GRCh38
rs1052153831 Chr.4: 44447798-44447798 on Build GRCh38
rs117297857 Chr.4: 44447630-44447630 on Build GRCh38
rs759644507 Chr.4: 44447542-44447542 on Build GRCh38
rs778189286 Chr.4: 44447621-44447621 on Build GRCh38
rs942912889 Chr.4: 44447783-44447783 on Build GRCh38
rs753521049 Chr.4: 44447600-44447600 on Build GRCh38
rs760544672 Chr.4: 44447846-44447846 on Build GRCh38
rs774377016 Chr.4: 44447548-44447548 on Build GRCh38
rs911454417 Chr.4: 44447780-44447780 on Build GRCh38
rs754419042 Chr.4: 44447607-44447607 on Build GRCh38
rs566017725 Chr.4: 44447475-44447475 on Build GRCh38
rs758905704 Chr.4: 44447617-44447617 on Build GRCh38
rs906466840 Chr.4: 44447434-44447434 on Build GRCh38
rs762454343 Chr.4: 44447875-44447875 on Build GRCh38
rs745421635 Chr.4: 44447473-44447473 on Build GRCh38
rs780193663 Chr.4: 44447823-44447823 on Build GRCh38
rs541069144 Chr.4: 44447882-44447882 on Build GRCh38
rs1044856796 Chr.4: 44447831-44447831 on Build GRCh38
rs781683655 Chr.4: 44447785-44447785 on Build GRCh38
rs142886340 Chr.4: 44447755-44447755 on Build GRCh38
rs1023475544 Chr.4: 44447575-44447575 on Build GRCh38
rs557968835 Chr.4: 44447497-44447497 on Build GRCh38
rs1022027412 Chr.4: 44447789-44447789 on Build GRCh38
rs768651447 Chr.4: 44447837-44447837 on Build GRCh38
rs539725620 Chr.4: 44447482-44447482 on Build GRCh38
rs746110451 Chr.4: 44447649-44447649 on Build GRCh38
rs752228283 Chr.4: 44447766-44447766 on Build GRCh38
rs770727878 Chr.4: 44447586-44447586 on Build GRCh38
rs905145774 Chr.4: 44447845-44447845 on Build GRCh38
rs955489221 Chr.4: 44447730-44447730 on Build GRCh38
rs977643242 Chr.4: 44447555-44447555 on Build GRCh38
rs932339337 Chr.4: 44447669-44447669 on Build GRCh38
rs574038147 Chr.4: 44447862-44447862 on Build GRCh38
rs766167112 Chr.4: 44447752-44447752 on Build GRCh38
rs770111942 Chr.4: 44447805-44447805 on Build GRCh38
rs1001511874 Chr.4: 44447582-44447582 on Build GRCh38
rs747676954 Chr.4: 44447611-44447611 on Build GRCh38
rs778300219 Chr.4: 44447768-44447768 on Build GRCh38
rs958661958 Chr.4: 44447443-44447443 on Build GRCh38
rs924751109 Chr.4: 44447577-44447577 on Build GRCh38
rs765408963 Chr.4: 44447547-44447547 on Build GRCh38
rs775710206 Chr.4: 44447658-44447658 on Build GRCh38
rs372138598 Chr.4: 44447589-44447589 on Build GRCh38
rs765968801 Chr.4: 44447591-44447591 on Build GRCh38
rs935117875 Chr.4: 44447792-44447792 on Build GRCh38
rs763521548 Chr.4: 44447877-44447877 on Build GRCh38
rs151062882 Chr.4: 44447777-44447777 on Build GRCh38
rs371026799 Chr.4: 44447438-44447438 on Build GRCh38
rs1048077981 Chr.4: 44447681-44447681 on Build GRCh38
rs751142804 Chr.4: 44447881-44447881 on Build GRCh38
rs776923781 Chr.4: 44447631-44447631 on Build GRCh38
rs868158851 Chr.4: 44447644-44447644 on Build GRCh38
rs773135071 Chr.4: 44447839-44447839 on Build GRCh38
rs1043515506 Chr.4: 44447469-44447469 on Build GRCh38
rs776376838 Chr.4: 44447861-44447861 on Build GRCh38
rs979121822 Chr.4: 44447786-44447786 on Build GRCh38
rs905889612 Chr.4: 44447576-44447576 on Build GRCh38
rs909655539 Chr.4: 44447683-44447683 on Build GRCh38
rs770921641 Chr.4: 44447860-44447860 on Build GRCh38
rs749341260 Chr.4: 44447836-44447836 on Build GRCh38
rs762985548 Chr.4: 44447666-44447666 on Build GRCh38
rs1012869015 Chr.4: 44447467-44447467 on Build GRCh38
rs747346931 Chr.4: 44447628-44447628 on Build GRCh38
rs1035658980 Chr.4: 44447441-44447441 on Build GRCh38
rs767468874 Chr.4: 44447672-44447672 on Build GRCh38
rs906323250 Chr.4: 44447758-44447758 on Build GRCh38
rs555582844 Chr.4: 44447790-44447790 on Build GRCh38
rs1017387594 Chr.4: 44447879-44447879 on Build GRCh38
rs1003415049 Chr.4: 44447763-44447763 on Build GRCh38
rs903623958 Chr.4: 44447470-44447470 on Build GRCh38
rs201961501 Chr.4: 44447867-44447867 on Build GRCh38
rs965670478 Chr.4: 44447601-44447601 on Build GRCh38
rs201210650 Chr.4: 44447678-44447678 on Build GRCh38
rs770203655 Chr.4: 44447656-44447656 on Build GRCh38
rs11944873 Chr.4: 44447518-44447518 on Build GRCh38
rs779207636 Chr.4: 44447551-44447551 on Build GRCh38
rs777013649 Chr.4: 44447530-44447530 on Build GRCh38
rs965575244 Chr.4: 44447876-44447876 on Build GRCh38

View Full Product Details