Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48483849-48484349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A4
Entrez Gene ID
201780
Gene Name
solute carrier family 10 member 4
Gene Aliases
P4
UniGene
Hs.132591
Gene Chromosome Location
Chr.4: 48483343-48489524 on Build GRCh38
Transcripts
NM_152679.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5570802 c.511G>A p.V171M Chr.4: 48484072-48484072 on Build GRCh38
7158550 c.588C>A p.L196L Chr.4: 48484149-48484149 on Build GRCh38
5995662 c.295A>T p.T99S Chr.4: 48483856-48483856 on Build GRCh38
3604283 c.517C>T p.L173L Chr.4: 48484078-48484078 on Build GRCh38
1485988 c.478G>A p.A160T Chr.4: 48484039-48484039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019911468 Chr.4: 48484244-48484244 on Build GRCh38
rs959960095 Chr.4: 48483879-48483879 on Build GRCh38
rs768419089 Chr.4: 48484265-48484267 on Build GRCh38
rs112585200 Chr.4: 48484046-48484046 on Build GRCh38
rs771955670 Chr.4: 48483977-48483977 on Build GRCh38
rs369167692 Chr.4: 48484138-48484138 on Build GRCh38
rs756316628 Chr.4: 48484158-48484158 on Build GRCh38
rs184059459 Chr.4: 48483975-48483975 on Build GRCh38
rs763472707 Chr.4: 48484015-48484015 on Build GRCh38
rs1035476926 Chr.4: 48483871-48483871 on Build GRCh38
rs925568161 Chr.4: 48484282-48484282 on Build GRCh38
rs945616290 Chr.4: 48483945-48483945 on Build GRCh38
rs770769951 Chr.4: 48483979-48483979 on Build GRCh38
rs933385813 Chr.4: 48484317-48484317 on Build GRCh38
rs879502049 Chr.4: 48484300-48484300 on Build GRCh38
rs141006219 Chr.4: 48484312-48484312 on Build GRCh38
rs539947639 Chr.4: 48484071-48484071 on Build GRCh38
rs964831123 Chr.4: 48484287-48484287 on Build GRCh38
rs6845250 Chr.4: 48484176-48484176 on Build GRCh38
rs745536626 Chr.4: 48484117-48484117 on Build GRCh38
rs757524706 Chr.4: 48483946-48483946 on Build GRCh38
rs886446517 Chr.4: 48484038-48484038 on Build GRCh38
rs753205598 Chr.4: 48484013-48484013 on Build GRCh38
rs778139760 Chr.4: 48483890-48483890 on Build GRCh38
rs766463451 Chr.4: 48484169-48484169 on Build GRCh38
rs943573839 Chr.4: 48483999-48483999 on Build GRCh38
rs189866168 Chr.4: 48484123-48484123 on Build GRCh38
rs988517423 Chr.4: 48483925-48483925 on Build GRCh38
rs561124613 Chr.4: 48484218-48484218 on Build GRCh38
rs764382889 Chr.4: 48484199-48484199 on Build GRCh38
rs781219189 Chr.4: 48483948-48483948 on Build GRCh38
rs758392580 Chr.4: 48484189-48484189 on Build GRCh38
rs764655825 Chr.4: 48484009-48484009 on Build GRCh38
rs553190797 Chr.4: 48484094-48484094 on Build GRCh38
rs536258981 Chr.4: 48483930-48483930 on Build GRCh38
rs200806965 Chr.4: 48484194-48484194 on Build GRCh38
rs547103558 Chr.4: 48484278-48484278 on Build GRCh38
rs573171087 Chr.4: 48484104-48484104 on Build GRCh38
rs776483776 Chr.4: 48484200-48484200 on Build GRCh38
rs777702602 Chr.4: 48484093-48484093 on Build GRCh38
rs769296941 Chr.4: 48484118-48484118 on Build GRCh38
rs774686147 Chr.4: 48484130-48484130 on Build GRCh38
rs959792194 Chr.4: 48484235-48484235 on Build GRCh38
rs777724620 Chr.4: 48484063-48484063 on Build GRCh38
rs572896419 Chr.4: 48484033-48484033 on Build GRCh38
rs1001396672 Chr.4: 48484177-48484177 on Build GRCh38
rs112201333 Chr.4: 48484316-48484316 on Build GRCh38
rs776277671 Chr.4: 48484096-48484096 on Build GRCh38
rs756012695 Chr.4: 48483894-48483894 on Build GRCh38
rs569593374 Chr.4: 48484155-48484155 on Build GRCh38
rs778854145 Chr.4: 48483967-48483967 on Build GRCh38
rs768435815 Chr.4: 48483984-48483989 on Build GRCh38
rs767780020 Chr.4: 48484151-48484151 on Build GRCh38
rs373281471 Chr.4: 48484201-48484201 on Build GRCh38
rs758462234 Chr.4: 48484078-48484078 on Build GRCh38
rs773166749 Chr.4: 48483978-48483978 on Build GRCh38
rs901950643 Chr.4: 48484016-48484016 on Build GRCh38
rs751960693 Chr.4: 48484028-48484028 on Build GRCh38
rs748073739 Chr.4: 48484067-48484067 on Build GRCh38
rs758721003 Chr.4: 48483880-48483880 on Build GRCh38
rs746027527 Chr.4: 48483958-48483958 on Build GRCh38
rs369977349 Chr.4: 48484195-48484195 on Build GRCh38
rs756154796 Chr.4: 48484047-48484047 on Build GRCh38
rs757467094 Chr.4: 48484031-48484031 on Build GRCh38
rs1047694654 Chr.4: 48484030-48484030 on Build GRCh38
rs764652339 Chr.4: 48484125-48484125 on Build GRCh38
rs750626613 Chr.4: 48484039-48484039 on Build GRCh38
rs774910326 Chr.4: 48484122-48484122 on Build GRCh38
rs912904349 Chr.4: 48483933-48483933 on Build GRCh38
rs368882632 Chr.4: 48483962-48483962 on Build GRCh38
rs1035828019 Chr.4: 48484209-48484209 on Build GRCh38
rs764419294 Chr.4: 48484017-48484017 on Build GRCh38
rs376484754 Chr.4: 48484156-48484156 on Build GRCh38
rs775987776 Chr.4: 48483990-48483990 on Build GRCh38
rs770859967 Chr.4: 48484095-48484095 on Build GRCh38
rs181510011 Chr.4: 48484242-48484242 on Build GRCh38

View Full Product Details