Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56435263-56435758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAICS
Entrez Gene ID
10606
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
Gene Aliases
ADE2, ADE2H1, AIRC, PAIS
UniGene
Hs.518774
Gene Chromosome Location
Chr.4: 56435749-56461368 on Build GRCh38
Transcripts
NM_006452.3 NM_002703.4 XM_011534387.1

Gene Symbol
PPAT
Entrez Gene ID
5471
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Gene Aliases
ATASE, GPAT, PRAT
UniGene
Hs.331420
Gene Chromosome Location
Chr.4: 56393362-56435636 on Build GRCh38
Transcripts
NM_006452.3 NM_002703.4 XM_011534387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373521 c.101C>T p.T34I Chr.4: 56435377-56435377 on Build GRCh38
4125290 c.59C>G p.S20* Chr.4: 56435419-56435419 on Build GRCh38
6275150 c.109C>A p.L37I Chr.4: 56435369-56435369 on Build GRCh38
1694422 c.33A>T p.E11D Chr.4: 56435445-56435445 on Build GRCh38
6469625 c.87A>G p.V29V Chr.4: 56435391-56435391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575699835 Chr.4: 56435585-56435585 on Build GRCh38
rs969535050 Chr.4: 56435666-56435666 on Build GRCh38
rs1008704280 Chr.4: 56435597-56435597 on Build GRCh38
rs776938734 Chr.4: 56435312-56435312 on Build GRCh38
rs540988880 Chr.4: 56435510-56435510 on Build GRCh38
rs888967127 Chr.4: 56435595-56435595 on Build GRCh38
rs569325646 Chr.4: 56435406-56435406 on Build GRCh38
rs3822001 Chr.4: 56435584-56435584 on Build GRCh38
rs756781025 Chr.4: 56435333-56435333 on Build GRCh38
rs375418740 Chr.4: 56435548-56435548 on Build GRCh38
rs772747599 Chr.4: 56435529-56435529 on Build GRCh38
rs1046405755 Chr.4: 56435315-56435315 on Build GRCh38
rs555078164 Chr.4: 56435383-56435383 on Build GRCh38
rs774980557 Chr.4: 56435622-56435622 on Build GRCh38
rs1031709970 Chr.4: 56435534-56435534 on Build GRCh38
rs902932887 Chr.4: 56435614-56435614 on Build GRCh38
rs754936603 Chr.4: 56435367-56435367 on Build GRCh38
rs754845652 Chr.4: 56435495-56435495 on Build GRCh38
rs765989683 Chr.4: 56435492-56435492 on Build GRCh38
rs919740742 Chr.4: 56435672-56435672 on Build GRCh38
rs201978310 Chr.4: 56435528-56435528 on Build GRCh38
rs554389201 Chr.4: 56435553-56435553 on Build GRCh38
rs761246399 Chr.4: 56435454-56435454 on Build GRCh38
rs1028821471 Chr.4: 56435468-56435468 on Build GRCh38
rs1018678364 Chr.4: 56435516-56435516 on Build GRCh38
rs774868196 Chr.4: 56435433-56435433 on Build GRCh38
rs781324117 Chr.4: 56435499-56435499 on Build GRCh38
rs763444167 Chr.4: 56435322-56435322 on Build GRCh38
rs998960998 Chr.4: 56435616-56435616 on Build GRCh38
rs564367114 Chr.4: 56435693-56435693 on Build GRCh38
rs745970882 Chr.4: 56435502-56435502 on Build GRCh38
rs755787806 Chr.4: 56435488-56435488 on Build GRCh38
rs60097571 Chr.4: 56435567-56435567 on Build GRCh38
rs989710463 Chr.4: 56435654-56435654 on Build GRCh38
rs948127213 Chr.4: 56435285-56435285 on Build GRCh38
rs972501082 Chr.4: 56435731-56435731 on Build GRCh38
rs1040422031 Chr.4: 56435705-56435705 on Build GRCh38
rs573844517 Chr.4: 56435473-56435473 on Build GRCh38
rs370765017 Chr.4: 56435424-56435424 on Build GRCh38
rs202181751 Chr.4: 56435503-56435503 on Build GRCh38
rs746907119 Chr.4: 56435319-56435320 on Build GRCh38
rs552713157 Chr.4: 56435340-56435340 on Build GRCh38
rs982173761 Chr.4: 56435568-56435568 on Build GRCh38
rs755376801 Chr.4: 56435504-56435504 on Build GRCh38
rs758097056 Chr.4: 56435610-56435610 on Build GRCh38
rs927219361 Chr.4: 56435546-56435546 on Build GRCh38
rs6843073 Chr.4: 56435515-56435515 on Build GRCh38
rs776654985 Chr.4: 56435401-56435401 on Build GRCh38
rs775798687 Chr.4: 56435419-56435419 on Build GRCh38
rs773422374 Chr.4: 56435304-56435304 on Build GRCh38
rs189152163 Chr.4: 56435549-56435549 on Build GRCh38
rs572353430 Chr.4: 56435626-56435626 on Build GRCh38
rs899040195 Chr.4: 56435418-56435418 on Build GRCh38
rs1032666518 Chr.4: 56435638-56435638 on Build GRCh38
rs769011905 Chr.4: 56435514-56435514 on Build GRCh38
rs778956686 Chr.4: 56435496-56435496 on Build GRCh38
rs1047854564 Chr.4: 56435557-56435557 on Build GRCh38
rs1019211343 Chr.4: 56435316-56435316 on Build GRCh38
rs142572514 Chr.4: 56435421-56435421 on Build GRCh38
rs533162729 Chr.4: 56435702-56435702 on Build GRCh38
rs749612783 Chr.4: 56435509-56435509 on Build GRCh38
rs958672881 Chr.4: 56435664-56435664 on Build GRCh38
rs769973255 Chr.4: 56435416-56435416 on Build GRCh38
rs549973708 Chr.4: 56435733-56435733 on Build GRCh38
rs1023812554 Chr.4: 56435660-56435660 on Build GRCh38
rs765642098 Chr.4: 56435314-56435314 on Build GRCh38
rs58548914 Chr.4: 56435519-56435519 on Build GRCh38
rs982206417 Chr.4: 56435668-56435668 on Build GRCh38
rs753664851 Chr.4: 56435493-56435493 on Build GRCh38
rs540958517 Chr.4: 56435670-56435670 on Build GRCh38
rs1009967280 Chr.4: 56435603-56435603 on Build GRCh38
rs747126936 Chr.4: 56435310-56435310 on Build GRCh38
rs988871758 Chr.4: 56435457-56435457 on Build GRCh38
rs541461683 Chr.4: 56435594-56435594 on Build GRCh38
rs757230006 Chr.4: 56435498-56435498 on Build GRCh38
rs777374484 Chr.4: 56435695-56435695 on Build GRCh38
rs779046474 Chr.4: 56435376-56435376 on Build GRCh38
rs1047810894 Chr.4: 56435684-56435684 on Build GRCh38
rs375080631 Chr.4: 56435482-56435482 on Build GRCh38
rs1050272780 Chr.4: 56435589-56435589 on Build GRCh38
rs754502923 Chr.4: 56435514-56435515 on Build GRCh38
rs1036360927 Chr.4: 56435577-56435577 on Build GRCh38
rs888895280 Chr.4: 56435483-56435483 on Build GRCh38
rs779886575 Chr.4: 56435352-56435352 on Build GRCh38
rs59010837 Chr.4: 56435570-56435570 on Build GRCh38
rs2176786 Chr.4: 56435698-56435698 on Build GRCh38
rs929815786 Chr.4: 56435680-56435680 on Build GRCh38
rs749162002 Chr.4: 56435366-56435366 on Build GRCh38
rs996731218 Chr.4: 56435587-56435587 on Build GRCh38
rs60112184 Chr.4: 56435602-56435602 on Build GRCh38
rs958555547 Chr.4: 56435643-56435643 on Build GRCh38
rs200428268 Chr.4: 56435324-56435324 on Build GRCh38
rs940689151 Chr.4: 56435575-56435575 on Build GRCh38
rs750095191 Chr.4: 56435337-56435337 on Build GRCh38
rs751044168 Chr.4: 56435328-56435328 on Build GRCh38
rs775773081 Chr.4: 56435319-56435319 on Build GRCh38
rs767037962 Chr.4: 56435335-56435335 on Build GRCh38
rs752689448 Chr.4: 56435497-56435497 on Build GRCh38

View Full Product Details