Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56448518-56448952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAICS
Entrez Gene ID
10606
Gene Name
phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase
Gene Aliases
ADE2, ADE2H1, AIRC, PAIS
UniGene
Hs.518774
Gene Chromosome Location
Chr.4: 56435749-56461368 on Build GRCh38
Transcripts
NM_001079524.1 NM_001079525.1 NM_006452.3 XM_011534366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1430312 c.592A>G p.I198V Chr.4: 56448538-56448538 on Build GRCh38
7176005 c.573G>C p.M191I Chr.4: 56448519-56448519 on Build GRCh38
3826027 c.598G>C p.E200Q Chr.4: 56448544-56448544 on Build GRCh38
6167907 c.603A>G p.I201M Chr.4: 56448549-56448549 on Build GRCh38
1056282 c.742C>T p.R248* Chr.4: 56448800-56448800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758746421 Chr.4: 56448841-56448842 on Build GRCh38
rs781279911 Chr.4: 56448673-56448673 on Build GRCh38
rs780099290 Chr.4: 56448598-56448598 on Build GRCh38
rs751597050 Chr.4: 56448846-56448846 on Build GRCh38
rs755977602 Chr.4: 56448597-56448597 on Build GRCh38
rs994962090 Chr.4: 56448721-56448723 on Build GRCh38
rs761243513 Chr.4: 56448801-56448801 on Build GRCh38
rs1054045389 Chr.4: 56448653-56448653 on Build GRCh38
rs775884727 Chr.4: 56448740-56448740 on Build GRCh38
rs1008543492 Chr.4: 56448744-56448744 on Build GRCh38
rs1027125737 Chr.4: 56448872-56448872 on Build GRCh38
rs777876017 Chr.4: 56448619-56448619 on Build GRCh38
rs370383263 Chr.4: 56448646-56448646 on Build GRCh38
rs190379455 Chr.4: 56448784-56448784 on Build GRCh38
rs562519099 Chr.4: 56448920-56448920 on Build GRCh38
rs985512623 Chr.4: 56448921-56448921 on Build GRCh38
rs763578528 Chr.4: 56448545-56448545 on Build GRCh38
rs966687439 Chr.4: 56448883-56448883 on Build GRCh38
rs773900006 Chr.4: 56448787-56448787 on Build GRCh38
rs750175213 Chr.4: 56448593-56448593 on Build GRCh38
rs927967891 Chr.4: 56448910-56448910 on Build GRCh38
rs894952468 Chr.4: 56448662-56448662 on Build GRCh38
rs56660002 Chr.4: 56448886-56448886 on Build GRCh38
rs562754907 Chr.4: 56448876-56448876 on Build GRCh38
rs1055676490 Chr.4: 56448686-56448686 on Build GRCh38
rs746160982 Chr.4: 56448706-56448706 on Build GRCh38
rs989966157 Chr.4: 56448866-56448866 on Build GRCh38
rs763421838 Chr.4: 56448753-56448753 on Build GRCh38
rs1015873524 Chr.4: 56448790-56448790 on Build GRCh38
rs796368719 Chr.4: 56448598-56448612 on Build GRCh38
rs753847420 Chr.4: 56448604-56448604 on Build GRCh38
rs982078334 Chr.4: 56448897-56448897 on Build GRCh38
rs6825812 Chr.4: 56448820-56448820 on Build GRCh38
rs1018806692 Chr.4: 56448765-56448765 on Build GRCh38
rs377203074 Chr.4: 56448629-56448629 on Build GRCh38
rs906235902 Chr.4: 56448762-56448762 on Build GRCh38
rs11549976 Chr.4: 56448739-56448739 on Build GRCh38
rs1046139657 Chr.4: 56448760-56448760 on Build GRCh38
rs755014046 Chr.4: 56448613-56448613 on Build GRCh38
rs750557169 Chr.4: 56448597-56448597 on Build GRCh38
rs751330271 Chr.4: 56448851-56448865 on Build GRCh38
rs199735757 Chr.4: 56448840-56448841 on Build GRCh38
rs1019653979 Chr.4: 56448922-56448922 on Build GRCh38
rs766254021 Chr.4: 56448815-56448815 on Build GRCh38
rs761357624 Chr.4: 56448842-56448843 on Build GRCh38
rs754959240 Chr.4: 56448840-56448840 on Build GRCh38
rs767134289 Chr.4: 56448587-56448587 on Build GRCh38
rs112629919 Chr.4: 56448613-56448627 on Build GRCh38
rs762387719 Chr.4: 56448542-56448542 on Build GRCh38
rs531428035 Chr.4: 56448899-56448899 on Build GRCh38
rs760497195 Chr.4: 56448809-56448809 on Build GRCh38
rs778771563 Chr.4: 56448842-56448842 on Build GRCh38
rs182572109 Chr.4: 56448925-56448925 on Build GRCh38
rs750075251 Chr.4: 56448808-56448808 on Build GRCh38
rs751073813 Chr.4: 56448583-56448583 on Build GRCh38

View Full Product Details