Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158895633-158896172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
UniGene
Hs.652441
Gene Chromosome Location
Chr.4: 158768995-158908050 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447877 c.3183G>A p.E1061E Chr.4: 158895782-158895782 on Build GRCh38
5632000 c.1195C>A p.L399I Chr.4: 158895816-158895816 on Build GRCh38
5995758 c.1131C>T p.C377C Chr.4: 158895752-158895752 on Build GRCh38
5060211 c.1236C>T p.V412V Chr.4: 158895857-158895857 on Build GRCh38
5014463 c.1196T>G p.L399R Chr.4: 158895817-158895817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952319311 Chr.4: 158895974-158895974 on Build GRCh38
rs765939971 Chr.4: 158895787-158895787 on Build GRCh38
rs915045892 Chr.4: 158896066-158896066 on Build GRCh38
rs749576660 Chr.4: 158895856-158895856 on Build GRCh38
rs771278684 Chr.4: 158895858-158895858 on Build GRCh38
rs34382174 Chr.4: 158895973-158895973 on Build GRCh38
rs924642261 Chr.4: 158896022-158896022 on Build GRCh38
rs985280183 Chr.4: 158895663-158895663 on Build GRCh38
rs147211971 Chr.4: 158896073-158896073 on Build GRCh38
rs946555059 Chr.4: 158896070-158896070 on Build GRCh38
rs759956960 Chr.4: 158895932-158895932 on Build GRCh38
rs986789200 Chr.4: 158895713-158895713 on Build GRCh38
rs993246259 Chr.4: 158895960-158895960 on Build GRCh38
rs375767543 Chr.4: 158895905-158895905 on Build GRCh38
rs756585371 Chr.4: 158895829-158895829 on Build GRCh38
rs778257465 Chr.4: 158895853-158895853 on Build GRCh38
rs567637059 Chr.4: 158895904-158895904 on Build GRCh38
rs201986504 Chr.4: 158895819-158895819 on Build GRCh38
rs904828002 Chr.4: 158895912-158895912 on Build GRCh38
rs917304176 Chr.4: 158896147-158896147 on Build GRCh38
rs200623773 Chr.4: 158895740-158895740 on Build GRCh38
rs761016860 Chr.4: 158896036-158896036 on Build GRCh38
rs1044928028 Chr.4: 158896072-158896072 on Build GRCh38
rs543083054 Chr.4: 158896131-158896131 on Build GRCh38
rs778867777 Chr.4: 158895874-158895874 on Build GRCh38
rs924674917 Chr.4: 158895806-158895806 on Build GRCh38
rs201372619 Chr.4: 158895762-158895762 on Build GRCh38
rs1034677079 Chr.4: 158895922-158895922 on Build GRCh38
rs751015157 Chr.4: 158895812-158895812 on Build GRCh38
rs910517430 Chr.4: 158895753-158895753 on Build GRCh38
rs769626331 Chr.4: 158895897-158895897 on Build GRCh38
rs1018289449 Chr.4: 158895998-158895998 on Build GRCh38
rs776592458 Chr.4: 158895894-158895894 on Build GRCh38
rs897570570 Chr.4: 158895931-158895931 on Build GRCh38
rs911100022 Chr.4: 158895699-158895699 on Build GRCh38
rs897497202 Chr.4: 158896122-158896122 on Build GRCh38
rs990236989 Chr.4: 158896045-158896045 on Build GRCh38
rs577297410 Chr.4: 158896097-158896097 on Build GRCh38
rs557371471 Chr.4: 158895771-158895771 on Build GRCh38
rs941590817 Chr.4: 158895966-158895966 on Build GRCh38
rs745905202 Chr.4: 158895884-158895884 on Build GRCh38
rs967894378 Chr.4: 158896082-158896082 on Build GRCh38
rs939304260 Chr.4: 158896085-158896085 on Build GRCh38
rs202019221 Chr.4: 158895783-158895783 on Build GRCh38
rs1052261781 Chr.4: 158895859-158895859 on Build GRCh38
rs761621214 Chr.4: 158895770-158895770 on Build GRCh38
rs764028732 Chr.4: 158895727-158895727 on Build GRCh38
rs768503956 Chr.4: 158895892-158895892 on Build GRCh38
rs976070777 Chr.4: 158896018-158896018 on Build GRCh38
rs750116846 Chr.4: 158895781-158895781 on Build GRCh38
rs867519538 Chr.4: 158896060-158896060 on Build GRCh38
rs979301664 Chr.4: 158896092-158896092 on Build GRCh38
rs748489884 Chr.4: 158895828-158895828 on Build GRCh38
rs966345225 Chr.4: 158896002-158896002 on Build GRCh38
rs558050940 Chr.4: 158896141-158896141 on Build GRCh38
rs781687627 Chr.4: 158895820-158895820 on Build GRCh38
rs934731368 Chr.4: 158896037-158896037 on Build GRCh38
rs192618522 Chr.4: 158895997-158895997 on Build GRCh38
rs896804699 Chr.4: 158895982-158895982 on Build GRCh38
rs993171741 Chr.4: 158896123-158896123 on Build GRCh38
rs764246243 Chr.4: 158896124-158896124 on Build GRCh38
rs956038809 Chr.4: 158896120-158896120 on Build GRCh38
rs774480682 Chr.4: 158895719-158895719 on Build GRCh38
rs952325559 Chr.4: 158895677-158895677 on Build GRCh38
rs760600238 Chr.4: 158895724-158895724 on Build GRCh38
rs370105773 Chr.4: 158895917-158895917 on Build GRCh38
rs946616299 Chr.4: 158895898-158895898 on Build GRCh38
rs771180731 Chr.4: 158895715-158895715 on Build GRCh38
rs184840625 Chr.4: 158896001-158896001 on Build GRCh38

View Full Product Details