Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158915088-158915600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5602995 c.540G>A p.K180K Chr.4: 158915176-158915176 on Build GRCh38
4138108 c.403G>A p.A135T Chr.4: 158915313-158915313 on Build GRCh38
5865985 c.523C>T p.P175S Chr.4: 158915193-158915193 on Build GRCh38
4547219 c.417G>T p.W139C Chr.4: 158915299-158915299 on Build GRCh38
1052706 c.525G>A p.P175P Chr.4: 158915191-158915191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182710806 Chr.4: 158915154-158915154 on Build GRCh38
rs781223408 Chr.4: 158915284-158915284 on Build GRCh38
rs369536072 Chr.4: 158915348-158915348 on Build GRCh38
rs1044722820 Chr.4: 158915393-158915393 on Build GRCh38
rs758312276 Chr.4: 158915368-158915368 on Build GRCh38
rs1035129154 Chr.4: 158915296-158915296 on Build GRCh38
rs754642476 Chr.4: 158915122-158915122 on Build GRCh38
rs879012850 Chr.4: 158915492-158915492 on Build GRCh38
rs768627086 Chr.4: 158915133-158915133 on Build GRCh38
rs993590070 Chr.4: 158915548-158915548 on Build GRCh38
rs922385596 Chr.4: 158915498-158915498 on Build GRCh38
rs562253193 Chr.4: 158915439-158915439 on Build GRCh38
rs895732603 Chr.4: 158915205-158915205 on Build GRCh38
rs771561802 Chr.4: 158915220-158915220 on Build GRCh38
rs78052401 Chr.4: 158915184-158915184 on Build GRCh38
rs368339639 Chr.4: 158915145-158915145 on Build GRCh38
rs764732646 Chr.4: 158915226-158915226 on Build GRCh38
rs561625731 Chr.4: 158915558-158915558 on Build GRCh38
rs976882042 Chr.4: 158915490-158915490 on Build GRCh38
rs374985341 Chr.4: 158915398-158915398 on Build GRCh38
rs200370568 Chr.4: 158915309-158915309 on Build GRCh38
rs762204226 Chr.4: 158915231-158915231 on Build GRCh38
rs781005416 Chr.4: 158915297-158915297 on Build GRCh38
rs764828904 Chr.4: 158915262-158915262 on Build GRCh38
rs572807293 Chr.4: 158915314-158915314 on Build GRCh38
rs757771741 Chr.4: 158915192-158915192 on Build GRCh38
rs772728934 Chr.4: 158915294-158915295 on Build GRCh38
rs939378761 Chr.4: 158915575-158915575 on Build GRCh38
rs267600064 Chr.4: 158915293-158915293 on Build GRCh38
rs10517695 Chr.4: 158915315-158915315 on Build GRCh38
rs750750550 Chr.4: 158915244-158915244 on Build GRCh38
rs771781181 Chr.4: 158915302-158915302 on Build GRCh38
rs934484434 Chr.4: 158915186-158915186 on Build GRCh38
rs756065939 Chr.4: 158915168-158915168 on Build GRCh38
rs778669506 Chr.4: 158915362-158915362 on Build GRCh38
rs375166042 Chr.4: 158915166-158915166 on Build GRCh38
rs745594193 Chr.4: 158915301-158915301 on Build GRCh38
rs373455711 Chr.4: 158915390-158915390 on Build GRCh38
rs749427245 Chr.4: 158915283-158915283 on Build GRCh38
rs1037745447 Chr.4: 158915525-158915525 on Build GRCh38
rs766537819 Chr.4: 158915250-158915250 on Build GRCh38
rs754279579 Chr.4: 158915140-158915140 on Build GRCh38
rs200938327 Chr.4: 158915190-158915190 on Build GRCh38
rs189786194 Chr.4: 158915144-158915144 on Build GRCh38
rs988364710 Chr.4: 158915506-158915506 on Build GRCh38
rs777647810 Chr.4: 158915170-158915170 on Build GRCh38
rs949957749 Chr.4: 158915390-158915391 on Build GRCh38
rs760219133 Chr.4: 158915224-158915224 on Build GRCh38
rs776268661 Chr.4: 158915313-158915313 on Build GRCh38
rs578126763 Chr.4: 158915379-158915379 on Build GRCh38
rs71609016 Chr.4: 158915482-158915482 on Build GRCh38
rs556094378 Chr.4: 158915295-158915295 on Build GRCh38
rs765693195 Chr.4: 158915232-158915232 on Build GRCh38
rs963701070 Chr.4: 158915472-158915472 on Build GRCh38
rs935460659 Chr.4: 158915503-158915503 on Build GRCh38
rs768331533 Chr.4: 158915311-158915311 on Build GRCh38
rs905170926 Chr.4: 158915568-158915568 on Build GRCh38
rs934448413 Chr.4: 158915350-158915350 on Build GRCh38
rs763185633 Chr.4: 158915319-158915319 on Build GRCh38
rs199776336 Chr.4: 158915505-158915505 on Build GRCh38
rs746718007 Chr.4: 158915384-158915384 on Build GRCh38
rs376450620 Chr.4: 158915256-158915256 on Build GRCh38
rs762381800 Chr.4: 158915304-158915304 on Build GRCh38
rs777318575 Chr.4: 158915230-158915230 on Build GRCh38
rs530312270 Chr.4: 158915331-158915331 on Build GRCh38
rs747031540 Chr.4: 158915167-158915167 on Build GRCh38
rs749189432 Chr.4: 158915181-158915181 on Build GRCh38
rs201167295 Chr.4: 158915285-158915285 on Build GRCh38
rs77199000 Chr.4: 158915533-158915533 on Build GRCh38
rs757235541 Chr.4: 158915361-158915361 on Build GRCh38
rs946978893 Chr.4: 158915537-158915537 on Build GRCh38
rs946695688 Chr.4: 158915277-158915277 on Build GRCh38
rs747170092 Chr.4: 158915433-158915433 on Build GRCh38
rs752649084 Chr.4: 158915356-158915356 on Build GRCh38
rs779340324 Chr.4: 158915249-158915249 on Build GRCh38
rs747561638 Chr.4: 158915406-158915406 on Build GRCh38
rs776396704 Chr.4: 158915162-158915164 on Build GRCh38
rs767647899 Chr.4: 158915355-158915355 on Build GRCh38
rs550026032 Chr.4: 158915223-158915223 on Build GRCh38
rs751670157 Chr.4: 158915252-158915252 on Build GRCh38
rs766697533 Chr.4: 158915322-158915322 on Build GRCh38
rs558298502 Chr.4: 158915354-158915354 on Build GRCh38
rs761567622 Chr.4: 158915177-158915177 on Build GRCh38
rs773646550 Chr.4: 158915316-158915316 on Build GRCh38
rs750267314 Chr.4: 158915366-158915366 on Build GRCh38
rs917086552 Chr.4: 158915380-158915380 on Build GRCh38
rs751825568 Chr.4: 158915344-158915344 on Build GRCh38
rs185999487 Chr.4: 158915195-158915195 on Build GRCh38
rs758536190 Chr.4: 158915156-158915156 on Build GRCh38

View Full Product Details