Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1258416-1258907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERT
Entrez Gene ID
7015
Gene Name
telomerase reverse transcriptase
Gene Aliases
CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TP2, TRT, hEST2, hTRT
UniGene
Hs.492203
Gene Chromosome Location
Chr.5: 1253167-1295626 on Build GRCh38
Transcripts
NM_001193376.1 NM_198253.2 XM_017009796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4486130 c.3015C>T p.L1005L Chr.5: 1258615-1258615 on Build GRCh38
3135502 c.2991G>A p.V997V Chr.5: 1258639-1258639 on Build GRCh38
5958331 c.2982C>G p.L994L Chr.5: 1258648-1258648 on Build GRCh38
7232273 c.2971-1G>T p.? Chr.5: 1258660-1258660 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs137919228 Chr.5: 1258792-1258792 on Build GRCh38
rs966548088 Chr.5: 1258802-1258802 on Build GRCh38
rs369172003 Chr.5: 1258843-1258843 on Build GRCh38
rs939507671 Chr.5: 1258652-1258652 on Build GRCh38
rs750187788 Chr.5: 1258663-1258663 on Build GRCh38
rs571161655 Chr.5: 1258801-1258801 on Build GRCh38
rs768976875 Chr.5: 1258580-1258580 on Build GRCh38
rs376266401 Chr.5: 1258639-1258639 on Build GRCh38
rs1035424242 Chr.5: 1258442-1258442 on Build GRCh38
rs371500615 Chr.5: 1258591-1258591 on Build GRCh38
rs774718903 Chr.5: 1258586-1258586 on Build GRCh38
rs35300412 Chr.5: 1258859-1258859 on Build GRCh38
rs1056756363 Chr.5: 1258833-1258833 on Build GRCh38
rs35092866 Chr.5: 1258552-1258552 on Build GRCh38
rs758294954 Chr.5: 1258681-1258681 on Build GRCh38
rs759222965 Chr.5: 1258872-1258872 on Build GRCh38
rs976724733 Chr.5: 1258756-1258756 on Build GRCh38
rs762178169 Chr.5: 1258648-1258648 on Build GRCh38
rs928391937 Chr.5: 1258873-1258873 on Build GRCh38
rs1023035784 Chr.5: 1258484-1258484 on Build GRCh38
rs985917683 Chr.5: 1258786-1258786 on Build GRCh38
rs999787664 Chr.5: 1258880-1258880 on Build GRCh38
rs770382447 Chr.5: 1258818-1258818 on Build GRCh38
rs186849858 Chr.5: 1258529-1258529 on Build GRCh38
rs145270195 Chr.5: 1258688-1258688 on Build GRCh38
rs866575708 Chr.5: 1258604-1258604 on Build GRCh38
rs895340018 Chr.5: 1258668-1258668 on Build GRCh38
rs765078035 Chr.5: 1258662-1258662 on Build GRCh38
rs375865342 Chr.5: 1258817-1258817 on Build GRCh38
rs955833048 Chr.5: 1258545-1258545 on Build GRCh38
rs573030083 Chr.5: 1258487-1258487 on Build GRCh38
rs746511287 Chr.5: 1258560-1258560 on Build GRCh38
rs941352340 Chr.5: 1258612-1258612 on Build GRCh38
rs773607043 Chr.5: 1258750-1258750 on Build GRCh38
rs772765233 Chr.5: 1258723-1258723 on Build GRCh38
rs544478890 Chr.5: 1258574-1258574 on Build GRCh38
rs758392233 Chr.5: 1258443-1258443 on Build GRCh38
rs35867091 Chr.5: 1258444-1258445 on Build GRCh38
rs775582757 Chr.5: 1258603-1258603 on Build GRCh38
rs374686436 Chr.5: 1258570-1258570 on Build GRCh38
rs776966669 Chr.5: 1258642-1258642 on Build GRCh38
rs112509836 Chr.5: 1258496-1258496 on Build GRCh38
rs978640977 Chr.5: 1258523-1258523 on Build GRCh38
rs112519466 Chr.5: 1258445-1258446 on Build GRCh38
rs953939979 Chr.5: 1258782-1258782 on Build GRCh38
rs373707255 Chr.5: 1258687-1258687 on Build GRCh38
rs746902908 Chr.5: 1258549-1258549 on Build GRCh38
rs1036120126 Chr.5: 1258654-1258654 on Build GRCh38
rs1029441101 Chr.5: 1258836-1258836 on Build GRCh38
rs866369624 Chr.5: 1258650-1258650 on Build GRCh38
rs533921916 Chr.5: 1258510-1258510 on Build GRCh38
rs906870073 Chr.5: 1258440-1258440 on Build GRCh38
rs878855303 Chr.5: 1258669-1258669 on Build GRCh38
rs760659103 Chr.5: 1258624-1258624 on Build GRCh38
rs777670571 Chr.5: 1258475-1258475 on Build GRCh38
rs953606922 Chr.5: 1258840-1258840 on Build GRCh38
rs143487428 Chr.5: 1258853-1258854 on Build GRCh38
rs750355020 Chr.5: 1258550-1258550 on Build GRCh38
rs754453683 Chr.5: 1258689-1258689 on Build GRCh38
rs1029889537 Chr.5: 1258764-1258764 on Build GRCh38
rs869039774 Chr.5: 1258574-1258574 on Build GRCh38
rs545138998 Chr.5: 1258697-1258697 on Build GRCh38
rs909345427 Chr.5: 1258850-1258850 on Build GRCh38
rs1011488595 Chr.5: 1258733-1258733 on Build GRCh38
rs1021448272 Chr.5: 1258530-1258530 on Build GRCh38
rs372276021 Chr.5: 1258558-1258558 on Build GRCh38
rs771262935 Chr.5: 1258579-1258579 on Build GRCh38
rs904298924 Chr.5: 1258871-1258871 on Build GRCh38
rs999539470 Chr.5: 1258797-1258797 on Build GRCh38
rs746348646 Chr.5: 1258590-1258590 on Build GRCh38
rs960072923 Chr.5: 1258470-1258470 on Build GRCh38
rs111466735 Chr.5: 1258572-1258572 on Build GRCh38
rs369797524 Chr.5: 1258707-1258707 on Build GRCh38
rs553820486 Chr.5: 1258441-1258441 on Build GRCh38
rs549308466 Chr.5: 1258806-1258806 on Build GRCh38

View Full Product Details