Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69262208-69262751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK7
Entrez Gene ID
1022
Gene Name
cyclin dependent kinase 7
Gene Aliases
CAK, CAK1, CDKN7, HCAK, MO15, STK1, p39MO15
UniGene
Hs.184298
Gene Chromosome Location
Chr.5: 69234795-69277430 on Build GRCh38
Transcripts
NM_001324069.1 NM_001324070.1 NM_001324071.1 NM_001324072.1 NM_001324074.1 NM_001324075.1 NM_001324077.1 NM_001324078.1 NM_001799.3 NR_136690.1 XM_011543094.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470491 c.534T>G p.Y178* Chr.5: 69262211-69262211 on Build GRCh38
3072102 c.617T>C p.L206S Chr.5: 69262294-69262294 on Build GRCh38
1197035 c.586A>G p.M196V Chr.5: 69262263-69262263 on Build GRCh38
6625594 c.602G>A p.C201Y Chr.5: 69262279-69262279 on Build GRCh38
6382709 c.591G>A p.W197* Chr.5: 69262268-69262268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187548867 Chr.5: 69262546-69262546 on Build GRCh38
rs1052393446 Chr.5: 69262705-69262705 on Build GRCh38
rs1026521289 Chr.5: 69262500-69262500 on Build GRCh38
rs112347522 Chr.5: 69262466-69262466 on Build GRCh38
rs1013424601 Chr.5: 69262281-69262281 on Build GRCh38
rs759480531 Chr.5: 69262241-69262242 on Build GRCh38
rs542802709 Chr.5: 69262432-69262432 on Build GRCh38
rs925912728 Chr.5: 69262680-69262684 on Build GRCh38
rs887734817 Chr.5: 69262592-69262592 on Build GRCh38
rs985386756 Chr.5: 69262410-69262410 on Build GRCh38
rs934643107 Chr.5: 69262692-69262692 on Build GRCh38
rs563554562 Chr.5: 69262369-69262369 on Build GRCh38
rs1004786471 Chr.5: 69262611-69262611 on Build GRCh38
rs1048752495 Chr.5: 69262581-69262581 on Build GRCh38
rs1015170158 Chr.5: 69262323-69262323 on Build GRCh38
rs367846887 Chr.5: 69262311-69262311 on Build GRCh38
rs910651266 Chr.5: 69262562-69262562 on Build GRCh38
rs201296615 Chr.5: 69262668-69262668 on Build GRCh38
rs998664631 Chr.5: 69262346-69262346 on Build GRCh38
rs149662675 Chr.5: 69262684-69262684 on Build GRCh38
rs554489775 Chr.5: 69262406-69262406 on Build GRCh38
rs868467163 Chr.5: 69262467-69262467 on Build GRCh38
rs70992912 Chr.5: 69262683-69262684 on Build GRCh38
rs34449359 Chr.5: 69262455-69262455 on Build GRCh38
rs548109846 Chr.5: 69262718-69262718 on Build GRCh38
rs542507570 Chr.5: 69262318-69262318 on Build GRCh38
rs968262224 Chr.5: 69262435-69262435 on Build GRCh38
rs949229911 Chr.5: 69262712-69262712 on Build GRCh38
rs35645434 Chr.5: 69262668-69262669 on Build GRCh38
rs750476115 Chr.5: 69262254-69262254 on Build GRCh38
rs779584978 Chr.5: 69262324-69262324 on Build GRCh38
rs10940209 Chr.5: 69262723-69262723 on Build GRCh38
rs1016302682 Chr.5: 69262404-69262404 on Build GRCh38
rs772048595 Chr.5: 69262326-69262326 on Build GRCh38
rs766676464 Chr.5: 69262229-69262230 on Build GRCh38
rs767340439 Chr.5: 69262334-69262336 on Build GRCh38
rs758216275 Chr.5: 69262336-69262338 on Build GRCh38
rs1004753998 Chr.5: 69262319-69262319 on Build GRCh38
rs748197935 Chr.5: 69262325-69262325 on Build GRCh38
rs756786767 Chr.5: 69262277-69262277 on Build GRCh38
rs865877801 Chr.5: 69262540-69262540 on Build GRCh38
rs529950542 Chr.5: 69262630-69262630 on Build GRCh38
rs759307878 Chr.5: 69262241-69262241 on Build GRCh38
rs780862489 Chr.5: 69262284-69262284 on Build GRCh38
rs978788858 Chr.5: 69262658-69262658 on Build GRCh38
rs887661329 Chr.5: 69262279-69262279 on Build GRCh38
rs954897009 Chr.5: 69262519-69262519 on Build GRCh38
rs375865945 Chr.5: 69262272-69262272 on Build GRCh38
rs564643497 Chr.5: 69262537-69262537 on Build GRCh38
rs769334476 Chr.5: 69262321-69262321 on Build GRCh38
rs773337070 Chr.5: 69262229-69262229 on Build GRCh38
rs756484013 Chr.5: 69262308-69262308 on Build GRCh38
rs954203450 Chr.5: 69262397-69262397 on Build GRCh38
rs758398367 Chr.5: 69262263-69262263 on Build GRCh38
rs913803176 Chr.5: 69262706-69262706 on Build GRCh38
rs961507998 Chr.5: 69262342-69262342 on Build GRCh38
rs915500278 Chr.5: 69262423-69262423 on Build GRCh38
rs543800599 Chr.5: 69262549-69262549 on Build GRCh38
rs763566784 Chr.5: 69262264-69262264 on Build GRCh38
rs750053473 Chr.5: 69262297-69262297 on Build GRCh38
rs1001894118 Chr.5: 69262402-69262402 on Build GRCh38
rs561716670 Chr.5: 69262593-69262593 on Build GRCh38
rs916295682 Chr.5: 69262558-69262558 on Build GRCh38
rs773425713 Chr.5: 69262334-69262334 on Build GRCh38
rs947711819 Chr.5: 69262561-69262561 on Build GRCh38
rs1036228414 Chr.5: 69262701-69262701 on Build GRCh38
rs752619178 Chr.5: 69262259-69262259 on Build GRCh38

View Full Product Details