Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132273081-132273585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDLIM4
Entrez Gene ID
8572
Gene Name
PDZ and LIM domain 4
Gene Aliases
RIL
UniGene
Hs.424312
Gene Chromosome Location
Chr.5: 132257658-132273454 on Build GRCh38
Transcripts
NM_001131027.1 NM_003687.3 XM_017010002.1

SNP Detail

SNP ID Chromosome Location
rs921675494 Chr.5: 132273318-132273319 on Build GRCh38
rs987411563 Chr.5: 132273405-132273405 on Build GRCh38
rs559281630 Chr.5: 132273433-132273433 on Build GRCh38
rs924760416 Chr.5: 132273148-132273148 on Build GRCh38
rs1040017114 Chr.5: 132273436-132273436 on Build GRCh38
rs1037451592 Chr.5: 132273397-132273397 on Build GRCh38
rs56992121 Chr.5: 132273445-132273445 on Build GRCh38
rs1035367693 Chr.5: 132273283-132273283 on Build GRCh38
rs1038784910 Chr.5: 132273309-132273309 on Build GRCh38
rs934645842 Chr.5: 132273150-132273150 on Build GRCh38
rs934760334 Chr.5: 132273396-132273396 on Build GRCh38
rs891016158 Chr.5: 132273186-132273186 on Build GRCh38
rs994613945 Chr.5: 132273133-132273133 on Build GRCh38
rs978911952 Chr.5: 132273239-132273239 on Build GRCh38
rs747443321 Chr.5: 132273140-132273140 on Build GRCh38
rs558026652 Chr.5: 132273444-132273444 on Build GRCh38
rs148781605 Chr.5: 132273156-132273157 on Build GRCh38
rs924678949 Chr.5: 132273382-132273382 on Build GRCh38
rs969290640 Chr.5: 132273355-132273355 on Build GRCh38
rs1039497989 Chr.5: 132273197-132273197 on Build GRCh38
rs778303930 Chr.5: 132273127-132273127 on Build GRCh38
rs58313718 Chr.5: 132273360-132273361 on Build GRCh38
rs1003867703 Chr.5: 132273277-132273277 on Build GRCh38
rs145772672 Chr.5: 132273486-132273486 on Build GRCh38
rs115707040 Chr.5: 132273192-132273192 on Build GRCh38
rs578199041 Chr.5: 132273177-132273177 on Build GRCh38
rs34545285 Chr.5: 132273507-132273508 on Build GRCh38
rs951185423 Chr.5: 132273266-132273266 on Build GRCh38
rs899789027 Chr.5: 132273441-132273441 on Build GRCh38
rs139455971 Chr.5: 132273358-132273359 on Build GRCh38
rs930512325 Chr.5: 132273327-132273328 on Build GRCh38
rs886830460 Chr.5: 132273501-132273501 on Build GRCh38
rs1027146291 Chr.5: 132273234-132273234 on Build GRCh38
rs558691603 Chr.5: 132273126-132273126 on Build GRCh38
rs1048998661 Chr.5: 132273149-132273149 on Build GRCh38
rs162881 Chr.5: 132273101-132273101 on Build GRCh38
rs991748840 Chr.5: 132273332-132273332 on Build GRCh38
rs141172553 Chr.5: 132273356-132273357 on Build GRCh38
rs1004770922 Chr.5: 132273389-132273389 on Build GRCh38
rs995384289 Chr.5: 132273200-132273200 on Build GRCh38
rs926224668 Chr.5: 132273251-132273251 on Build GRCh38
rs944473973 Chr.5: 132273287-132273287 on Build GRCh38
rs542653079 Chr.5: 132273344-132273344 on Build GRCh38
rs746161982 Chr.5: 132273198-132273198 on Build GRCh38
rs757407560 Chr.5: 132273167-132273167 on Build GRCh38
rs944430938 Chr.5: 132273113-132273113 on Build GRCh38
rs911897532 Chr.5: 132273418-132273418 on Build GRCh38
rs1014310982 Chr.5: 132273194-132273194 on Build GRCh38
rs970237160 Chr.5: 132273201-132273201 on Build GRCh38
rs960255462 Chr.5: 132273301-132273301 on Build GRCh38
rs978977270 Chr.5: 132273356-132273356 on Build GRCh38
rs140648821 Chr.5: 132273245-132273245 on Build GRCh38
rs956311281 Chr.5: 132273276-132273276 on Build GRCh38
rs544011424 Chr.5: 132273190-132273190 on Build GRCh38

View Full Product Details