Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141179700-141180192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB8
Entrez Gene ID
56128
Gene Name
protocadherin beta 8
Gene Aliases
PCDH-BETA8, PCDH3I
UniGene
Hs.283803
Gene Chromosome Location
Chr.5: 141177790-141180500 on Build GRCh38
Transcripts
NM_019120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232794 c.2058T>G p.S686S Chr.5: 141180092-141180092 on Build GRCh38
7117784 c.1893G>A p.L631L Chr.5: 141179927-141179927 on Build GRCh38
1062699 c.1896C>T p.S632S Chr.5: 141179930-141179930 on Build GRCh38
3919035 c.1721C>T p.T574I Chr.5: 141179755-141179755 on Build GRCh38
6309145 c.1714C>G p.P572A Chr.5: 141179748-141179748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782697594 Chr.5: 141179959-141179959 on Build GRCh38
rs782024596 Chr.5: 141179787-141179787 on Build GRCh38
rs782067091 Chr.5: 141179783-141179783 on Build GRCh38
rs782043465 Chr.5: 141179883-141179883 on Build GRCh38
rs138527738 Chr.5: 141180140-141180140 on Build GRCh38
rs781988348 Chr.5: 141180007-141180007 on Build GRCh38
rs782287941 Chr.5: 141180076-141180076 on Build GRCh38
rs782612510 Chr.5: 141179815-141179815 on Build GRCh38
rs200922876 Chr.5: 141180041-141180041 on Build GRCh38
rs782304914 Chr.5: 141180108-141180108 on Build GRCh38
rs781802748 Chr.5: 141179747-141179747 on Build GRCh38
rs782352967 Chr.5: 141179836-141179836 on Build GRCh38
rs373937766 Chr.5: 141180146-141180146 on Build GRCh38
rs139550075 Chr.5: 141180047-141180047 on Build GRCh38
rs17844502 Chr.5: 141179942-141179942 on Build GRCh38
rs781996928 Chr.5: 141179834-141179834 on Build GRCh38
rs782623210 Chr.5: 141179973-141179973 on Build GRCh38
rs781919932 Chr.5: 141179777-141179777 on Build GRCh38
rs370085881 Chr.5: 141179738-141179738 on Build GRCh38
rs34866056 Chr.5: 141179912-141179912 on Build GRCh38
rs782358695 Chr.5: 141179998-141179998 on Build GRCh38
rs782552370 Chr.5: 141179868-141179868 on Build GRCh38
rs782208929 Chr.5: 141179996-141179996 on Build GRCh38
rs142690275 Chr.5: 141179933-141179933 on Build GRCh38
rs369925528 Chr.5: 141180085-141180085 on Build GRCh38
rs782547439 Chr.5: 141179721-141179721 on Build GRCh38
rs782203531 Chr.5: 141179835-141179835 on Build GRCh38
rs147107612 Chr.5: 141179770-141179770 on Build GRCh38
rs781836739 Chr.5: 141180078-141180078 on Build GRCh38
rs782098169 Chr.5: 141179745-141179745 on Build GRCh38
rs782092870 Chr.5: 141180107-141180107 on Build GRCh38
rs17844504 Chr.5: 141179991-141179991 on Build GRCh38
rs147123510 Chr.5: 141180125-141180125 on Build GRCh38
rs2740582 Chr.5: 141179951-141179951 on Build GRCh38
rs782628632 Chr.5: 141179873-141179873 on Build GRCh38
rs781919824 Chr.5: 141180002-141180002 on Build GRCh38
rs782724408 Chr.5: 141179866-141179866 on Build GRCh38
rs571141703 Chr.5: 141179989-141179989 on Build GRCh38
rs782650929 Chr.5: 141180075-141180075 on Build GRCh38
rs782736535 Chr.5: 141180076-141180076 on Build GRCh38
rs782389783 Chr.5: 141179952-141179952 on Build GRCh38
rs781983619 Chr.5: 141180071-141180071 on Build GRCh38
rs782423761 Chr.5: 141180097-141180097 on Build GRCh38
rs782190775 Chr.5: 141180148-141180148 on Build GRCh38
rs782295910 Chr.5: 141179821-141179821 on Build GRCh38
rs782585940 Chr.5: 141180092-141180092 on Build GRCh38
rs781797785 Chr.5: 141180122-141180122 on Build GRCh38
rs375739229 Chr.5: 141180164-141180164 on Build GRCh38
rs781858287 Chr.5: 141179915-141179915 on Build GRCh38
rs782490850 Chr.5: 141180131-141180131 on Build GRCh38
rs782028134 Chr.5: 141180150-141180150 on Build GRCh38
rs778535463 Chr.5: 141180119-141180119 on Build GRCh38
rs145707096 Chr.5: 141179734-141179734 on Build GRCh38
rs782249589 Chr.5: 141180052-141180052 on Build GRCh38
rs373119743 Chr.5: 141180086-141180086 on Build GRCh38
rs267600430 Chr.5: 141180161-141180161 on Build GRCh38
rs374710779 Chr.5: 141179756-141179756 on Build GRCh38
rs782396899 Chr.5: 141179945-141179945 on Build GRCh38
rs781889748 Chr.5: 141179962-141179962 on Build GRCh38
rs782462443 Chr.5: 141179751-141179751 on Build GRCh38
rs112099599 Chr.5: 141180144-141180144 on Build GRCh38
rs782085543 Chr.5: 141180117-141180117 on Build GRCh38
rs563679514 Chr.5: 141179867-141179867 on Build GRCh38
rs777458231 Chr.5: 141180067-141180067 on Build GRCh38
rs17844501 Chr.5: 141179885-141179885 on Build GRCh38
rs17844507 Chr.5: 141180098-141180098 on Build GRCh38
rs370441568 Chr.5: 141180128-141180128 on Build GRCh38
rs782154872 Chr.5: 141180065-141180065 on Build GRCh38
rs782781685 Chr.5: 141180171-141180171 on Build GRCh38
rs782097283 Chr.5: 141179906-141179906 on Build GRCh38
rs781945778 Chr.5: 141180064-141180064 on Build GRCh38
rs782423303 Chr.5: 141180006-141180006 on Build GRCh38
rs782634308 Chr.5: 141179992-141179992 on Build GRCh38
rs201726568 Chr.5: 141179762-141179762 on Build GRCh38
rs782164666 Chr.5: 141180166-141180166 on Build GRCh38
rs781938170 Chr.5: 141180114-141180114 on Build GRCh38
rs782463343 Chr.5: 141180035-141180035 on Build GRCh38
rs781948265 Chr.5: 141180101-141180101 on Build GRCh38
rs2697541 Chr.5: 141180015-141180015 on Build GRCh38
rs782107526 Chr.5: 141179788-141179788 on Build GRCh38
rs150243744 Chr.5: 141179753-141179753 on Build GRCh38
rs782719532 Chr.5: 141179803-141179803 on Build GRCh38
rs757018919 Chr.5: 141180105-141180105 on Build GRCh38
rs150564946 Chr.5: 141179986-141179986 on Build GRCh38
rs782362169 Chr.5: 141179826-141179826 on Build GRCh38
rs782178135 Chr.5: 141179816-141179816 on Build GRCh38
rs782256171 Chr.5: 141179884-141179884 on Build GRCh38
rs782198861 Chr.5: 141180095-141180095 on Build GRCh38
rs782285661 Chr.5: 141179990-141179990 on Build GRCh38
rs781894460 Chr.5: 141179805-141179805 on Build GRCh38
rs537745447 Chr.5: 141179969-141179969 on Build GRCh38
rs782583919 Chr.5: 141179728-141179728 on Build GRCh38
rs782724794 Chr.5: 141179917-141179917 on Build GRCh38
rs545745677 Chr.5: 141180165-141180165 on Build GRCh38
rs548331717 Chr.5: 141179936-141179936 on Build GRCh38
rs781864846 Chr.5: 141180033-141180033 on Build GRCh38
rs377688984 Chr.5: 141179944-141179944 on Build GRCh38
rs377233474 Chr.5: 141180110-141180110 on Build GRCh38
rs781815915 Chr.5: 141179791-141179791 on Build GRCh38
rs782391266 Chr.5: 141180056-141180056 on Build GRCh38
rs543400913 Chr.5: 141179846-141179846 on Build GRCh38
rs782719463 Chr.5: 141180016-141180016 on Build GRCh38
rs782428679 Chr.5: 141179931-141179931 on Build GRCh38
rs781842190 Chr.5: 141179864-141179864 on Build GRCh38
rs782322523 Chr.5: 141179871-141179871 on Build GRCh38
rs782654812 Chr.5: 141180087-141180087 on Build GRCh38
rs782177986 Chr.5: 141179804-141179805 on Build GRCh38
rs142723933 Chr.5: 141180162-141180162 on Build GRCh38
rs782129569 Chr.5: 141179853-141179853 on Build GRCh38
rs138970173 Chr.5: 141179761-141179761 on Build GRCh38
rs17844506 Chr.5: 141180074-141180074 on Build GRCh38
rs781878337 Chr.5: 141180115-141180115 on Build GRCh38
rs782255854 Chr.5: 141179877-141179877 on Build GRCh38
rs782483601 Chr.5: 141179814-141179814 on Build GRCh38
rs782064898 Chr.5: 141180003-141180003 on Build GRCh38
rs781892339 Chr.5: 141180017-141180017 on Build GRCh38
rs199819904 Chr.5: 141179810-141179810 on Build GRCh38
rs755868664 Chr.5: 141180029-141180029 on Build GRCh38
rs782667532 Chr.5: 141179819-141179819 on Build GRCh38
rs563401373 Chr.5: 141180157-141180157 on Build GRCh38
rs781942957 Chr.5: 141179837-141179837 on Build GRCh38
rs374505292 Chr.5: 141179773-141179773 on Build GRCh38
rs781799409 Chr.5: 141179985-141179985 on Build GRCh38
rs572196294 Chr.5: 141179766-141179766 on Build GRCh38
rs781894851 Chr.5: 141179966-141179966 on Build GRCh38
rs782764386 Chr.5: 141179789-141179789 on Build GRCh38
rs782808193 Chr.5: 141180069-141180069 on Build GRCh38
rs141057693 Chr.5: 141179746-141179746 on Build GRCh38
rs782163367 Chr.5: 141179963-141179963 on Build GRCh38
rs781872796 Chr.5: 141179967-141179967 on Build GRCh38
rs782363146 Chr.5: 141179776-141179776 on Build GRCh38
rs782215807 Chr.5: 141180088-141180088 on Build GRCh38
rs782259875 Chr.5: 141179727-141179727 on Build GRCh38
rs782596438 Chr.5: 141179763-141179763 on Build GRCh38
rs146928691 Chr.5: 141180172-141180172 on Build GRCh38
rs267600429 Chr.5: 141179882-141179882 on Build GRCh38
rs61735944 Chr.5: 141180014-141180014 on Build GRCh38
rs782215919 Chr.5: 141179774-141179774 on Build GRCh38
rs782627939 Chr.5: 141179724-141179724 on Build GRCh38
rs35928232 Chr.5: 141179879-141179879 on Build GRCh38
rs149161573 Chr.5: 141180079-141180079 on Build GRCh38
rs782710772 Chr.5: 141179908-141179908 on Build GRCh38
rs782180053 Chr.5: 141180046-141180046 on Build GRCh38
rs782519470 Chr.5: 141179934-141179934 on Build GRCh38
rs782662625 Chr.5: 141180126-141180126 on Build GRCh38
rs782435311 Chr.5: 141179865-141179865 on Build GRCh38
rs17844508 Chr.5: 141180138-141180138 on Build GRCh38
rs782386871 Chr.5: 141180100-141180100 on Build GRCh38
rs17844498 Chr.5: 141179739-141179739 on Build GRCh38
rs781833997 Chr.5: 141179927-141179927 on Build GRCh38
rs781953674 Chr.5: 141179741-141179741 on Build GRCh38
rs141755837 Chr.5: 141179955-141179955 on Build GRCh38
rs144189906 Chr.5: 141180057-141180057 on Build GRCh38
rs782133409 Chr.5: 141180155-141180155 on Build GRCh38
rs782748024 Chr.5: 141180030-141180030 on Build GRCh38
rs112759368 Chr.5: 141179903-141179903 on Build GRCh38
rs782818286 Chr.5: 141179845-141179845 on Build GRCh38
rs753615504 Chr.5: 141180073-141180073 on Build GRCh38
rs201448607 Chr.5: 141179971-141179971 on Build GRCh38
rs782704020 Chr.5: 141180080-141180080 on Build GRCh38
rs201243979 Chr.5: 141179797-141179797 on Build GRCh38
rs782723370 Chr.5: 141179863-141179863 on Build GRCh38
rs782027000 Chr.5: 141179946-141179946 on Build GRCh38
rs782480554 Chr.5: 141180091-141180091 on Build GRCh38
rs782549414 Chr.5: 141180020-141180020 on Build GRCh38
rs782664391 Chr.5: 141179768-141179768 on Build GRCh38
rs782747311 Chr.5: 141180120-141180120 on Build GRCh38
rs782098562 Chr.5: 141179957-141179957 on Build GRCh38
rs565807496 Chr.5: 141179910-141179910 on Build GRCh38
rs782155242 Chr.5: 141179840-141179840 on Build GRCh38
rs782311995 Chr.5: 141180061-141180061 on Build GRCh38

View Full Product Details