Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150219520-150220018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32E
Entrez Gene ID
81611
Gene Name
acidic nuclear phosphoprotein 32 family member E
Gene Aliases
LANP-L, LANPL
UniGene
Hs.656466
Gene Chromosome Location
Chr.1: 150218417-150236156 on Build GRCh38
Transcripts
NM_001136478.3 NM_001136479.2 NM_001280559.1 NM_001280560.1 NM_030920.4 XM_005245513.3 XM_005245514.3 XM_017002418.1

SNP Detail

SNP ID Chromosome Location
rs782472308 Chr.1: 150219639-150219639 on Build GRCh38
rs937925455 Chr.1: 150219962-150219962 on Build GRCh38
rs111859228 Chr.1: 150219577-150219577 on Build GRCh38
rs587672294 Chr.1: 150219973-150219973 on Build GRCh38
rs1035092222 Chr.1: 150219572-150219572 on Build GRCh38
rs979028516 Chr.1: 150219642-150219642 on Build GRCh38
rs905153945 Chr.1: 150219922-150219922 on Build GRCh38
rs1009693531 Chr.1: 150219605-150219605 on Build GRCh38
rs147121925 Chr.1: 150219812-150219812 on Build GRCh38
rs587631758 Chr.1: 150219821-150219821 on Build GRCh38
rs587608801 Chr.1: 150219923-150219923 on Build GRCh38
rs587642495 Chr.1: 150219672-150219672 on Build GRCh38
rs587768263 Chr.1: 150219871-150219871 on Build GRCh38
rs967510179 Chr.1: 150219640-150219640 on Build GRCh38
rs941172216 Chr.1: 150219859-150219859 on Build GRCh38
rs587686946 Chr.1: 150219841-150219841 on Build GRCh38
rs587669830 Chr.1: 150219784-150219784 on Build GRCh38
rs782251222 Chr.1: 150219971-150219971 on Build GRCh38
rs975472066 Chr.1: 150219691-150219691 on Build GRCh38
rs80110309 Chr.1: 150219660-150219660 on Build GRCh38
rs186412915 Chr.1: 150219730-150219730 on Build GRCh38
rs782669280 Chr.1: 150219885-150219885 on Build GRCh38
rs964275576 Chr.1: 150219661-150219661 on Build GRCh38
rs781839052 Chr.1: 150219624-150219624 on Build GRCh38

View Full Product Details