Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18224420-18224966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEK
Entrez Gene ID
7913
Gene Name
DEK proto-oncogene
Gene Aliases
D6S231E
UniGene
Hs.484813
Gene Chromosome Location
Chr.6: 18224169-18264568 on Build GRCh38
Transcripts
NM_001134709.1 NM_003472.3

SNP Detail

SNP ID Chromosome Location
rs529223116 Chr.6: 18224699-18224699 on Build GRCh38
rs1022929691 Chr.6: 18224731-18224731 on Build GRCh38
rs890529585 Chr.6: 18224681-18224681 on Build GRCh38
rs923528869 Chr.6: 18224619-18224619 on Build GRCh38
rs752044843 Chr.6: 18224463-18224464 on Build GRCh38
rs985265021 Chr.6: 18224560-18224560 on Build GRCh38
rs563640642 Chr.6: 18224801-18224801 on Build GRCh38
rs964973766 Chr.6: 18224464-18224464 on Build GRCh38
rs910751189 Chr.6: 18224563-18224564 on Build GRCh38
rs111415771 Chr.6: 18224487-18224487 on Build GRCh38
rs940982040 Chr.6: 18224803-18224803 on Build GRCh38
rs978242814 Chr.6: 18224750-18224750 on Build GRCh38
rs760811824 Chr.6: 18224612-18224612 on Build GRCh38
rs907150353 Chr.6: 18224873-18224873 on Build GRCh38
rs897070439 Chr.6: 18224624-18224624 on Build GRCh38
rs890999876 Chr.6: 18224916-18224916 on Build GRCh38
rs766585662 Chr.6: 18224691-18224691 on Build GRCh38
rs990601662 Chr.6: 18224789-18224789 on Build GRCh38
rs773541899 Chr.6: 18224543-18224543 on Build GRCh38
rs1026343837 Chr.6: 18224889-18224889 on Build GRCh38
rs1050868004 Chr.6: 18224908-18224908 on Build GRCh38
rs1038055946 Chr.6: 18224822-18224822 on Build GRCh38
rs1013166489 Chr.6: 18224820-18224820 on Build GRCh38
rs149949880 Chr.6: 18224773-18224773 on Build GRCh38
rs1029466226 Chr.6: 18224673-18224673 on Build GRCh38
rs1024107960 Chr.6: 18224872-18224872 on Build GRCh38
rs915405725 Chr.6: 18224495-18224495 on Build GRCh38
rs140400691 Chr.6: 18224568-18224568 on Build GRCh38
rs868740330 Chr.6: 18224513-18224513 on Build GRCh38
rs3476 Chr.6: 18224942-18224942 on Build GRCh38
rs1041399583 Chr.6: 18224659-18224659 on Build GRCh38
rs1038658494 Chr.6: 18224621-18224621 on Build GRCh38
rs1017558015 Chr.6: 18224712-18224712 on Build GRCh38
rs1054776316 Chr.6: 18224567-18224567 on Build GRCh38
rs965108712 Chr.6: 18224719-18224719 on Build GRCh38
rs542161056 Chr.6: 18224536-18224542 on Build GRCh38
rs752512964 Chr.6: 18224765-18224765 on Build GRCh38
rs575609291 Chr.6: 18224535-18224535 on Build GRCh38
rs989505412 Chr.6: 18224729-18224729 on Build GRCh38
rs764061142 Chr.6: 18224818-18224818 on Build GRCh38
rs941278080 Chr.6: 18224601-18224601 on Build GRCh38
rs146494116 Chr.6: 18224886-18224886 on Build GRCh38
rs1008962108 Chr.6: 18224695-18224695 on Build GRCh38
rs1018124759 Chr.6: 18224929-18224929 on Build GRCh38
rs929926427 Chr.6: 18224895-18224895 on Build GRCh38
rs754680402 Chr.6: 18224674-18224675 on Build GRCh38
rs966999546 Chr.6: 18224744-18224744 on Build GRCh38
rs984780624 Chr.6: 18224907-18224907 on Build GRCh38
rs374662564 Chr.6: 18224626-18224626 on Build GRCh38
rs112325094 Chr.6: 18224596-18224596 on Build GRCh38
rs545875456 Chr.6: 18224580-18224580 on Build GRCh38
rs993940188 Chr.6: 18224661-18224661 on Build GRCh38
rs927669611 Chr.6: 18224541-18224541 on Build GRCh38
rs902829200 Chr.6: 18224665-18224665 on Build GRCh38
rs976230161 Chr.6: 18224943-18224943 on Build GRCh38
rs945503001 Chr.6: 18224531-18224531 on Build GRCh38
rs952205233 Chr.6: 18224559-18224559 on Build GRCh38
rs541819408 Chr.6: 18224475-18224475 on Build GRCh38
rs993414335 Chr.6: 18224884-18224884 on Build GRCh38
rs939079594 Chr.6: 18224782-18224782 on Build GRCh38
rs945598690 Chr.6: 18224637-18224637 on Build GRCh38
rs542914199 Chr.6: 18224867-18224867 on Build GRCh38
rs1024245263 Chr.6: 18224490-18224490 on Build GRCh38
rs943548203 Chr.6: 18224585-18224585 on Build GRCh38
rs939110843 Chr.6: 18224548-18224548 on Build GRCh38

View Full Product Details